Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 64010414-64010944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMD6
Entrez Gene ID
9861
Gene Name
proteasome 26S subunit, non-ATPase 6
Gene Aliases
Rpn7, S10, SGA-113M, p42A, p44S10
UniGene
Hs.152536
Gene Chromosome Location
Chr.3: 64010549-64024010 on Build GRCh38
Transcripts
NM_001271779.1 NM_001271780.1 NM_001271781.1 NM_014814.2 XM_005265619.1 XM_011534288.1 XM_017007569.1 NR_038286.1

Gene Symbol
PSMD6-AS2
Entrez Gene ID
100507062
Gene Name
PSMD6 antisense RNA 2
Gene Aliases
-
UniGene
Hs.152536
Gene Chromosome Location
Chr.3: 64004022-64012243 on Build GRCh38
Transcripts
NM_001271779.1 NM_001271780.1 NM_001271781.1 NM_014814.2 XM_005265619.1 XM_011534288.1 XM_017007569.1 NR_038286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4630893 c.947T>A p.V316E Chr.3: 64010890-64010890 on Build GRCh38
1753361 c.1078G>A p.D360N Chr.3: 64010760-64010760 on Build GRCh38
1131363 c.1105A>C p.T369P Chr.3: 64010733-64010733 on Build GRCh38
4630892 c.1061T>A p.V354E Chr.3: 64010890-64010890 on Build GRCh38
5798767 c.1146A>T p.K382N Chr.3: 64010692-64010692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373997221 Chr.3: 64010901-64010901 on Build GRCh38
rs773513107 Chr.3: 64010779-64010779 on Build GRCh38
rs200913854 Chr.3: 64010870-64010870 on Build GRCh38
rs190515045 Chr.3: 64010458-64010458 on Build GRCh38
rs771038707 Chr.3: 64010646-64010646 on Build GRCh38
rs776717867 Chr.3: 64010798-64010798 on Build GRCh38
rs571187628 Chr.3: 64010554-64010554 on Build GRCh38
rs748663278 Chr.3: 64010700-64010700 on Build GRCh38
rs747862951 Chr.3: 64010921-64010921 on Build GRCh38
rs879131810 Chr.3: 64010598-64010598 on Build GRCh38
rs929991205 Chr.3: 64010448-64010448 on Build GRCh38
rs975051900 Chr.3: 64010831-64010831 on Build GRCh38
rs572335618 Chr.3: 64010816-64010816 on Build GRCh38
rs747030579 Chr.3: 64010795-64010795 on Build GRCh38
rs754698499 Chr.3: 64010763-64010763 on Build GRCh38
rs76056570 Chr.3: 64010470-64010470 on Build GRCh38
rs754610524 Chr.3: 64010858-64010858 on Build GRCh38
rs778394033 Chr.3: 64010787-64010787 on Build GRCh38
rs748550060 Chr.3: 64010772-64010772 on Build GRCh38
rs202088427 Chr.3: 64010584-64010585 on Build GRCh38
rs1046025 Chr.3: 64010586-64010586 on Build GRCh38
rs534470073 Chr.3: 64010527-64010527 on Build GRCh38
rs763830203 Chr.3: 64010663-64010663 on Build GRCh38
rs773162770 Chr.3: 64010632-64010632 on Build GRCh38
rs564037062 Chr.3: 64010853-64010853 on Build GRCh38
rs769787602 Chr.3: 64010781-64010782 on Build GRCh38
rs754508395 Chr.3: 64010770-64010772 on Build GRCh38
rs758819604 Chr.3: 64010786-64010786 on Build GRCh38
rs911903522 Chr.3: 64010460-64010460 on Build GRCh38
rs867330284 Chr.3: 64010824-64010824 on Build GRCh38
rs758868081 Chr.3: 64010673-64010674 on Build GRCh38
rs181913793 Chr.3: 64010484-64010484 on Build GRCh38
rs199911104 Chr.3: 64010650-64010650 on Build GRCh38
rs761721292 Chr.3: 64010836-64010836 on Build GRCh38
rs530652687 Chr.3: 64010843-64010843 on Build GRCh38
rs556808057 Chr.3: 64010608-64010608 on Build GRCh38
rs574942701 Chr.3: 64010614-64010614 on Build GRCh38
rs185008244 Chr.3: 64010494-64010494 on Build GRCh38
rs373282641 Chr.3: 64010814-64010814 on Build GRCh38
rs375750857 Chr.3: 64010771-64010771 on Build GRCh38
rs755696075 Chr.3: 64010748-64010748 on Build GRCh38
rs946581280 Chr.3: 64010530-64010530 on Build GRCh38
rs749315951 Chr.3: 64010503-64010503 on Build GRCh38
rs772645567 Chr.3: 64010618-64010618 on Build GRCh38
rs766095729 Chr.3: 64010756-64010756 on Build GRCh38
rs371594487 Chr.3: 64010608-64010609 on Build GRCh38
rs779561861 Chr.3: 64010765-64010766 on Build GRCh38
rs372288372 Chr.3: 64010695-64010695 on Build GRCh38
rs1007693143 Chr.3: 64010634-64010634 on Build GRCh38
rs752982040 Chr.3: 64010863-64010863 on Build GRCh38
rs571575391 Chr.3: 64010606-64010607 on Build GRCh38
rs749021246 Chr.3: 64010808-64010809 on Build GRCh38
rs753412173 Chr.3: 64010856-64010856 on Build GRCh38
rs1019339403 Chr.3: 64010805-64010805 on Build GRCh38
rs1056495671 Chr.3: 64010587-64010587 on Build GRCh38
rs147494745 Chr.3: 64010713-64010713 on Build GRCh38
rs950691937 Chr.3: 64010443-64010443 on Build GRCh38
rs750691116 Chr.3: 64010653-64010656 on Build GRCh38
rs982784557 Chr.3: 64010803-64010803 on Build GRCh38
rs759788983 Chr.3: 64010651-64010651 on Build GRCh38
rs896171848 Chr.3: 64010591-64010591 on Build GRCh38
rs372965754 Chr.3: 64010833-64010834 on Build GRCh38
rs559276166 Chr.3: 64010549-64010549 on Build GRCh38
rs915997558 Chr.3: 64010894-64010894 on Build GRCh38
rs760341335 Chr.3: 64010898-64010898 on Build GRCh38
rs966084263 Chr.3: 64010822-64010822 on Build GRCh38
rs938788211 Chr.3: 64010489-64010489 on Build GRCh38
rs77843045 Chr.3: 64010916-64010916 on Build GRCh38
rs894616508 Chr.3: 64010536-64010537 on Build GRCh38
rs751276017 Chr.3: 64010767-64010768 on Build GRCh38
rs376618605 Chr.3: 64010851-64010851 on Build GRCh38
rs760695968 Chr.3: 64010874-64010874 on Build GRCh38
rs369229462 Chr.3: 64010838-64010838 on Build GRCh38
rs747674753 Chr.3: 64010772-64010773 on Build GRCh38
rs979131456 Chr.3: 64010545-64010545 on Build GRCh38
rs369084169 Chr.3: 64010797-64010797 on Build GRCh38
rs543173006 Chr.3: 64010886-64010886 on Build GRCh38
rs577922025 Chr.3: 64010562-64010562 on Build GRCh38
rs376072140 Chr.3: 64010586-64010586 on Build GRCh38
rs546208245 Chr.3: 64010826-64010826 on Build GRCh38
rs554103023 Chr.3: 64010716-64010716 on Build GRCh38
rs761592937 Chr.3: 64010704-64010704 on Build GRCh38
rs760867687 Chr.3: 64010643-64010643 on Build GRCh38
rs113068109 Chr.3: 64010767-64010767 on Build GRCh38
rs777832768 Chr.3: 64010784-64010785 on Build GRCh38
rs747503832 Chr.3: 64010871-64010871 on Build GRCh38
rs760559278 Chr.3: 64010852-64010852 on Build GRCh38
rs3217453 Chr.3: 64010770-64010771 on Build GRCh38
rs145261156 Chr.3: 64010740-64010740 on Build GRCh38

View Full Product Details