Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68004132-68004644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM19A1
Entrez Gene ID
407738
Gene Name
family with sequence similarity 19 member A1, C-C motif chemokine like
Gene Aliases
TAFA-1, TAFA1
UniGene
Hs.655061
Gene Chromosome Location
Chr.3: 67991592-68546056 on Build GRCh38
Transcripts
NM_001252216.1 NM_213609.3 XM_017006387.1 XM_017006388.1 XM_017006390.1

SNP Detail

SNP ID Chromosome Location
rs772965009 Chr.3: 68004488-68004488 on Build GRCh38
rs528690406 Chr.3: 68004455-68004455 on Build GRCh38
rs989139152 Chr.3: 68004504-68004504 on Build GRCh38
rs553010239 Chr.3: 68004194-68004194 on Build GRCh38
rs949695889 Chr.3: 68004406-68004406 on Build GRCh38
rs188011743 Chr.3: 68004249-68004249 on Build GRCh38
rs954818235 Chr.3: 68004198-68004198 on Build GRCh38
rs200797756 Chr.3: 68004558-68004558 on Build GRCh38
rs935005640 Chr.3: 68004270-68004270 on Build GRCh38
rs770670070 Chr.3: 68004496-68004496 on Build GRCh38
rs572973181 Chr.3: 68004229-68004229 on Build GRCh38
rs759410994 Chr.3: 68004592-68004592 on Build GRCh38
rs199502322 Chr.3: 68004557-68004557 on Build GRCh38
rs1054357610 Chr.3: 68004465-68004465 on Build GRCh38
rs752932805 Chr.3: 68004508-68004508 on Build GRCh38
rs551277729 Chr.3: 68004506-68004506 on Build GRCh38
rs1004094592 Chr.3: 68004341-68004341 on Build GRCh38
rs939545461 Chr.3: 68004563-68004563 on Build GRCh38
rs774132853 Chr.3: 68004523-68004523 on Build GRCh38
rs562250511 Chr.3: 68004441-68004441 on Build GRCh38
rs1047320385 Chr.3: 68004356-68004356 on Build GRCh38
rs752166375 Chr.3: 68004557-68004559 on Build GRCh38
rs190870041 Chr.3: 68004494-68004494 on Build GRCh38
rs146295415 Chr.3: 68004424-68004424 on Build GRCh38
rs901274639 Chr.3: 68004474-68004474 on Build GRCh38
rs756393667 Chr.3: 68004600-68004600 on Build GRCh38
rs756517597 Chr.3: 68004556-68004559 on Build GRCh38
rs889700938 Chr.3: 68004514-68004514 on Build GRCh38
rs1017266667 Chr.3: 68004516-68004516 on Build GRCh38
rs66481633 Chr.3: 68004556-68004558 on Build GRCh38
rs869213358 Chr.3: 68004558-68004560 on Build GRCh38
rs964160542 Chr.3: 68004427-68004427 on Build GRCh38
rs567296408 Chr.3: 68004541-68004541 on Build GRCh38
rs559765312 Chr.3: 68004440-68004440 on Build GRCh38
rs1005560266 Chr.3: 68004396-68004396 on Build GRCh38
rs368584411 Chr.3: 68004421-68004421 on Build GRCh38
rs148594850 Chr.3: 68004492-68004492 on Build GRCh38
rs1047637476 Chr.3: 68004568-68004568 on Build GRCh38
rs926999767 Chr.3: 68004559-68004559 on Build GRCh38
rs549089454 Chr.3: 68004429-68004429 on Build GRCh38
rs201393977 Chr.3: 68004556-68004556 on Build GRCh38
rs998168450 Chr.3: 68004476-68004476 on Build GRCh38
rs996922623 Chr.3: 68004437-68004437 on Build GRCh38
rs893873250 Chr.3: 68004211-68004211 on Build GRCh38
rs183239664 Chr.3: 68004588-68004588 on Build GRCh38
rs746369394 Chr.3: 68004212-68004213 on Build GRCh38
rs916995394 Chr.3: 68004374-68004374 on Build GRCh38
rs17140929 Chr.3: 68004552-68004552 on Build GRCh38
rs530762157 Chr.3: 68004306-68004306 on Build GRCh38
rs544091791 Chr.3: 68004383-68004383 on Build GRCh38
rs551906250 Chr.3: 68004468-68004468 on Build GRCh38
rs1044837078 Chr.3: 68004550-68004550 on Build GRCh38
rs182493887 Chr.3: 68004248-68004248 on Build GRCh38
rs1053344875 Chr.3: 68004273-68004273 on Build GRCh38
rs780557699 Chr.3: 68004292-68004292 on Build GRCh38
rs1005506654 Chr.3: 68004515-68004516 on Build GRCh38
rs935669011 Chr.3: 68004571-68004571 on Build GRCh38
rs906681245 Chr.3: 68004324-68004324 on Build GRCh38
rs979728589 Chr.3: 68004555-68004555 on Build GRCh38
rs1045666961 Chr.3: 68004311-68004311 on Build GRCh38
rs527789867 Chr.3: 68004542-68004542 on Build GRCh38
rs141926967 Chr.3: 68004401-68004401 on Build GRCh38
rs747764543 Chr.3: 68004405-68004405 on Build GRCh38
rs887326205 Chr.3: 68004362-68004362 on Build GRCh38
rs767787927 Chr.3: 68004471-68004471 on Build GRCh38
rs915030818 Chr.3: 68004282-68004282 on Build GRCh38
rs905467906 Chr.3: 68004192-68004192 on Build GRCh38

View Full Product Details