Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69084853-69085359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6IP5
Entrez Gene ID
10550
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 5
Gene Aliases
DERP11, GTRAP3-18, HSPC127, JWA, PRAF3, Yip6b, addicsin, hp22, jmx
UniGene
Hs.518060
Gene Chromosome Location
Chr.3: 69084939-69106088 on Build GRCh38
Transcripts
NM_006407.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604846 c.157_159delATG p.M54delM Chr.3: 69085204-69085206 on Build GRCh38
1485462 c.176+1_176+2insT p.? Chr.3: 69085224-69085225 on Build GRCh38
4845808 c.15C>G p.I5M Chr.3: 69085062-69085062 on Build GRCh38
7200614 c.41T>G p.F14C Chr.3: 69085088-69085088 on Build GRCh38
7094897 c.42C>A p.F14L Chr.3: 69085089-69085089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954891572 Chr.3: 69084959-69084959 on Build GRCh38
rs774190139 Chr.3: 69085269-69085269 on Build GRCh38
rs767557247 Chr.3: 69084998-69084999 on Build GRCh38
rs751081080 Chr.3: 69085008-69085008 on Build GRCh38
rs199950458 Chr.3: 69085216-69085216 on Build GRCh38
rs774102206 Chr.3: 69085153-69085153 on Build GRCh38
rs760167852 Chr.3: 69085203-69085203 on Build GRCh38
rs771494996 Chr.3: 69085245-69085246 on Build GRCh38
rs370577404 Chr.3: 69085176-69085176 on Build GRCh38
rs111700508 Chr.3: 69085033-69085033 on Build GRCh38
rs770813056 Chr.3: 69085270-69085271 on Build GRCh38
rs761806347 Chr.3: 69085170-69085170 on Build GRCh38
rs753102364 Chr.3: 69085236-69085236 on Build GRCh38
rs762750764 Chr.3: 69085048-69085048 on Build GRCh38
rs201200564 Chr.3: 69085114-69085114 on Build GRCh38
rs749916432 Chr.3: 69085191-69085191 on Build GRCh38
rs911170237 Chr.3: 69084897-69084898 on Build GRCh38
rs779456513 Chr.3: 69085229-69085229 on Build GRCh38
rs562358092 Chr.3: 69084877-69084877 on Build GRCh38
rs976649023 Chr.3: 69085285-69085285 on Build GRCh38
rs947976176 Chr.3: 69084980-69084980 on Build GRCh38
rs779700699 Chr.3: 69085076-69085076 on Build GRCh38
rs202151518 Chr.3: 69085030-69085030 on Build GRCh38
rs753485326 Chr.3: 69085217-69085217 on Build GRCh38
rs372636665 Chr.3: 69085093-69085093 on Build GRCh38
rs768390281 Chr.3: 69085139-69085139 on Build GRCh38
rs748273789 Chr.3: 69085035-69085035 on Build GRCh38
rs557684428 Chr.3: 69085082-69085082 on Build GRCh38
rs569349303 Chr.3: 69084979-69084979 on Build GRCh38
rs1016275373 Chr.3: 69085072-69085072 on Build GRCh38
rs922240863 Chr.3: 69085298-69085298 on Build GRCh38
rs774935049 Chr.3: 69085034-69085034 on Build GRCh38
rs146642345 Chr.3: 69084971-69084971 on Build GRCh38
rs765981304 Chr.3: 69085207-69085207 on Build GRCh38
rs747667916 Chr.3: 69085219-69085221 on Build GRCh38
rs1029234297 Chr.3: 69085165-69085165 on Build GRCh38
rs781292331 Chr.3: 69085259-69085259 on Build GRCh38
rs759030081 Chr.3: 69085241-69085241 on Build GRCh38
rs780853153 Chr.3: 69085025-69085025 on Build GRCh38
rs989310986 Chr.3: 69085246-69085246 on Build GRCh38
rs761540758 Chr.3: 69085273-69085273 on Build GRCh38
rs746150381 Chr.3: 69085261-69085261 on Build GRCh38
rs775204291 Chr.3: 69085044-69085044 on Build GRCh38
rs1031559588 Chr.3: 69085293-69085293 on Build GRCh38
rs756689357 Chr.3: 69085010-69085010 on Build GRCh38
rs542239988 Chr.3: 69085324-69085324 on Build GRCh38
rs769999590 Chr.3: 69085263-69085263 on Build GRCh38
rs974949779 Chr.3: 69084923-69084923 on Build GRCh38
rs575760008 Chr.3: 69084882-69084882 on Build GRCh38
rs957918716 Chr.3: 69084884-69084884 on Build GRCh38
rs777893041 Chr.3: 69085233-69085242 on Build GRCh38
rs528001807 Chr.3: 69085075-69085075 on Build GRCh38
rs899160016 Chr.3: 69085118-69085118 on Build GRCh38
rs772673187 Chr.3: 69085102-69085102 on Build GRCh38
rs968748264 Chr.3: 69084898-69084898 on Build GRCh38
rs756070702 Chr.3: 69085230-69085230 on Build GRCh38
rs149212853 Chr.3: 69085007-69085007 on Build GRCh38
rs751545675 Chr.3: 69085151-69085151 on Build GRCh38
rs190980113 Chr.3: 69084890-69084890 on Build GRCh38
rs11542220 Chr.3: 69085137-69085137 on Build GRCh38
rs570981345 Chr.3: 69084903-69084903 on Build GRCh38
rs888595897 Chr.3: 69084995-69084996 on Build GRCh38
rs923698208 Chr.3: 69084962-69084962 on Build GRCh38
rs767055034 Chr.3: 69085053-69085053 on Build GRCh38
rs779589390 Chr.3: 69085094-69085094 on Build GRCh38
rs576545934 Chr.3: 69085279-69085279 on Build GRCh38
rs771000582 Chr.3: 69085257-69085257 on Build GRCh38
rs780762487 Chr.3: 69084996-69084998 on Build GRCh38
rs758331284 Chr.3: 69085001-69085001 on Build GRCh38
rs376728432 Chr.3: 69085221-69085221 on Build GRCh38
rs747095530 Chr.3: 69085111-69085111 on Build GRCh38
rs906410779 Chr.3: 69084991-69084991 on Build GRCh38
rs989176555 Chr.3: 69084975-69084975 on Build GRCh38
rs534878576 Chr.3: 69085067-69085067 on Build GRCh38
rs543035958 Chr.3: 69085270-69085270 on Build GRCh38
rs987678042 Chr.3: 69085275-69085275 on Build GRCh38
rs764022062 Chr.3: 69085049-69085049 on Build GRCh38
rs747063034 Chr.3: 69085247-69085247 on Build GRCh38
rs745570847 Chr.3: 69085026-69085026 on Build GRCh38
rs749621819 Chr.3: 69085077-69085077 on Build GRCh38
rs987653929 Chr.3: 69084961-69084961 on Build GRCh38
rs79392449 Chr.3: 69084883-69084883 on Build GRCh38
rs755761324 Chr.3: 69085027-69085027 on Build GRCh38
rs778281752 Chr.3: 69085244-69085244 on Build GRCh38
rs1044958983 Chr.3: 69084986-69084986 on Build GRCh38
rs749945961 Chr.3: 69085058-69085058 on Build GRCh38
rs776844556 Chr.3: 69085112-69085112 on Build GRCh38
rs766616578 Chr.3: 69085037-69085037 on Build GRCh38
rs368886970 Chr.3: 69085036-69085036 on Build GRCh38
rs903202730 Chr.3: 69085107-69085107 on Build GRCh38
rs767431039 Chr.3: 69085180-69085180 on Build GRCh38
rs376490250 Chr.3: 69085104-69085104 on Build GRCh38

View Full Product Details