Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 71683613-71684116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4E3
Entrez Gene ID
317649
Gene Name
eukaryotic translation initiation factor 4E family member 3
Gene Aliases
eIF-4E3, eIF4E-3
UniGene
Hs.581355
Gene Chromosome Location
Chr.3: 71679289-71754831 on Build GRCh38
Transcripts
NM_001134649.2 NM_001134650.1 NM_001134651.1 NM_001282886.1 NM_173359.4 XM_011533651.2 XM_017006278.1 XM_017006279.1 XM_017006280.1 XM_017006281.1 XM_017006282.1

SNP Detail

SNP ID Chromosome Location
rs1044743571 Chr.3: 71683876-71683876 on Build GRCh38
rs964433415 Chr.3: 71683765-71683765 on Build GRCh38
rs1011421700 Chr.3: 71683879-71683879 on Build GRCh38
rs1003296908 Chr.3: 71683650-71683650 on Build GRCh38
rs947744657 Chr.3: 71683872-71683872 on Build GRCh38
rs796971040 Chr.3: 71684026-71684026 on Build GRCh38
rs2276757 Chr.3: 71683945-71683945 on Build GRCh38
rs1052208234 Chr.3: 71683904-71683905 on Build GRCh38
rs764999312 Chr.3: 71683865-71683865 on Build GRCh38
rs34947242 Chr.3: 71683817-71683818 on Build GRCh38
rs922813660 Chr.3: 71683789-71683789 on Build GRCh38
rs966539665 Chr.3: 71683685-71683685 on Build GRCh38
rs758197616 Chr.3: 71684012-71684012 on Build GRCh38
rs997465631 Chr.3: 71684067-71684067 on Build GRCh38
rs142713172 Chr.3: 71683790-71683790 on Build GRCh38
rs976161741 Chr.3: 71683735-71683735 on Build GRCh38
rs552274362 Chr.3: 71683727-71683727 on Build GRCh38
rs1030811557 Chr.3: 71683697-71683697 on Build GRCh38
rs983058993 Chr.3: 71684066-71684066 on Build GRCh38
rs1037476571 Chr.3: 71683846-71683846 on Build GRCh38
rs938308473 Chr.3: 71683819-71683819 on Build GRCh38
rs1030978010 Chr.3: 71683678-71683678 on Build GRCh38
rs955577061 Chr.3: 71684058-71684058 on Build GRCh38
rs750252432 Chr.3: 71683962-71683962 on Build GRCh38
rs956759123 Chr.3: 71683725-71683725 on Build GRCh38
rs546218740 Chr.3: 71683805-71683805 on Build GRCh38
rs546183592 Chr.3: 71683916-71683916 on Build GRCh38
rs903116774 Chr.3: 71683996-71683996 on Build GRCh38
rs1055590708 Chr.3: 71683822-71683822 on Build GRCh38
rs564446929 Chr.3: 71684081-71684081 on Build GRCh38
rs894219910 Chr.3: 71683878-71683878 on Build GRCh38
rs967285980 Chr.3: 71683656-71683656 on Build GRCh38
rs3202957 Chr.3: 71684031-71684031 on Build GRCh38
rs747367887 Chr.3: 71684057-71684058 on Build GRCh38
rs150586974 Chr.3: 71683817-71683817 on Build GRCh38
rs535783297 Chr.3: 71684003-71684003 on Build GRCh38
rs367606108 Chr.3: 71683803-71683803 on Build GRCh38
rs751136153 Chr.3: 71683798-71683798 on Build GRCh38
rs183814575 Chr.3: 71684039-71684039 on Build GRCh38
rs1029679688 Chr.3: 71684046-71684046 on Build GRCh38
rs546475405 Chr.3: 71683764-71683764 on Build GRCh38
rs975684383 Chr.3: 71683785-71683785 on Build GRCh38
rs544126983 Chr.3: 71683658-71683658 on Build GRCh38
rs1036452403 Chr.3: 71683813-71683813 on Build GRCh38
rs767401845 Chr.3: 71683938-71683938 on Build GRCh38
rs752902092 Chr.3: 71683783-71683783 on Build GRCh38
rs1003428091 Chr.3: 71683903-71683903 on Build GRCh38
rs1022837701 Chr.3: 71684008-71684008 on Build GRCh38
rs759637590 Chr.3: 71683705-71683705 on Build GRCh38
rs569037564 Chr.3: 71684076-71684076 on Build GRCh38
rs577159247 Chr.3: 71683670-71683670 on Build GRCh38

View Full Product Details