Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87227182-87227682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP2B
Entrez Gene ID
25978
Gene Name
charged multivesicular body protein 2B
Gene Aliases
ALS17, CHMP2.5, DMT1, VPS2-2, VPS2B
UniGene
Hs.476930
Gene Chromosome Location
Chr.3: 87227263-87255548 on Build GRCh38
Transcripts
NM_001244644.1 NM_014043.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1162456 c.27C>T p.T9T Chr.3: 87227549-87227549 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771264230 Chr.3: 87227580-87227580 on Build GRCh38
rs1019631363 Chr.3: 87227276-87227276 on Build GRCh38
rs754459872 Chr.3: 87227570-87227570 on Build GRCh38
rs199854996 Chr.3: 87227489-87227489 on Build GRCh38
rs775571812 Chr.3: 87227502-87227502 on Build GRCh38
rs747484307 Chr.3: 87227574-87227574 on Build GRCh38
rs921275036 Chr.3: 87227554-87227554 on Build GRCh38
rs563173960 Chr.3: 87227210-87227210 on Build GRCh38
rs754708487 Chr.3: 87227527-87227527 on Build GRCh38
rs1027585624 Chr.3: 87227263-87227263 on Build GRCh38
rs200140890 Chr.3: 87227510-87227510 on Build GRCh38
rs908650178 Chr.3: 87227278-87227278 on Build GRCh38
rs11540917 Chr.3: 87227428-87227428 on Build GRCh38
rs1036097398 Chr.3: 87227201-87227201 on Build GRCh38
rs1002590853 Chr.3: 87227392-87227392 on Build GRCh38
rs921222813 Chr.3: 87227294-87227294 on Build GRCh38
rs766100499 Chr.3: 87227517-87227517 on Build GRCh38
rs965434696 Chr.3: 87227548-87227548 on Build GRCh38
rs773633818 Chr.3: 87227487-87227487 on Build GRCh38
rs944125378 Chr.3: 87227281-87227281 on Build GRCh38
rs756651997 Chr.3: 87227448-87227448 on Build GRCh38
rs1043312096 Chr.3: 87227474-87227474 on Build GRCh38
rs775727022 Chr.3: 87227597-87227597 on Build GRCh38
rs1002642029 Chr.3: 87227208-87227208 on Build GRCh38
rs367697846 Chr.3: 87227513-87227513 on Build GRCh38
rs984666292 Chr.3: 87227319-87227319 on Build GRCh38
rs763080668 Chr.3: 87227598-87227598 on Build GRCh38
rs961904685 Chr.3: 87227605-87227605 on Build GRCh38
rs1013188255 Chr.3: 87227447-87227447 on Build GRCh38
rs766591028 Chr.3: 87227386-87227386 on Build GRCh38
rs757832453 Chr.3: 87227476-87227477 on Build GRCh38
rs778570281 Chr.3: 87227477-87227477 on Build GRCh38
rs371801344 Chr.3: 87227567-87227567 on Build GRCh38
rs1041559270 Chr.3: 87227637-87227637 on Build GRCh38
rs534996932 Chr.3: 87227212-87227212 on Build GRCh38
rs947283089 Chr.3: 87227628-87227628 on Build GRCh38
rs778515285 Chr.3: 87227538-87227538 on Build GRCh38
rs906967958 Chr.3: 87227202-87227202 on Build GRCh38
rs35413339 Chr.3: 87227564-87227564 on Build GRCh38
rs893029176 Chr.3: 87227329-87227329 on Build GRCh38
rs947264382 Chr.3: 87227341-87227341 on Build GRCh38
rs571134459 Chr.3: 87227233-87227233 on Build GRCh38
rs750390377 Chr.3: 87227512-87227512 on Build GRCh38
rs1050330114 Chr.3: 87227316-87227316 on Build GRCh38
rs902912650 Chr.3: 87227211-87227211 on Build GRCh38
rs751800748 Chr.3: 87227218-87227218 on Build GRCh38
rs768627605 Chr.3: 87227487-87227487 on Build GRCh38
rs746915597 Chr.3: 87227485-87227494 on Build GRCh38
rs372901949 Chr.3: 87227232-87227232 on Build GRCh38
rs976757018 Chr.3: 87227293-87227293 on Build GRCh38
rs368463124 Chr.3: 87227486-87227486 on Build GRCh38
rs374964592 Chr.3: 87227583-87227583 on Build GRCh38
rs751723331 Chr.3: 87227397-87227397 on Build GRCh38
rs983344183 Chr.3: 87227269-87227269 on Build GRCh38
rs938399805 Chr.3: 87227645-87227645 on Build GRCh38
rs748625123 Chr.3: 87227479-87227479 on Build GRCh38
rs902965639 Chr.3: 87227374-87227374 on Build GRCh38
rs914533855 Chr.3: 87227612-87227612 on Build GRCh38
rs903413856 Chr.3: 87227481-87227481 on Build GRCh38
rs931916738 Chr.3: 87227295-87227295 on Build GRCh38
rs972012209 Chr.3: 87227619-87227619 on Build GRCh38
rs767269731 Chr.3: 87227437-87227437 on Build GRCh38
rs886058905 Chr.3: 87227485-87227485 on Build GRCh38
rs1035345982 Chr.3: 87227227-87227227 on Build GRCh38
rs573036388 Chr.3: 87227469-87227469 on Build GRCh38
rs772653746 Chr.3: 87227480-87227480 on Build GRCh38
rs776777126 Chr.3: 87227491-87227491 on Build GRCh38
rs11540914 Chr.3: 87227488-87227488 on Build GRCh38
rs762989447 Chr.3: 87227505-87227505 on Build GRCh38
rs574954693 Chr.3: 87227364-87227364 on Build GRCh38
rs769984979 Chr.3: 87227584-87227584 on Build GRCh38
rs779680258 Chr.3: 87227484-87227485 on Build GRCh38
rs958953316 Chr.3: 87227230-87227230 on Build GRCh38
rs947569272 Chr.3: 87227463-87227463 on Build GRCh38
rs77328592 Chr.3: 87227372-87227372 on Build GRCh38
rs1016170868 Chr.3: 87227536-87227536 on Build GRCh38
rs2279720 Chr.3: 87227549-87227549 on Build GRCh38
rs777217524 Chr.3: 87227616-87227616 on Build GRCh38
rs750699594 Chr.3: 87227429-87227429 on Build GRCh38
rs909163449 Chr.3: 87227334-87227334 on Build GRCh38
rs767454705 Chr.3: 87227511-87227511 on Build GRCh38
rs953203889 Chr.3: 87227515-87227515 on Build GRCh38
rs965814372 Chr.3: 87227279-87227279 on Build GRCh38
rs376821391 Chr.3: 87227492-87227492 on Build GRCh38
rs1032371345 Chr.3: 87227425-87227425 on Build GRCh38
rs36098294 Chr.3: 87227421-87227421 on Build GRCh38
rs768671850 Chr.3: 87227602-87227602 on Build GRCh38
rs556556377 Chr.3: 87227318-87227318 on Build GRCh38
rs938394465 Chr.3: 87227393-87227393 on Build GRCh38
rs538600777 Chr.3: 87227264-87227264 on Build GRCh38
rs753473616 Chr.3: 87227521-87227521 on Build GRCh38

View Full Product Details