Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 87991300-87991843 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR1F
Entrez Gene ID
3355
Gene Name
5-hydroxytryptamine receptor 1F
Gene Aliases
5-HT-1F, 5-HT1F, 5HT6, HTR1EL, MR77
UniGene
Hs.248136
Gene Chromosome Location
Chr.3: 87792767-87994856 on Build GRCh38
Transcripts
NM_000866.4 NM_001322208.1 NM_001322209.1 NM_001322210.1 XM_005264751.3 XM_011533664.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5691460 c.901G>T p.A301S Chr.3: 87991650-87991650 on Build GRCh38
4327759 c.922C>T p.P308S Chr.3: 87991671-87991671 on Build GRCh38
5865355 c.1012C>T p.L338F Chr.3: 87991761-87991761 on Build GRCh38
7199600 c.611T>C p.I204T Chr.3: 87991360-87991360 on Build GRCh38
1425595 c.617G>C p.R206T Chr.3: 87991366-87991366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867170750 Chr.3: 87991529-87991529 on Build GRCh38
rs775925663 Chr.3: 87991559-87991559 on Build GRCh38
rs751480837 Chr.3: 87991775-87991775 on Build GRCh38
rs757005164 Chr.3: 87991659-87991659 on Build GRCh38
rs777903863 Chr.3: 87991349-87991349 on Build GRCh38
rs748771062 Chr.3: 87991795-87991795 on Build GRCh38
rs374597659 Chr.3: 87991796-87991796 on Build GRCh38
rs745971003 Chr.3: 87991383-87991383 on Build GRCh38
rs376954240 Chr.3: 87991454-87991454 on Build GRCh38
rs772705392 Chr.3: 87991429-87991429 on Build GRCh38
rs200158656 Chr.3: 87991331-87991331 on Build GRCh38
rs753056910 Chr.3: 87991343-87991343 on Build GRCh38
rs762138823 Chr.3: 87991590-87991590 on Build GRCh38
rs780942771 Chr.3: 87991784-87991784 on Build GRCh38
rs759114776 Chr.3: 87991447-87991447 on Build GRCh38
rs748130736 Chr.3: 87991525-87991525 on Build GRCh38
rs746808571 Chr.3: 87991653-87991653 on Build GRCh38
rs756278839 Chr.3: 87991502-87991502 on Build GRCh38
rs748859479 Chr.3: 87991712-87991712 on Build GRCh38
rs766748036 Chr.3: 87991759-87991759 on Build GRCh38
rs760961399 Chr.3: 87991755-87991755 on Build GRCh38
rs776206926 Chr.3: 87991549-87991549 on Build GRCh38
rs533792234 Chr.3: 87991642-87991642 on Build GRCh38
rs781305737 Chr.3: 87991370-87991370 on Build GRCh38
rs768707290 Chr.3: 87991385-87991385 on Build GRCh38
rs767724180 Chr.3: 87991614-87991614 on Build GRCh38
rs201820111 Chr.3: 87991484-87991484 on Build GRCh38
rs750258109 Chr.3: 87991786-87991786 on Build GRCh38
rs755746856 Chr.3: 87991630-87991630 on Build GRCh38
rs6301 Chr.3: 87991532-87991532 on Build GRCh38
rs780356858 Chr.3: 87991413-87991414 on Build GRCh38
rs899591113 Chr.3: 87991688-87991688 on Build GRCh38
rs781460836 Chr.3: 87991643-87991644 on Build GRCh38
rs748579647 Chr.3: 87991673-87991673 on Build GRCh38
rs748767043 Chr.3: 87991693-87991693 on Build GRCh38
rs373080175 Chr.3: 87991471-87991471 on Build GRCh38
rs779736434 Chr.3: 87991792-87991792 on Build GRCh38
rs773522735 Chr.3: 87991748-87991748 on Build GRCh38
rs950196292 Chr.3: 87991417-87991418 on Build GRCh38
rs751914938 Chr.3: 87991479-87991479 on Build GRCh38
rs779438575 Chr.3: 87991808-87991808 on Build GRCh38
rs777602073 Chr.3: 87991534-87991534 on Build GRCh38
rs191093250 Chr.3: 87991556-87991556 on Build GRCh38
rs770614075 Chr.3: 87991548-87991548 on Build GRCh38
rs751838243 Chr.3: 87991359-87991359 on Build GRCh38
rs779588189 Chr.3: 87991637-87991637 on Build GRCh38
rs1039379923 Chr.3: 87991680-87991680 on Build GRCh38
rs201728270 Chr.3: 87991571-87991571 on Build GRCh38
rs1024864304 Chr.3: 87991769-87991769 on Build GRCh38
rs747247849 Chr.3: 87991498-87991499 on Build GRCh38
rs971347378 Chr.3: 87991487-87991487 on Build GRCh38
rs756225984 Chr.3: 87991616-87991616 on Build GRCh38
rs200867459 Chr.3: 87991438-87991438 on Build GRCh38
rs780856447 Chr.3: 87991667-87991667 on Build GRCh38
rs764036964 Chr.3: 87991773-87991773 on Build GRCh38
rs142679464 Chr.3: 87991514-87991514 on Build GRCh38
rs754250380 Chr.3: 87991330-87991330 on Build GRCh38
rs963774192 Chr.3: 87991376-87991376 on Build GRCh38
rs370338613 Chr.3: 87991464-87991464 on Build GRCh38
rs745309208 Chr.3: 87991550-87991550 on Build GRCh38
rs755754943 Chr.3: 87991790-87991790 on Build GRCh38
rs917853441 Chr.3: 87991461-87991461 on Build GRCh38
rs113447772 Chr.3: 87991519-87991519 on Build GRCh38
rs868218434 Chr.3: 87991599-87991599 on Build GRCh38
rs981437033 Chr.3: 87991787-87991787 on Build GRCh38
rs772000643 Chr.3: 87991416-87991416 on Build GRCh38
rs767913928 Chr.3: 87991356-87991356 on Build GRCh38
rs757234278 Chr.3: 87991362-87991362 on Build GRCh38
rs747897098 Chr.3: 87991409-87991409 on Build GRCh38
rs376368065 Chr.3: 87991542-87991542 on Build GRCh38
rs750500923 Chr.3: 87991497-87991497 on Build GRCh38
rs970665863 Chr.3: 87991782-87991782 on Build GRCh38
rs1027221571 Chr.3: 87991703-87991703 on Build GRCh38
rs867150601 Chr.3: 87991821-87991821 on Build GRCh38
rs200052855 Chr.3: 87991365-87991365 on Build GRCh38
rs753964138 Chr.3: 87991767-87991767 on Build GRCh38
rs758325682 Chr.3: 87991632-87991632 on Build GRCh38
rs774464006 Chr.3: 87991524-87991525 on Build GRCh38
rs769190304 Chr.3: 87991715-87991715 on Build GRCh38
rs201702555 Chr.3: 87991620-87991620 on Build GRCh38
rs757026676 Chr.3: 87991778-87991778 on Build GRCh38
rs776177421 Chr.3: 87991440-87991440 on Build GRCh38
rs141307791 Chr.3: 87991437-87991437 on Build GRCh38
rs763427983 Chr.3: 87991568-87991568 on Build GRCh38
rs374593102 Chr.3: 87991408-87991408 on Build GRCh38
rs762174194 Chr.3: 87991740-87991740 on Build GRCh38
rs765774121 Chr.3: 87991342-87991342 on Build GRCh38
rs772357190 Chr.3: 87991743-87991743 on Build GRCh38
rs200883374 Chr.3: 87991628-87991628 on Build GRCh38
rs764540990 Chr.3: 87991473-87991473 on Build GRCh38
rs745540576 Chr.3: 87991673-87991673 on Build GRCh38
rs774770360 Chr.3: 87991574-87991574 on Build GRCh38
rs769484463 Chr.3: 87991687-87991687 on Build GRCh38
rs758773315 Chr.3: 87991347-87991347 on Build GRCh38
rs759594314 Chr.3: 87991771-87991771 on Build GRCh38
rs558842950 Chr.3: 87991695-87991695 on Build GRCh38
rs767696253 Chr.3: 87991485-87991485 on Build GRCh38

View Full Product Details