Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 97877574-97878092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
UniGene
Hs.733378
Gene Chromosome Location
Chr.3: 97821930-97944986 on Build GRCh38
Transcripts
NM_153605.3 XM_005247117.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
731369 c.923G>T p.G308V Chr.3: 97877961-97877961 on Build GRCh38
5072491 c.974C>T p.P325L Chr.3: 97878012-97878012 on Build GRCh38
6635869 c.958C>T p.R320C Chr.3: 97877996-97877996 on Build GRCh38
1738847 c.553C>T p.R185C Chr.3: 97877591-97877591 on Build GRCh38
6028008 c.583_586delAGAG p.E196fs*19 Chr.3: 97877621-97877624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974814163 Chr.3: 97877609-97877609 on Build GRCh38
rs761303579 Chr.3: 97877621-97877624 on Build GRCh38
rs551119284 Chr.3: 97877717-97877717 on Build GRCh38
rs774414540 Chr.3: 97878043-97878043 on Build GRCh38
rs528260943 Chr.3: 97877716-97877716 on Build GRCh38
rs368806699 Chr.3: 97878003-97878003 on Build GRCh38
rs779885026 Chr.3: 97878061-97878063 on Build GRCh38
rs755936350 Chr.3: 97877727-97877727 on Build GRCh38
rs868662752 Chr.3: 97877828-97877828 on Build GRCh38
rs754798718 Chr.3: 97877854-97877854 on Build GRCh38
rs375789891 Chr.3: 97877804-97877804 on Build GRCh38
rs745841441 Chr.3: 97877625-97877625 on Build GRCh38
rs781148931 Chr.3: 97877623-97877623 on Build GRCh38
rs965735766 Chr.3: 97877893-97877893 on Build GRCh38
rs753867444 Chr.3: 97877848-97877848 on Build GRCh38
rs776033804 Chr.3: 97878027-97878027 on Build GRCh38
rs770316569 Chr.3: 97877901-97877901 on Build GRCh38
rs754930461 Chr.3: 97877968-97877968 on Build GRCh38
rs563102328 Chr.3: 97877618-97877618 on Build GRCh38
rs748209514 Chr.3: 97877607-97877607 on Build GRCh38
rs151245559 Chr.3: 97877996-97877996 on Build GRCh38
rs759983402 Chr.3: 97877883-97877883 on Build GRCh38
rs897854490 Chr.3: 97878046-97878046 on Build GRCh38
rs149984020 Chr.3: 97878035-97878035 on Build GRCh38
rs761041275 Chr.3: 97877703-97877703 on Build GRCh38
rs773750813 Chr.3: 97877702-97877702 on Build GRCh38
rs774714035 Chr.3: 97877998-97877998 on Build GRCh38
rs1017340759 Chr.3: 97877892-97877892 on Build GRCh38
rs750331738 Chr.3: 97877802-97877804 on Build GRCh38
rs757944490 Chr.3: 97878013-97878013 on Build GRCh38
rs775764188 Chr.3: 97877907-97877907 on Build GRCh38
rs764797513 Chr.3: 97877645-97877647 on Build GRCh38
rs373734953 Chr.3: 97878024-97878024 on Build GRCh38
rs768639623 Chr.3: 97877762-97877762 on Build GRCh38
rs746493073 Chr.3: 97877875-97877875 on Build GRCh38
rs764210342 Chr.3: 97877931-97877931 on Build GRCh38
rs751877357 Chr.3: 97877938-97877938 on Build GRCh38
rs770511997 Chr.3: 97877881-97877881 on Build GRCh38
rs747856194 Chr.3: 97877786-97877786 on Build GRCh38
rs774206870 Chr.3: 97877678-97877678 on Build GRCh38
rs974644180 Chr.3: 97877975-97877975 on Build GRCh38
rs993885581 Chr.3: 97878057-97878057 on Build GRCh38
rs368735896 Chr.3: 97877847-97877847 on Build GRCh38
rs747991615 Chr.3: 97877866-97877866 on Build GRCh38
rs374133102 Chr.3: 97877724-97877724 on Build GRCh38
rs147308479 Chr.3: 97877793-97877793 on Build GRCh38
rs763388038 Chr.3: 97877911-97877911 on Build GRCh38
rs143958468 Chr.3: 97877645-97877645 on Build GRCh38
rs922957749 Chr.3: 97877636-97877636 on Build GRCh38
rs905280472 Chr.3: 97878033-97878033 on Build GRCh38
rs767176261 Chr.3: 97877694-97877694 on Build GRCh38
rs764379457 Chr.3: 97877825-97877825 on Build GRCh38
rs767887744 Chr.3: 97878056-97878056 on Build GRCh38
rs771413363 Chr.3: 97877990-97877990 on Build GRCh38
rs745385703 Chr.3: 97878005-97878005 on Build GRCh38
rs775106340 Chr.3: 97877655-97877655 on Build GRCh38
rs975724785 Chr.3: 97877963-97877963 on Build GRCh38
rs55680275 Chr.3: 97877991-97877991 on Build GRCh38
rs763121420 Chr.3: 97877822-97877822 on Build GRCh38
rs765635108 Chr.3: 97877808-97877808 on Build GRCh38
rs908800691 Chr.3: 97877927-97877927 on Build GRCh38
rs752485058 Chr.3: 97877980-97877980 on Build GRCh38
rs780056239 Chr.3: 97877750-97877750 on Build GRCh38
rs139232160 Chr.3: 97877997-97877997 on Build GRCh38
rs767756346 Chr.3: 97877952-97877952 on Build GRCh38
rs550393891 Chr.3: 97878032-97878032 on Build GRCh38
rs777497838 Chr.3: 97877873-97877873 on Build GRCh38
rs559542940 Chr.3: 97877683-97877683 on Build GRCh38
rs757417397 Chr.3: 97877834-97877834 on Build GRCh38
rs201775543 Chr.3: 97877670-97877670 on Build GRCh38
rs749098258 Chr.3: 97877665-97877665 on Build GRCh38
rs993400317 Chr.3: 97877859-97877859 on Build GRCh38
rs773363339 Chr.3: 97878066-97878066 on Build GRCh38
rs771560077 Chr.3: 97877871-97877871 on Build GRCh38
rs569935278 Chr.3: 97877802-97877802 on Build GRCh38
rs935596696 Chr.3: 97877981-97877981 on Build GRCh38
rs751533960 Chr.3: 97877830-97877830 on Build GRCh38
rs759858687 Chr.3: 97877807-97877807 on Build GRCh38
rs762047991 Chr.3: 97878054-97878054 on Build GRCh38
rs777096541 Chr.3: 97877882-97877882 on Build GRCh38
rs774117006 Chr.3: 97877783-97877783 on Build GRCh38
rs750556921 Chr.3: 97877965-97877965 on Build GRCh38
rs749612609 Chr.3: 97878034-97878034 on Build GRCh38
rs185535219 Chr.3: 97877792-97877792 on Build GRCh38
rs1055517681 Chr.3: 97877765-97877765 on Build GRCh38
rs140793528 Chr.3: 97877857-97877857 on Build GRCh38
rs1027971013 Chr.3: 97877860-97877860 on Build GRCh38
rs761768617 Chr.3: 97877940-97877940 on Build GRCh38
rs779001629 Chr.3: 97877969-97877969 on Build GRCh38
rs777186582 Chr.3: 97877806-97877806 on Build GRCh38
rs564242205 Chr.3: 97877761-97877761 on Build GRCh38
rs936622792 Chr.3: 97878062-97878062 on Build GRCh38
rs181428974 Chr.3: 97877600-97877600 on Build GRCh38
rs753081418 Chr.3: 97877814-97877814 on Build GRCh38
rs780640303 Chr.3: 97878002-97878002 on Build GRCh38
rs370317726 Chr.3: 97877719-97877719 on Build GRCh38
rs758280998 Chr.3: 97877895-97877896 on Build GRCh38
rs1056880828 Chr.3: 97878041-97878041 on Build GRCh38

View Full Product Details