Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98017495-98018000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRR3
Entrez Gene ID
200959
Gene Name
gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene)
Gene Aliases
-
UniGene
Hs.534578
Gene Chromosome Location
Chr.3: 97986683-98035304 on Build GRCh38
Transcripts
NM_001105580.2 NR_047685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4930439 c.239-2A>T p.? Chr.3: 98017724-98017724 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs6788085 Chr.3: 98017960-98017960 on Build GRCh38
rs768547473 Chr.3: 98017611-98017612 on Build GRCh38
rs200552743 Chr.3: 98017739-98017739 on Build GRCh38
rs1044416263 Chr.3: 98017813-98017813 on Build GRCh38
rs183356048 Chr.3: 98017829-98017829 on Build GRCh38
rs774386005 Chr.3: 98017698-98017698 on Build GRCh38
rs758089977 Chr.3: 98017750-98017750 on Build GRCh38
rs770302737 Chr.3: 98017660-98017660 on Build GRCh38
rs191200032 Chr.3: 98017717-98017717 on Build GRCh38
rs369171965 Chr.3: 98017697-98017697 on Build GRCh38
rs778671207 Chr.3: 98017637-98017637 on Build GRCh38
rs531611239 Chr.3: 98017758-98017758 on Build GRCh38
rs755476490 Chr.3: 98017849-98017849 on Build GRCh38
rs1008617275 Chr.3: 98017746-98017746 on Build GRCh38
rs763973417 Chr.3: 98017752-98017752 on Build GRCh38
rs983507037 Chr.3: 98017874-98017874 on Build GRCh38
rs577042338 Chr.3: 98017628-98017628 on Build GRCh38
rs779574993 Chr.3: 98017865-98017865 on Build GRCh38
rs144957328 Chr.3: 98017770-98017770 on Build GRCh38
rs920097797 Chr.3: 98017975-98017975 on Build GRCh38
rs780849229 Chr.3: 98017772-98017772 on Build GRCh38
rs985159490 Chr.3: 98017610-98017610 on Build GRCh38
rs372528547 Chr.3: 98017640-98017640 on Build GRCh38
rs780803553 Chr.3: 98017664-98017664 on Build GRCh38
rs373783184 Chr.3: 98017704-98017704 on Build GRCh38
rs149891736 Chr.3: 98017560-98017560 on Build GRCh38
rs746346442 Chr.3: 98017666-98017666 on Build GRCh38
rs970763486 Chr.3: 98017571-98017571 on Build GRCh38
rs891598918 Chr.3: 98017832-98017832 on Build GRCh38
rs201490254 Chr.3: 98017638-98017638 on Build GRCh38
rs767657087 Chr.3: 98017624-98017624 on Build GRCh38
rs559730023 Chr.3: 98017644-98017644 on Build GRCh38
rs562149526 Chr.3: 98017728-98017728 on Build GRCh38
rs553912185 Chr.3: 98017605-98017605 on Build GRCh38
rs751149700 Chr.3: 98017753-98017753 on Build GRCh38
rs902417734 Chr.3: 98017762-98017762 on Build GRCh38
rs970439703 Chr.3: 98017861-98017861 on Build GRCh38
rs1021767462 Chr.3: 98017872-98017872 on Build GRCh38
rs758821449 Chr.3: 98017886-98017886 on Build GRCh38
rs762143862 Chr.3: 98017700-98017700 on Build GRCh38
rs1038491451 Chr.3: 98017692-98017692 on Build GRCh38
rs907095495 Chr.3: 98017824-98017824 on Build GRCh38
rs990929495 Chr.3: 98017519-98017519 on Build GRCh38
rs546039152 Chr.3: 98017682-98017682 on Build GRCh38
rs754943982 Chr.3: 98017631-98017631 on Build GRCh38
rs910764796 Chr.3: 98017639-98017639 on Build GRCh38
rs752953407 Chr.3: 98017881-98017881 on Build GRCh38
rs770491801 Chr.3: 98017679-98017679 on Build GRCh38
rs948706472 Chr.3: 98017789-98017789 on Build GRCh38
rs75634562 Chr.3: 98017709-98017709 on Build GRCh38
rs199520403 Chr.3: 98017737-98017737 on Build GRCh38
rs375897248 Chr.3: 98017657-98017657 on Build GRCh38
rs1032173454 Chr.3: 98017817-98017817 on Build GRCh38
rs961600847 Chr.3: 98017904-98017904 on Build GRCh38
rs1050234603 Chr.3: 98017714-98017714 on Build GRCh38
rs186038993 Chr.3: 98017691-98017691 on Build GRCh38
rs959426128 Chr.3: 98017825-98017825 on Build GRCh38
rs878900124 Chr.3: 98017647-98017647 on Build GRCh38
rs149023607 Chr.3: 98017888-98017888 on Build GRCh38

View Full Product Details