Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100364714-100365226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOMM70
Entrez Gene ID
9868
Gene Name
translocase of outer mitochondrial membrane 70
Gene Aliases
TOMM70A, Tom70
UniGene
Hs.227253
Gene Chromosome Location
Chr.3: 100363459-100401398 on Build GRCh38
Transcripts
NM_014820.4

SNP Detail

SNP ID Chromosome Location
rs985356723 Chr.3: 100364745-100364745 on Build GRCh38
rs566146621 Chr.3: 100365139-100365139 on Build GRCh38
rs938996271 Chr.3: 100365134-100365134 on Build GRCh38
rs993457311 Chr.3: 100365108-100365108 on Build GRCh38
rs1044745002 Chr.3: 100365119-100365119 on Build GRCh38
rs866968486 Chr.3: 100364988-100364988 on Build GRCh38
rs368789432 Chr.3: 100365133-100365133 on Build GRCh38
rs547632588 Chr.3: 100365110-100365110 on Build GRCh38
rs1026602322 Chr.3: 100365109-100365109 on Build GRCh38
rs922944586 Chr.3: 100364831-100364831 on Build GRCh38
rs1002259460 Chr.3: 100364956-100364956 on Build GRCh38
rs952407898 Chr.3: 100365115-100365115 on Build GRCh38
rs976930155 Chr.3: 100364879-100364879 on Build GRCh38
rs540683394 Chr.3: 100364850-100364851 on Build GRCh38
rs3772687 Chr.3: 100364801-100364801 on Build GRCh38
rs377316003 Chr.3: 100364854-100364855 on Build GRCh38
rs1801865 Chr.3: 100365136-100365136 on Build GRCh38
rs185834394 Chr.3: 100364762-100364762 on Build GRCh38
rs543137297 Chr.3: 100364875-100364875 on Build GRCh38
rs964845229 Chr.3: 100365140-100365140 on Build GRCh38
rs1045103524 Chr.3: 100364737-100364737 on Build GRCh38
rs910687418 Chr.3: 100364775-100364775 on Build GRCh38
rs74796833 Chr.3: 100364838-100364838 on Build GRCh38
rs896517377 Chr.3: 100365079-100365079 on Build GRCh38
rs984678691 Chr.3: 100365010-100365010 on Build GRCh38
rs1003427224 Chr.3: 100364755-100364755 on Build GRCh38
rs905673749 Chr.3: 100364936-100364936 on Build GRCh38
rs1006433610 Chr.3: 100365117-100365117 on Build GRCh38
rs145959428 Chr.3: 100364844-100364844 on Build GRCh38
rs934300981 Chr.3: 100364835-100364835 on Build GRCh38
rs1045610 Chr.3: 100365176-100365176 on Build GRCh38
rs539676443 Chr.3: 100365188-100365188 on Build GRCh38
rs554407778 Chr.3: 100364807-100364807 on Build GRCh38
rs1018206557 Chr.3: 100365118-100365118 on Build GRCh38
rs909652398 Chr.3: 100365028-100365028 on Build GRCh38
rs947272817 Chr.3: 100364849-100364849 on Build GRCh38
rs115227601 Chr.3: 100365009-100365009 on Build GRCh38
rs771853980 Chr.3: 100364764-100364764 on Build GRCh38
rs536359198 Chr.3: 100365149-100365149 on Build GRCh38

View Full Product Details