Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101230761-101231265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPG2
Entrez Gene ID
50939
Gene Name
interphotoreceptor matrix proteoglycan 2
Gene Aliases
IPM200, RP56, SPACRCAN, VMD5
UniGene
Hs.209249
Gene Chromosome Location
Chr.3: 101222546-101320575 on Build GRCh38
Transcripts
NM_016247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5863094 c.3415C>T p.P1139S Chr.3: 101230964-101230964 on Build GRCh38
727558 c.3342C>A p.V1114V Chr.3: 101231037-101231037 on Build GRCh38
2915315 c.3258G>A p.W1086* Chr.3: 101231121-101231121 on Build GRCh38
5863096 c.3238C>T p.R1080W Chr.3: 101231141-101231141 on Build GRCh38
1035753 c.3262C>T p.R1088* Chr.3: 101231117-101231117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144305775 Chr.3: 101230991-101230991 on Build GRCh38
rs1008858158 Chr.3: 101231198-101231198 on Build GRCh38
rs536621712 Chr.3: 101230799-101230799 on Build GRCh38
rs200847823 Chr.3: 101231130-101231130 on Build GRCh38
rs755996200 Chr.3: 101231002-101231002 on Build GRCh38
rs751596849 Chr.3: 101231101-101231101 on Build GRCh38
rs752837379 Chr.3: 101230983-101230983 on Build GRCh38
rs765312212 Chr.3: 101230979-101230979 on Build GRCh38
rs778413772 Chr.3: 101231032-101231032 on Build GRCh38
rs922232164 Chr.3: 101230859-101230859 on Build GRCh38
rs1014130120 Chr.3: 101231193-101231193 on Build GRCh38
rs746457874 Chr.3: 101231157-101231157 on Build GRCh38
rs778665787 Chr.3: 101231118-101231118 on Build GRCh38
rs531246692 Chr.3: 101231116-101231116 on Build GRCh38
rs548034695 Chr.3: 101231201-101231201 on Build GRCh38
rs372156074 Chr.3: 101230907-101230907 on Build GRCh38
rs886057686 Chr.3: 101231024-101231024 on Build GRCh38
rs756288071 Chr.3: 101231237-101231237 on Build GRCh38
rs182467523 Chr.3: 101231160-101231160 on Build GRCh38
rs190315698 Chr.3: 101231091-101231091 on Build GRCh38
rs772761291 Chr.3: 101230926-101230926 on Build GRCh38
rs777063202 Chr.3: 101231074-101231074 on Build GRCh38
rs756885011 Chr.3: 101230966-101230967 on Build GRCh38
rs367649636 Chr.3: 101231004-101231004 on Build GRCh38
rs373381142 Chr.3: 101231141-101231141 on Build GRCh38
rs187262692 Chr.3: 101230933-101230933 on Build GRCh38
rs1033215292 Chr.3: 101230800-101230800 on Build GRCh38
rs759763352 Chr.3: 101231075-101231075 on Build GRCh38
rs2271973 Chr.3: 101230824-101230824 on Build GRCh38
rs778475195 Chr.3: 101231019-101231021 on Build GRCh38
rs775572865 Chr.3: 101231082-101231082 on Build GRCh38
rs771526450 Chr.3: 101231055-101231055 on Build GRCh38
rs370201339 Chr.3: 101231100-101231100 on Build GRCh38
rs755393105 Chr.3: 101230891-101230891 on Build GRCh38
rs866534427 Chr.3: 101231059-101231059 on Build GRCh38
rs140621379 Chr.3: 101231054-101231054 on Build GRCh38
rs747756597 Chr.3: 101231045-101231045 on Build GRCh38
rs1055884959 Chr.3: 101230832-101230832 on Build GRCh38
rs553162401 Chr.3: 101230805-101230805 on Build GRCh38
rs748968343 Chr.3: 101231008-101231008 on Build GRCh38
rs764233004 Chr.3: 101231184-101231184 on Build GRCh38
rs758079098 Chr.3: 101231136-101231136 on Build GRCh38
rs200306665 Chr.3: 101231126-101231126 on Build GRCh38
rs771581663 Chr.3: 101231158-101231158 on Build GRCh38
rs866612545 Chr.3: 101231083-101231083 on Build GRCh38
rs555007756 Chr.3: 101230805-101230805 on Build GRCh38
rs756049687 Chr.3: 101231112-101231112 on Build GRCh38
rs771675308 Chr.3: 101231050-101231050 on Build GRCh38
rs201105142 Chr.3: 101231081-101231081 on Build GRCh38
rs532707468 Chr.3: 101231190-101231190 on Build GRCh38
rs766832557 Chr.3: 101230947-101230947 on Build GRCh38
rs113543478 Chr.3: 101230920-101230920 on Build GRCh38
rs781172642 Chr.3: 101230993-101230993 on Build GRCh38
rs754210120 Chr.3: 101230948-101230948 on Build GRCh38
rs746310179 Chr.3: 101231165-101231165 on Build GRCh38
rs376775316 Chr.3: 101230922-101230922 on Build GRCh38
rs368858737 Chr.3: 101231170-101231170 on Build GRCh38
rs761167704 Chr.3: 101230925-101230925 on Build GRCh38
rs759836540 Chr.3: 101230962-101230962 on Build GRCh38
rs777459431 Chr.3: 101231140-101231140 on Build GRCh38
rs925697655 Chr.3: 101230793-101230793 on Build GRCh38
rs763192653 Chr.3: 101231171-101231171 on Build GRCh38
rs770024660 Chr.3: 101231169-101231169 on Build GRCh38
rs1020215065 Chr.3: 101231207-101231207 on Build GRCh38
rs749940265 Chr.3: 101231155-101231156 on Build GRCh38
rs348867 Chr.3: 101230998-101230998 on Build GRCh38
rs199867882 Chr.3: 101231117-101231117 on Build GRCh38
rs558831716 Chr.3: 101230882-101230882 on Build GRCh38
rs369601149 Chr.3: 101230935-101230935 on Build GRCh38
rs537543538 Chr.3: 101230826-101230826 on Build GRCh38
rs754672236 Chr.3: 101231017-101231017 on Build GRCh38

View Full Product Details