Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108984597-108985109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MORC1
Entrez Gene ID
27136
Gene Name
MORC family CW-type zinc finger 1
Gene Aliases
CT33, MORC, ZCW6
UniGene
Hs.591296
Gene Chromosome Location
Chr.3: 108958239-109118146 on Build GRCh38
Transcripts
NM_014429.3 XM_005247362.2 XM_011512691.1 XM_011512692.1 XM_011512693.1 XM_011512694.1 XM_011512696.2 XM_011512697.2 XM_017006169.1 XM_017006170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3759599 c.2300G>A p.S767N Chr.3: 108984740-108984740 on Build GRCh38
6860861 c.2297_2300delGAAG p.R766fs*18 Chr.3: 108984740-108984743 on Build GRCh38
2917199 c.2278G>T p.D760Y Chr.3: 108984762-108984762 on Build GRCh38
3992627 c.2281G>A p.V761I Chr.3: 108984759-108984759 on Build GRCh38
5794447 c.2258-1G>C p.? Chr.3: 108984783-108984783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536595324 Chr.3: 108984866-108984866 on Build GRCh38
rs538731228 Chr.3: 108984733-108984733 on Build GRCh38
rs186810942 Chr.3: 108984938-108984938 on Build GRCh38
rs765356511 Chr.3: 108984981-108984981 on Build GRCh38
rs746706857 Chr.3: 108984743-108984743 on Build GRCh38
rs754885131 Chr.3: 108984975-108984975 on Build GRCh38
rs576342332 Chr.3: 108984927-108984927 on Build GRCh38
rs560979945 Chr.3: 108985025-108985025 on Build GRCh38
rs375787915 Chr.3: 108984851-108984851 on Build GRCh38
rs192681521 Chr.3: 108985086-108985086 on Build GRCh38
rs200908423 Chr.3: 108984820-108984820 on Build GRCh38
rs973653312 Chr.3: 108984937-108984937 on Build GRCh38
rs950218351 Chr.3: 108985056-108985056 on Build GRCh38
rs755314127 Chr.3: 108984815-108984815 on Build GRCh38
rs572109643 Chr.3: 108984993-108984993 on Build GRCh38
rs747638308 Chr.3: 108984732-108984732 on Build GRCh38
rs963565207 Chr.3: 108984677-108984677 on Build GRCh38
rs998600859 Chr.3: 108984632-108984632 on Build GRCh38
rs780614332 Chr.3: 108984698-108984698 on Build GRCh38
rs201342080 Chr.3: 108984679-108984679 on Build GRCh38
rs34598598 Chr.3: 108984814-108984815 on Build GRCh38
rs201641494 Chr.3: 108984791-108984791 on Build GRCh38
rs2290057 Chr.3: 108984740-108984740 on Build GRCh38
rs576200826 Chr.3: 108984645-108984645 on Build GRCh38
rs377523915 Chr.3: 108984822-108984822 on Build GRCh38
rs777317586 Chr.3: 108984826-108984826 on Build GRCh38
rs894738670 Chr.3: 108985013-108985014 on Build GRCh38
rs561376355 Chr.3: 108984689-108984689 on Build GRCh38
rs765159379 Chr.3: 108984785-108984785 on Build GRCh38
rs368973330 Chr.3: 108984750-108984750 on Build GRCh38
rs200767011 Chr.3: 108984699-108984699 on Build GRCh38
rs781548351 Chr.3: 108984686-108984686 on Build GRCh38
rs16855104 Chr.3: 108984660-108984660 on Build GRCh38
rs969562560 Chr.3: 108984957-108984957 on Build GRCh38
rs935022003 Chr.3: 108984990-108984990 on Build GRCh38
rs752768593 Chr.3: 108984986-108984986 on Build GRCh38
rs757293361 Chr.3: 108984684-108984684 on Build GRCh38
rs915500515 Chr.3: 108985002-108985002 on Build GRCh38
rs753747347 Chr.3: 108984672-108984672 on Build GRCh38
rs2593960 Chr.3: 108984855-108984855 on Build GRCh38
rs115155867 Chr.3: 108984891-108984891 on Build GRCh38
rs946948840 Chr.3: 108985007-108985007 on Build GRCh38
rs776364556 Chr.3: 108984759-108984759 on Build GRCh38
rs761945775 Chr.3: 108984798-108984798 on Build GRCh38
rs746547787 Chr.3: 108984833-108984833 on Build GRCh38
rs558345026 Chr.3: 108984951-108984951 on Build GRCh38
rs370312012 Chr.3: 108984693-108984693 on Build GRCh38
rs963319495 Chr.3: 108984900-108984900 on Build GRCh38
rs138140588 Chr.3: 108984859-108984859 on Build GRCh38
rs540630222 Chr.3: 108985014-108985014 on Build GRCh38
rs199712379 Chr.3: 108984706-108984706 on Build GRCh38
rs530325672 Chr.3: 108985038-108985038 on Build GRCh38
rs772904951 Chr.3: 108984849-108984849 on Build GRCh38
rs183063404 Chr.3: 108984676-108984676 on Build GRCh38
rs934139099 Chr.3: 108985036-108985036 on Build GRCh38
rs373973608 Chr.3: 108984809-108984809 on Build GRCh38
rs921750518 Chr.3: 108984871-108984871 on Build GRCh38
rs543739273 Chr.3: 108985063-108985063 on Build GRCh38
rs760405721 Chr.3: 108984808-108984808 on Build GRCh38
rs911608130 Chr.3: 108984928-108984928 on Build GRCh38
rs927611441 Chr.3: 108984736-108984736 on Build GRCh38
rs2593959 Chr.3: 108984701-108984701 on Build GRCh38
rs759250853 Chr.3: 108984778-108984778 on Build GRCh38
rs1014887560 Chr.3: 108984892-108984892 on Build GRCh38
rs760453387 Chr.3: 108984669-108984669 on Build GRCh38
rs751588875 Chr.3: 108984681-108984681 on Build GRCh38
rs774230651 Chr.3: 108985040-108985041 on Build GRCh38
rs779463218 Chr.3: 108984819-108984819 on Build GRCh38
rs2593958 Chr.3: 108984671-108984671 on Build GRCh38
rs767744282 Chr.3: 108984800-108984800 on Build GRCh38
rs758658718 Chr.3: 108984992-108984992 on Build GRCh38
rs1045883442 Chr.3: 108985081-108985081 on Build GRCh38
rs945779455 Chr.3: 108984945-108984945 on Build GRCh38

View Full Product Details