Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 111594532-111595043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD96
Entrez Gene ID
10225
Gene Name
CD96 molecule
Gene Aliases
TACTILE
UniGene
Hs.142023
Gene Chromosome Location
Chr.3: 111542079-111665996 on Build GRCh38
Transcripts
NM_001318889.1 NM_005816.4 NM_198196.2 NR_134917.1 XM_005247063.3 XM_006713469.3 XM_006713470.3 XM_017005521.1 XM_017005522.1 NM_024508.4

Gene Symbol
ZBED2
Entrez Gene ID
79413
Gene Name
zinc finger BED-type containing 2
Gene Aliases
-
UniGene
Hs.136912
Gene Chromosome Location
Chr.3: 111592900-111595335 on Build GRCh38
Transcripts
NM_001318889.1 NM_005816.4 NM_198196.2 NR_134917.1 XM_005247063.3 XM_006713469.3 XM_006713470.3 XM_017005521.1 XM_017005522.1 NM_024508.4

SNP Detail

SNP ID Chromosome Location
rs76361131 Chr.3: 111594572-111594572 on Build GRCh38
rs1008168540 Chr.3: 111594831-111594831 on Build GRCh38
rs746588124 Chr.3: 111594935-111594935 on Build GRCh38
rs778996478 Chr.3: 111594938-111594938 on Build GRCh38
rs972017891 Chr.3: 111594910-111594910 on Build GRCh38
rs139751860 Chr.3: 111594958-111594958 on Build GRCh38
rs1029489783 Chr.3: 111594729-111594729 on Build GRCh38
rs10804491 Chr.3: 111594610-111594610 on Build GRCh38
rs867558539 Chr.3: 111594590-111594590 on Build GRCh38
rs1033213820 Chr.3: 111594796-111594796 on Build GRCh38
rs909709789 Chr.3: 111594639-111594639 on Build GRCh38
rs938213 Chr.3: 111595007-111595007 on Build GRCh38
rs554647118 Chr.3: 111594843-111594843 on Build GRCh38
rs938170068 Chr.3: 111594568-111594568 on Build GRCh38
rs556997018 Chr.3: 111594926-111594926 on Build GRCh38
rs566208829 Chr.3: 111594682-111594682 on Build GRCh38
rs1055697485 Chr.3: 111594673-111594673 on Build GRCh38
rs762677933 Chr.3: 111594573-111594573 on Build GRCh38
rs552827671 Chr.3: 111594648-111594648 on Build GRCh38
rs982937874 Chr.3: 111594707-111594707 on Build GRCh38
rs755141346 Chr.3: 111594880-111594880 on Build GRCh38
rs919315942 Chr.3: 111594601-111594601 on Build GRCh38
rs930719611 Chr.3: 111594609-111594609 on Build GRCh38
rs545563829 Chr.3: 111594934-111594934 on Build GRCh38
rs926830981 Chr.3: 111594558-111594558 on Build GRCh38
rs996674811 Chr.3: 111594709-111594709 on Build GRCh38
rs772199895 Chr.3: 111594975-111594975 on Build GRCh38
rs992772632 Chr.3: 111595012-111595012 on Build GRCh38
rs889270886 Chr.3: 111594805-111594805 on Build GRCh38
rs752494296 Chr.3: 111594569-111594569 on Build GRCh38
rs999894200 Chr.3: 111594736-111594736 on Build GRCh38
rs955669891 Chr.3: 111594778-111594783 on Build GRCh38
rs902906416 Chr.3: 111594713-111594713 on Build GRCh38
rs1009633399 Chr.3: 111594791-111594791 on Build GRCh38
rs537141777 Chr.3: 111594874-111594874 on Build GRCh38
rs2036997 Chr.3: 111594970-111594970 on Build GRCh38
rs778359935 Chr.3: 111595008-111595008 on Build GRCh38
rs576925623 Chr.3: 111594927-111594927 on Build GRCh38
rs949822758 Chr.3: 111594596-111594596 on Build GRCh38
rs748337686 Chr.3: 111594964-111594964 on Build GRCh38
rs186033314 Chr.3: 111594695-111594695 on Build GRCh38
rs1041384213 Chr.3: 111594708-111594708 on Build GRCh38
rs937212297 Chr.3: 111594652-111594652 on Build GRCh38
rs984891830 Chr.3: 111594973-111594973 on Build GRCh38
rs574825807 Chr.3: 111594870-111594870 on Build GRCh38

View Full Product Details