Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 111845743-111846252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLDB2
Entrez Gene ID
90102
Gene Name
pleckstrin homology like domain family B member 2
Gene Aliases
LL5b, LL5beta
UniGene
Hs.603252
Gene Chromosome Location
Chr.3: 111732480-111976517 on Build GRCh38
Transcripts
NM_001134437.1 NM_001185106.1 NM_153268.3

Gene Symbol
PLCXD2
Entrez Gene ID
257068
Gene Name
phosphatidylinositol specific phospholipase C X domain containing 2
Gene Aliases
-
UniGene
Hs.477114
Gene Chromosome Location
Chr.3: 111674676-111846447 on Build GRCh38
Transcripts
NM_001134437.1 NM_001185106.1 NM_153268.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4454911 c.60A>G p.Q20Q Chr.3: 111845928-111845928 on Build GRCh38
5756140 c.896G>T p.R299I Chr.3: 111845850-111845850 on Build GRCh38
1693158 c.880C>T p.P294S Chr.3: 111845834-111845834 on Build GRCh38
3585226 c.36G>C p.K12N Chr.3: 111845904-111845904 on Build GRCh38
1036227 c.20_21delAG p.E9fs*9 Chr.3: 111845888-111845889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766458500 Chr.3: 111846003-111846003 on Build GRCh38
rs267599542 Chr.3: 111845880-111845880 on Build GRCh38
rs1025389887 Chr.3: 111845825-111845825 on Build GRCh38
rs1021914376 Chr.3: 111846031-111846031 on Build GRCh38
rs748576905 Chr.3: 111845954-111845954 on Build GRCh38
rs773092803 Chr.3: 111845925-111845925 on Build GRCh38
rs924693186 Chr.3: 111846027-111846027 on Build GRCh38
rs866518282 Chr.3: 111846172-111846172 on Build GRCh38
rs933034421 Chr.3: 111846205-111846205 on Build GRCh38
rs549952387 Chr.3: 111846030-111846031 on Build GRCh38
rs531816644 Chr.3: 111845836-111845836 on Build GRCh38
rs772560284 Chr.3: 111845846-111845846 on Build GRCh38
rs139783116 Chr.3: 111845833-111845833 on Build GRCh38
rs756243624 Chr.3: 111845927-111845927 on Build GRCh38
rs564922506 Chr.3: 111846000-111846000 on Build GRCh38
rs1025823808 Chr.3: 111846151-111846151 on Build GRCh38
rs748544895 Chr.3: 111845843-111845843 on Build GRCh38
rs373228894 Chr.3: 111845865-111845865 on Build GRCh38
rs372180305 Chr.3: 111845818-111845818 on Build GRCh38
rs57510764 Chr.3: 111845768-111845768 on Build GRCh38
rs752646696 Chr.3: 111845786-111845786 on Build GRCh38
rs1039176132 Chr.3: 111845990-111845990 on Build GRCh38
rs527590952 Chr.3: 111846025-111846025 on Build GRCh38
rs200623527 Chr.3: 111845794-111845794 on Build GRCh38
rs752052953 Chr.3: 111845973-111845992 on Build GRCh38
rs766498960 Chr.3: 111845902-111845905 on Build GRCh38
rs762990101 Chr.3: 111845879-111845882 on Build GRCh38
rs762757292 Chr.3: 111845895-111845895 on Build GRCh38
rs937557 Chr.3: 111845992-111845992 on Build GRCh38
rs779252062 Chr.3: 111845838-111845838 on Build GRCh38
rs756255482 Chr.3: 111845802-111845802 on Build GRCh38
rs547326185 Chr.3: 111846028-111846028 on Build GRCh38
rs566447047 Chr.3: 111846171-111846171 on Build GRCh38
rs752553376 Chr.3: 111845893-111845893 on Build GRCh38
rs941679966 Chr.3: 111846158-111846158 on Build GRCh38
rs937558 Chr.3: 111846138-111846138 on Build GRCh38
rs753993588 Chr.3: 111845934-111845934 on Build GRCh38
rs377738910 Chr.3: 111845896-111845896 on Build GRCh38
rs753656346 Chr.3: 111845774-111845774 on Build GRCh38
rs755205495 Chr.3: 111845946-111845946 on Build GRCh38
rs868022313 Chr.3: 111845826-111845826 on Build GRCh38
rs372254649 Chr.3: 111845775-111845775 on Build GRCh38
rs900873372 Chr.3: 111846102-111846102 on Build GRCh38
rs201186190 Chr.3: 111845884-111845884 on Build GRCh38
rs758803127 Chr.3: 111845985-111845985 on Build GRCh38
rs779098923 Chr.3: 111845952-111845952 on Build GRCh38
rs749638648 Chr.3: 111845821-111845821 on Build GRCh38
rs773454750 Chr.3: 111845812-111845812 on Build GRCh38
rs369583710 Chr.3: 111845859-111845859 on Build GRCh38
rs370186491 Chr.3: 111845899-111845899 on Build GRCh38
rs758364258 Chr.3: 111845790-111845790 on Build GRCh38
rs769859774 Chr.3: 111846156-111846156 on Build GRCh38
rs376727397 Chr.3: 111845845-111845845 on Build GRCh38
rs1001570340 Chr.3: 111846049-111846049 on Build GRCh38
rs904501653 Chr.3: 111846044-111846044 on Build GRCh38
rs766582575 Chr.3: 111845931-111845931 on Build GRCh38
rs201415940 Chr.3: 111845881-111845881 on Build GRCh38
rs144138155 Chr.3: 111846086-111846086 on Build GRCh38
rs983771708 Chr.3: 111846143-111846143 on Build GRCh38
rs368997068 Chr.3: 111845797-111845797 on Build GRCh38
rs765061373 Chr.3: 111845890-111845890 on Build GRCh38
rs893735455 Chr.3: 111846065-111846065 on Build GRCh38
rs139342193 Chr.3: 111846066-111846066 on Build GRCh38
rs1053524464 Chr.3: 111846052-111846052 on Build GRCh38

View Full Product Details