Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112151575-112152010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC9C1
Entrez Gene ID
285335
Gene Name
solute carrier family 9 member C1
Gene Aliases
NHE, NHE-10, SLC9A10, sperm-NHE
UniGene
Hs.680112
Gene Chromosome Location
Chr.3: 112140898-112294258 on Build GRCh38
Transcripts
NM_001320531.1 NM_183061.2 NR_135297.1 XM_006713603.2 XM_011512718.1 XM_011512719.1 XM_011512720.1 XM_011512721.1 XM_011512725.2 XM_017006246.1 XM_017006247.1 XM_017006248.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6342670 c.3469C>A p.L1157M Chr.3: 112151912-112151912 on Build GRCh38
3118261 c.3436G>A p.A1146T Chr.3: 112151945-112151945 on Build GRCh38
5654703 c.3457C>G p.P1153A Chr.3: 112151924-112151924 on Build GRCh38
7421207 c.3434C>A p.A1145D Chr.3: 112151947-112151947 on Build GRCh38
4775998 c.3499C>T p.P1167S Chr.3: 112151882-112151882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039713979 Chr.3: 112151644-112151644 on Build GRCh38
rs988564082 Chr.3: 112151954-112151954 on Build GRCh38
rs760429274 Chr.3: 112151976-112151976 on Build GRCh38
rs746233266 Chr.3: 112151833-112151833 on Build GRCh38
rs376085181 Chr.3: 112151940-112151940 on Build GRCh38
rs769099787 Chr.3: 112151884-112151884 on Build GRCh38
rs779331823 Chr.3: 112151959-112151959 on Build GRCh38
rs1051794134 Chr.3: 112151705-112151705 on Build GRCh38
rs756267726 Chr.3: 112151945-112151945 on Build GRCh38
rs757584639 Chr.3: 112151824-112151824 on Build GRCh38
rs756838412 Chr.3: 112151671-112151671 on Build GRCh38
rs886558144 Chr.3: 112151727-112151727 on Build GRCh38
rs1037667445 Chr.3: 112151746-112151746 on Build GRCh38
rs749848774 Chr.3: 112151864-112151864 on Build GRCh38
rs922108802 Chr.3: 112151647-112151647 on Build GRCh38
rs748638400 Chr.3: 112151966-112151966 on Build GRCh38
rs368893280 Chr.3: 112151888-112151888 on Build GRCh38
rs771722882 Chr.3: 112151890-112151890 on Build GRCh38
rs772568670 Chr.3: 112151974-112151974 on Build GRCh38
rs967141935 Chr.3: 112151786-112151786 on Build GRCh38
rs757355441 Chr.3: 112151929-112151929 on Build GRCh38
rs200185816 Chr.3: 112151952-112151952 on Build GRCh38
rs184621890 Chr.3: 112151689-112151689 on Build GRCh38
rs1023333439 Chr.3: 112151821-112151821 on Build GRCh38
rs780250904 Chr.3: 112151851-112151851 on Build GRCh38
rs549677947 Chr.3: 112151946-112151946 on Build GRCh38
rs773834980 Chr.3: 112151865-112151865 on Build GRCh38
rs753556924 Chr.3: 112151920-112151920 on Build GRCh38
rs759569049 Chr.3: 112151923-112151923 on Build GRCh38
rs995406976 Chr.3: 112151799-112151799 on Build GRCh38
rs770258591 Chr.3: 112151845-112151845 on Build GRCh38
rs760242484 Chr.3: 112151915-112151915 on Build GRCh38
rs772979340 Chr.3: 112151910-112151910 on Build GRCh38
rs771640982 Chr.3: 112151969-112151969 on Build GRCh38
rs781126084 Chr.3: 112151936-112151936 on Build GRCh38
rs201829027 Chr.3: 112151924-112151924 on Build GRCh38
rs766240649 Chr.3: 112151917-112151917 on Build GRCh38
rs768627682 Chr.3: 112151828-112151828 on Build GRCh38
rs573793957 Chr.3: 112151748-112151755 on Build GRCh38
rs145087190 Chr.3: 112151860-112151860 on Build GRCh38
rs947627573 Chr.3: 112151613-112151613 on Build GRCh38
rs112577278 Chr.3: 112151802-112151802 on Build GRCh38
rs899179936 Chr.3: 112151754-112151754 on Build GRCh38
rs879223928 Chr.3: 112151766-112151766 on Build GRCh38
rs1035431529 Chr.3: 112151947-112151947 on Build GRCh38
rs1032617035 Chr.3: 112151870-112151870 on Build GRCh38
rs1004930840 Chr.3: 112151739-112151739 on Build GRCh38
rs890401764 Chr.3: 112151687-112151687 on Build GRCh38
rs781410178 Chr.3: 112151827-112151827 on Build GRCh38
rs371646327 Chr.3: 112151818-112151818 on Build GRCh38

View Full Product Details