Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113296635-113297140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP44
Entrez Gene ID
55779
Gene Name
cilia and flagella associated protein 44
Gene Aliases
WDR52
UniGene
Hs.584936
Gene Chromosome Location
Chr.3: 113286930-113445190 on Build GRCh38
Transcripts
NM_001164496.1 XM_005247617.3 XM_006713696.2 XM_011512977.2 XM_011512978.2 XM_011512979.2 XM_017006842.1 XM_017006843.1 XM_017006844.1 XM_017006845.1 XM_017006846.1 XM_017006847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195028 c.2567G>A p.R856H Chr.3: 113296840-113296840 on Build GRCh38
5750878 c.2642G>A p.R881Q Chr.3: 113296765-113296765 on Build GRCh38
5624523 c.931C>A p.R311S Chr.3: 113296841-113296841 on Build GRCh38
5624522 c.2566C>A p.R856S Chr.3: 113296841-113296841 on Build GRCh38
5397828 c.2578C>T p.L860L Chr.3: 113296829-113296829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1022784812 Chr.3: 113297019-113297019 on Build GRCh38
rs181057895 Chr.3: 113297061-113297061 on Build GRCh38
rs112007652 Chr.3: 113296955-113296955 on Build GRCh38
rs564766641 Chr.3: 113296898-113296898 on Build GRCh38
rs914336833 Chr.3: 113296821-113296821 on Build GRCh38
rs928308717 Chr.3: 113296795-113296795 on Build GRCh38
rs75960443 Chr.3: 113297096-113297096 on Build GRCh38
rs574753062 Chr.3: 113296746-113296746 on Build GRCh38
rs808950 Chr.3: 113296706-113296706 on Build GRCh38
rs900552685 Chr.3: 113296792-113296792 on Build GRCh38
rs200900144 Chr.3: 113296877-113296877 on Build GRCh38
rs189524927 Chr.3: 113296765-113296765 on Build GRCh38
rs981062805 Chr.3: 113296775-113296775 on Build GRCh38
rs932897327 Chr.3: 113296914-113296914 on Build GRCh38
rs144500668 Chr.3: 113296701-113296701 on Build GRCh38
rs1022267617 Chr.3: 113296853-113296853 on Build GRCh38
rs570455884 Chr.3: 113296698-113296698 on Build GRCh38
rs758988568 Chr.3: 113296747-113296747 on Build GRCh38
rs749059989 Chr.3: 113296828-113296828 on Build GRCh38
rs576834835 Chr.3: 113296933-113296933 on Build GRCh38
rs933391072 Chr.3: 113297119-113297119 on Build GRCh38
rs774436326 Chr.3: 113296868-113296868 on Build GRCh38
rs533743755 Chr.3: 113296668-113296668 on Build GRCh38
rs779948250 Chr.3: 113296826-113296826 on Build GRCh38
rs712530 Chr.3: 113297109-113297109 on Build GRCh38
rs112325705 Chr.3: 113296706-113296706 on Build GRCh38
rs552014961 Chr.3: 113296669-113296669 on Build GRCh38
rs1015665516 Chr.3: 113296956-113296956 on Build GRCh38
rs981344147 Chr.3: 113297072-113297072 on Build GRCh38
rs148547745 Chr.3: 113296800-113296800 on Build GRCh38
rs909749586 Chr.3: 113297058-113297058 on Build GRCh38
rs2270781 Chr.3: 113296813-113296813 on Build GRCh38
rs918546017 Chr.3: 113296675-113296675 on Build GRCh38
rs754336666 Chr.3: 113297074-113297074 on Build GRCh38
rs929859112 Chr.3: 113296693-113296693 on Build GRCh38
rs142935053 Chr.3: 113297002-113297002 on Build GRCh38
rs201886712 Chr.3: 113296840-113296840 on Build GRCh38
rs1010432113 Chr.3: 113296846-113296846 on Build GRCh38
rs969912039 Chr.3: 113297044-113297044 on Build GRCh38
rs185216206 Chr.3: 113297089-113297089 on Build GRCh38
rs891150491 Chr.3: 113296944-113296944 on Build GRCh38
rs984072129 Chr.3: 113297025-113297026 on Build GRCh38
rs368393292 Chr.3: 113296867-113296867 on Build GRCh38
rs995348101 Chr.3: 113297000-113297000 on Build GRCh38
rs992516654 Chr.3: 113296928-113296928 on Build GRCh38
rs1034136558 Chr.3: 113296888-113296888 on Build GRCh38
rs1022817426 Chr.3: 113296754-113296754 on Build GRCh38
rs764605240 Chr.3: 113296766-113296766 on Build GRCh38
rs898598217 Chr.3: 113296995-113296995 on Build GRCh38
rs745591378 Chr.3: 113296806-113296806 on Build GRCh38
rs780669248 Chr.3: 113296805-113296805 on Build GRCh38
rs575516332 Chr.3: 113296909-113296909 on Build GRCh38
rs759664024 Chr.3: 113296684-113296684 on Build GRCh38
rs752018447 Chr.3: 113296776-113296776 on Build GRCh38

View Full Product Details