Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119608685-119609193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA1A
Entrez Gene ID
51365
Gene Name
phospholipase A1 member A
Gene Aliases
PS-PLA1, PSPLA1
UniGene
Hs.437451
Gene Chromosome Location
Chr.3: 119597848-119629811 on Build GRCh38
Transcripts
NM_001206960.1 NM_001206961.1 NM_001293225.1 NM_015900.3 NR_120610.1 XM_017006572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3120070 c.417G>A p.L139L Chr.3: 119608911-119608911 on Build GRCh38
4714877 c.400delA p.N135fs*2 Chr.3: 119608894-119608894 on Build GRCh38
6747656 c.336G>A p.T112T Chr.3: 119608830-119608830 on Build GRCh38
1418035 c.436T>G p.F146V Chr.3: 119608930-119608930 on Build GRCh38
6163081 c.328C>A p.R110S Chr.3: 119608822-119608822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754255086 Chr.3: 119608903-119608903 on Build GRCh38
rs768879250 Chr.3: 119608862-119608862 on Build GRCh38
rs767055392 Chr.3: 119608990-119608990 on Build GRCh38
rs761774913 Chr.3: 119608967-119608967 on Build GRCh38
rs201244962 Chr.3: 119608750-119608750 on Build GRCh38
rs867103446 Chr.3: 119608817-119608817 on Build GRCh38
rs752227229 Chr.3: 119608915-119608915 on Build GRCh38
rs750748287 Chr.3: 119608810-119608810 on Build GRCh38
rs199894684 Chr.3: 119608756-119608756 on Build GRCh38
rs772138872 Chr.3: 119608970-119608970 on Build GRCh38
rs768327394 Chr.3: 119608736-119608736 on Build GRCh38
rs776583416 Chr.3: 119608769-119608769 on Build GRCh38
rs551983611 Chr.3: 119609164-119609164 on Build GRCh38
rs984762167 Chr.3: 119608824-119608824 on Build GRCh38
rs368245262 Chr.3: 119608807-119608807 on Build GRCh38
rs566332572 Chr.3: 119609086-119609086 on Build GRCh38
rs1047402171 Chr.3: 119609089-119609089 on Build GRCh38
rs147462697 Chr.3: 119608809-119608809 on Build GRCh38
rs772873262 Chr.3: 119608753-119608753 on Build GRCh38
rs1057334526 Chr.3: 119608721-119608721 on Build GRCh38
rs748054913 Chr.3: 119608748-119608748 on Build GRCh38
rs376515521 Chr.3: 119608720-119608720 on Build GRCh38
rs74582733 Chr.3: 119608723-119608723 on Build GRCh38
rs747721461 Chr.3: 119608831-119608831 on Build GRCh38
rs756705282 Chr.3: 119608917-119608917 on Build GRCh38
rs9860404 Chr.3: 119609067-119609067 on Build GRCh38
rs765031126 Chr.3: 119608717-119608717 on Build GRCh38
rs540239329 Chr.3: 119609052-119609052 on Build GRCh38
rs397875901 Chr.3: 119608762-119608762 on Build GRCh38
rs754667968 Chr.3: 119608911-119608911 on Build GRCh38
rs1038275236 Chr.3: 119609014-119609014 on Build GRCh38
rs142605269 Chr.3: 119609166-119609166 on Build GRCh38
rs61733987 Chr.3: 119608823-119608823 on Build GRCh38
rs1005521194 Chr.3: 119609104-119609104 on Build GRCh38
rs948951191 Chr.3: 119609000-119609000 on Build GRCh38
rs369466528 Chr.3: 119608984-119608984 on Build GRCh38
rs768869454 Chr.3: 119608763-119608763 on Build GRCh38
rs199983369 Chr.3: 119608804-119608804 on Build GRCh38
rs1015981966 Chr.3: 119608793-119608793 on Build GRCh38
rs145457987 Chr.3: 119608829-119608829 on Build GRCh38
rs901711914 Chr.3: 119609036-119609036 on Build GRCh38
rs375246969 Chr.3: 119608925-119608925 on Build GRCh38
rs762021809 Chr.3: 119608988-119608990 on Build GRCh38
rs139965899 Chr.3: 119608801-119608801 on Build GRCh38
rs773193409 Chr.3: 119608983-119608983 on Build GRCh38
rs756539203 Chr.3: 119608814-119608814 on Build GRCh38
rs746927050 Chr.3: 119608922-119608922 on Build GRCh38
rs5852212 Chr.3: 119608758-119608758 on Build GRCh38
rs778778801 Chr.3: 119608913-119608913 on Build GRCh38
rs780538330 Chr.3: 119609110-119609110 on Build GRCh38
rs771903126 Chr.3: 119608833-119608833 on Build GRCh38
rs777808563 Chr.3: 119608839-119608839 on Build GRCh38
rs1723969 Chr.3: 119608806-119608806 on Build GRCh38
rs774016862 Chr.3: 119608746-119608746 on Build GRCh38
rs149097224 Chr.3: 119608863-119608863 on Build GRCh38
rs539659283 Chr.3: 119609101-119609101 on Build GRCh38
rs142153170 Chr.3: 119608865-119608865 on Build GRCh38
rs775739657 Chr.3: 119608791-119608791 on Build GRCh38
rs777172711 Chr.3: 119608918-119608918 on Build GRCh38
rs760588762 Chr.3: 119608781-119608781 on Build GRCh38
rs1013085487 Chr.3: 119608762-119608762 on Build GRCh38
rs528036437 Chr.3: 119608850-119608850 on Build GRCh38
rs766806936 Chr.3: 119608890-119608890 on Build GRCh38
rs768860913 Chr.3: 119608953-119608953 on Build GRCh38
rs774594490 Chr.3: 119608957-119608957 on Build GRCh38
rs746760433 Chr.3: 119608849-119608849 on Build GRCh38
rs748774228 Chr.3: 119608728-119608728 on Build GRCh38
rs761106273 Chr.3: 119608870-119608870 on Build GRCh38
rs540932793 Chr.3: 119608837-119608837 on Build GRCh38
rs114103322 Chr.3: 119608830-119608830 on Build GRCh38
rs371605005 Chr.3: 119608866-119608866 on Build GRCh38
rs779821204 Chr.3: 119608725-119608725 on Build GRCh38
rs765398614 Chr.3: 119608991-119608992 on Build GRCh38
rs745720363 Chr.3: 119608948-119608948 on Build GRCh38
rs776442517 Chr.3: 119608855-119608855 on Build GRCh38
rs750706510 Chr.3: 119609167-119609167 on Build GRCh38
rs750986427 Chr.3: 119608867-119608867 on Build GRCh38
rs916090673 Chr.3: 119608999-119608999 on Build GRCh38
rs1057364396 Chr.3: 119608949-119608949 on Build GRCh38
rs150705035 Chr.3: 119608788-119608788 on Build GRCh38
rs763094806 Chr.3: 119608856-119608856 on Build GRCh38
rs1016952641 Chr.3: 119609121-119609121 on Build GRCh38
rs770604782 Chr.3: 119609055-119609055 on Build GRCh38
rs780908128 Chr.3: 119608927-119608927 on Build GRCh38

View Full Product Details