Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119676962-119677498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX17
Entrez Gene ID
10063
Gene Name
COX17, cytochrome c oxidase copper chaperone
Gene Aliases
-
UniGene
Hs.534383
Gene Chromosome Location
Chr.3: 119659975-119677441 on Build GRCh38
Transcripts
NM_005694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3845921 c.96G>A p.A32A Chr.3: 119677215-119677215 on Build GRCh38
3845922 c.20C>G p.S7* Chr.3: 119677291-119677291 on Build GRCh38
6634322 c.51G>T p.K17N Chr.3: 119677260-119677260 on Build GRCh38
6634323 c.106T>C p.C36R Chr.3: 119677205-119677205 on Build GRCh38
295010 c.23A>C p.N8T Chr.3: 119677288-119677288 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs191351133 Chr.3: 119677065-119677065 on Build GRCh38
rs776316905 Chr.3: 119677257-119677259 on Build GRCh38
rs902680092 Chr.3: 119677117-119677117 on Build GRCh38
rs183531260 Chr.3: 119677092-119677092 on Build GRCh38
rs573859174 Chr.3: 119677168-119677168 on Build GRCh38
rs962660760 Chr.3: 119677204-119677204 on Build GRCh38
rs538625306 Chr.3: 119677110-119677110 on Build GRCh38
rs879781586 Chr.3: 119677076-119677089 on Build GRCh38
rs902705814 Chr.3: 119677386-119677386 on Build GRCh38
rs929823828 Chr.3: 119677271-119677271 on Build GRCh38
rs567442873 Chr.3: 119677096-119677096 on Build GRCh38
rs372993809 Chr.3: 119677105-119677125 on Build GRCh38
rs779218982 Chr.3: 119677281-119677281 on Build GRCh38
rs771090370 Chr.3: 119677003-119677004 on Build GRCh38
rs754728319 Chr.3: 119677340-119677340 on Build GRCh38
rs61736422 Chr.3: 119677239-119677239 on Build GRCh38
rs537274114 Chr.3: 119677148-119677148 on Build GRCh38
rs367671485 Chr.3: 119677063-119677063 on Build GRCh38
rs955085168 Chr.3: 119677395-119677395 on Build GRCh38
rs777924465 Chr.3: 119677293-119677293 on Build GRCh38
rs896241863 Chr.3: 119677416-119677416 on Build GRCh38
rs538288513 Chr.3: 119677434-119677434 on Build GRCh38
rs944764005 Chr.3: 119677353-119677353 on Build GRCh38
rs528249091 Chr.3: 119677377-119677377 on Build GRCh38
rs34292626 Chr.3: 119677338-119677339 on Build GRCh38
rs71156765 Chr.3: 119677027-119677028 on Build GRCh38
rs1011142600 Chr.3: 119677372-119677372 on Build GRCh38
rs771065921 Chr.3: 119677198-119677198 on Build GRCh38
rs760102276 Chr.3: 119677217-119677217 on Build GRCh38
rs755802772 Chr.3: 119677166-119677166 on Build GRCh38
rs760634772 Chr.3: 119677378-119677378 on Build GRCh38
rs748360418 Chr.3: 119677178-119677178 on Build GRCh38
rs370907975 Chr.3: 119677163-119677163 on Build GRCh38
rs772401538 Chr.3: 119677154-119677174 on Build GRCh38
rs1020008196 Chr.3: 119677023-119677023 on Build GRCh38
rs548709252 Chr.3: 119677072-119677092 on Build GRCh38
rs1035155716 Chr.3: 119677412-119677412 on Build GRCh38
rs770040932 Chr.3: 119677230-119677230 on Build GRCh38
rs761545191 Chr.3: 119677326-119677326 on Build GRCh38
rs985056988 Chr.3: 119677144-119677144 on Build GRCh38
rs933648951 Chr.3: 119677140-119677140 on Build GRCh38
rs11558038 Chr.3: 119677307-119677307 on Build GRCh38
rs537362660 Chr.3: 119677458-119677458 on Build GRCh38
rs7618526 Chr.3: 119677071-119677071 on Build GRCh38
rs11558040 Chr.3: 119677306-119677306 on Build GRCh38
rs760501748 Chr.3: 119677336-119677336 on Build GRCh38
rs534724994 Chr.3: 119677097-119677097 on Build GRCh38
rs761364347 Chr.3: 119677263-119677263 on Build GRCh38
rs577993070 Chr.3: 119677278-119677278 on Build GRCh38
rs541134728 Chr.3: 119677191-119677191 on Build GRCh38
rs571310893 Chr.3: 119677406-119677406 on Build GRCh38
rs780939639 Chr.3: 119677154-119677154 on Build GRCh38
rs771261337 Chr.3: 119677083-119677089 on Build GRCh38
rs896973999 Chr.3: 119677246-119677246 on Build GRCh38
rs13067694 Chr.3: 119677008-119677008 on Build GRCh38
rs563464418 Chr.3: 119677299-119677299 on Build GRCh38
rs749618468 Chr.3: 119677308-119677308 on Build GRCh38
rs902953556 Chr.3: 119677053-119677053 on Build GRCh38
rs749505426 Chr.3: 119677004-119677008 on Build GRCh38
rs558206349 Chr.3: 119677126-119677126 on Build GRCh38
rs746006976 Chr.3: 119677361-119677361 on Build GRCh38
rs545372777 Chr.3: 119677284-119677284 on Build GRCh38
rs141108342 Chr.3: 119677304-119677304 on Build GRCh38
rs576658447 Chr.3: 119677127-119677127 on Build GRCh38
rs1014055093 Chr.3: 119677018-119677018 on Build GRCh38
rs571158479 Chr.3: 119677107-119677107 on Build GRCh38
rs567286103 Chr.3: 119677033-119677035 on Build GRCh38
rs776062046 Chr.3: 119677234-119677234 on Build GRCh38
rs778117192 Chr.3: 119677123-119677129 on Build GRCh38
rs758599418 Chr.3: 119677289-119677289 on Build GRCh38
rs750401989 Chr.3: 119677164-119677164 on Build GRCh38
rs867299579 Chr.3: 119677276-119677276 on Build GRCh38
rs369344618 Chr.3: 119677157-119677157 on Build GRCh38
rs999303613 Chr.3: 119677125-119677125 on Build GRCh38
rs141671893 Chr.3: 119677435-119677435 on Build GRCh38
rs757217777 Chr.3: 119677300-119677300 on Build GRCh38
rs376284865 Chr.3: 119677103-119677109 on Build GRCh38
rs867217884 Chr.3: 119677028-119677028 on Build GRCh38
rs774076220 Chr.3: 119677318-119677318 on Build GRCh38
rs768999715 Chr.3: 119677255-119677255 on Build GRCh38
rs373100398 Chr.3: 119677116-119677129 on Build GRCh38
rs775928124 Chr.3: 119677187-119677201 on Build GRCh38
rs751766550 Chr.3: 119677349-119677349 on Build GRCh38
rs373552751 Chr.3: 119677344-119677344 on Build GRCh38
rs776964268 Chr.3: 119677212-119677212 on Build GRCh38
rs766355272 Chr.3: 119677337-119677337 on Build GRCh38
rs191836063 Chr.3: 119677445-119677445 on Build GRCh38
rs895615754 Chr.3: 119677013-119677013 on Build GRCh38
rs756138478 Chr.3: 119677279-119677279 on Build GRCh38
rs772366536 Chr.3: 119677366-119677366 on Build GRCh38
rs189940878 Chr.3: 119677440-119677440 on Build GRCh38
rs761097475 Chr.3: 119677190-119677190 on Build GRCh38
rs867156677 Chr.3: 119677089-119677089 on Build GRCh38
rs747141970 Chr.3: 119677197-119677197 on Build GRCh38
rs553331091 Chr.3: 119677024-119677027 on Build GRCh38
rs751276782 Chr.3: 119677260-119677260 on Build GRCh38
rs368546522 Chr.3: 119677316-119677316 on Build GRCh38
rs370422554 Chr.3: 119677331-119677331 on Build GRCh38
rs751329108 Chr.3: 119677109-119677129 on Build GRCh38
rs555561766 Chr.3: 119677165-119677165 on Build GRCh38
rs546679052 Chr.3: 119677106-119677106 on Build GRCh38
rs375003489 Chr.3: 119677153-119677153 on Build GRCh38
rs546426061 Chr.3: 119677390-119677390 on Build GRCh38
rs999710931 Chr.3: 119677410-119677410 on Build GRCh38
rs1022464855 Chr.3: 119677098-119677098 on Build GRCh38
rs1044362660 Chr.3: 119677383-119677383 on Build GRCh38
rs368987137 Chr.3: 119677019-119677025 on Build GRCh38
rs753796656 Chr.3: 119677338-119677338 on Build GRCh38
rs767054140 Chr.3: 119677270-119677270 on Build GRCh38
rs966900048 Chr.3: 119677374-119677374 on Build GRCh38
rs763886228 Chr.3: 119677242-119677242 on Build GRCh38
rs911995721 Chr.3: 119677351-119677351 on Build GRCh38
rs78442596 Chr.3: 119677373-119677373 on Build GRCh38
rs772350414 Chr.3: 119677190-119677190 on Build GRCh38
rs753675980 Chr.3: 119677288-119677288 on Build GRCh38
rs947030500 Chr.3: 119677382-119677382 on Build GRCh38
rs757565261 Chr.3: 119677350-119677350 on Build GRCh38
rs781099452 Chr.3: 119677360-119677360 on Build GRCh38
rs973999600 Chr.3: 119677432-119677432 on Build GRCh38
rs9854767 Chr.3: 119677075-119677075 on Build GRCh38

View Full Product Details