Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121667928-121668367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGB1
Entrez Gene ID
2804
Gene Name
golgin B1
Gene Aliases
GCP, GCP372, GOLIM1
UniGene
Hs.213389
Gene Chromosome Location
Chr.3: 121663199-121749908 on Build GRCh38
Transcripts
NM_001256486.1 NM_001256487.1 NM_001256488.1 NM_004487.4 XM_005247371.4 XM_005247372.4 XM_005247373.1 XM_006713587.1 XM_006713588.1 XM_006713589.1 XM_006713590.1 XM_006713591.1 XM_011512699.2 XM_017006189.1 XM_017006190.1 XM_017006191.1 XM_017006192.1 XM_017006193.1 XM_017006194.1 XM_017006195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7099201 c.9351G>C p.K3117N Chr.3: 121668114-121668114 on Build GRCh38
1036981 c.9351G>T p.K3117N Chr.3: 121668114-121668114 on Build GRCh38
6453276 c.9358G>A p.V3120I Chr.3: 121668107-121668107 on Build GRCh38
6492008 c.9371G>A p.S3124N Chr.3: 121668094-121668094 on Build GRCh38
4636082 c.9339T>C p.A3113A Chr.3: 121668126-121668126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756464844 Chr.3: 121668119-121668119 on Build GRCh38
rs752713984 Chr.3: 121668334-121668334 on Build GRCh38
rs748558505 Chr.3: 121668042-121668042 on Build GRCh38
rs535874392 Chr.3: 121668017-121668017 on Build GRCh38
rs138791437 Chr.3: 121668315-121668315 on Build GRCh38
rs761235655 Chr.3: 121668252-121668252 on Build GRCh38
rs56109033 Chr.3: 121668213-121668213 on Build GRCh38
rs767945340 Chr.3: 121667969-121667969 on Build GRCh38
rs201193439 Chr.3: 121668083-121668083 on Build GRCh38
rs1026672421 Chr.3: 121668217-121668217 on Build GRCh38
rs748561642 Chr.3: 121668177-121668177 on Build GRCh38
rs758905646 Chr.3: 121668189-121668189 on Build GRCh38
rs544491760 Chr.3: 121668288-121668288 on Build GRCh38
rs755051134 Chr.3: 121668142-121668142 on Build GRCh38
rs776172927 Chr.3: 121668069-121668069 on Build GRCh38
rs766705145 Chr.3: 121668121-121668121 on Build GRCh38
rs763837909 Chr.3: 121668089-121668089 on Build GRCh38
rs748156123 Chr.3: 121668313-121668313 on Build GRCh38
rs867250568 Chr.3: 121668133-121668133 on Build GRCh38
rs544737213 Chr.3: 121668193-121668193 on Build GRCh38
rs1016468722 Chr.3: 121668071-121668071 on Build GRCh38
rs145580930 Chr.3: 121668203-121668203 on Build GRCh38
rs779496010 Chr.3: 121668156-121668156 on Build GRCh38
rs999500521 Chr.3: 121668233-121668233 on Build GRCh38
rs765160103 Chr.3: 121668076-121668076 on Build GRCh38
rs553004218 Chr.3: 121668070-121668070 on Build GRCh38
rs776196044 Chr.3: 121668202-121668202 on Build GRCh38
rs772275331 Chr.3: 121668044-121668044 on Build GRCh38
rs561430200 Chr.3: 121668304-121668304 on Build GRCh38
rs765349484 Chr.3: 121668006-121668006 on Build GRCh38
rs779495513 Chr.3: 121668040-121668040 on Build GRCh38
rs751349367 Chr.3: 121668095-121668095 on Build GRCh38
rs542920121 Chr.3: 121668294-121668294 on Build GRCh38
rs989425971 Chr.3: 121668335-121668335 on Build GRCh38
rs778046115 Chr.3: 121668190-121668190 on Build GRCh38
rs770545810 Chr.3: 121668053-121668053 on Build GRCh38
rs1013838792 Chr.3: 121668099-121668099 on Build GRCh38
rs780332325 Chr.3: 121668010-121668010 on Build GRCh38
rs760841645 Chr.3: 121668172-121668172 on Build GRCh38
rs753972405 Chr.3: 121668127-121668127 on Build GRCh38
rs747358478 Chr.3: 121668191-121668191 on Build GRCh38
rs1031026998 Chr.3: 121668249-121668249 on Build GRCh38
rs995408108 Chr.3: 121668215-121668215 on Build GRCh38
rs1012462535 Chr.3: 121668140-121668140 on Build GRCh38
rs958149899 Chr.3: 121668254-121668254 on Build GRCh38
rs772946964 Chr.3: 121668046-121668046 on Build GRCh38
rs755610551 Chr.3: 121668037-121668037 on Build GRCh38
rs901472570 Chr.3: 121668009-121668009 on Build GRCh38
rs746678875 Chr.3: 121668047-121668047 on Build GRCh38

View Full Product Details