Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123620126-123620633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYLK
Entrez Gene ID
4638
Gene Name
myosin light chain kinase
Gene Aliases
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
UniGene
Hs.477375
Gene Chromosome Location
Chr.3: 123612296-123884302 on Build GRCh38
Transcripts
NM_001321309.1 NM_053025.3 NM_053026.3 NM_053027.3 NM_053028.3 XM_011512860.2 XM_011512861.2 XM_017006469.1 XM_017006470.1 XM_017006471.1 XM_017006472.1 XM_017006473.1 NR_038266.2 NR_121654.1

Gene Symbol
MYLK-AS1
Entrez Gene ID
100506826
Gene Name
MYLK antisense RNA 1
Gene Aliases
-
UniGene
Hs.556600
Gene Chromosome Location
Chr.3: 123585513-123630821 on Build GRCh38
Transcripts
NM_001321309.1 NM_053025.3 NM_053026.3 NM_053027.3 NM_053028.3 XM_011512860.2 XM_011512861.2 XM_017006469.1 XM_017006470.1 XM_017006471.1 XM_017006472.1 XM_017006473.1 NR_038266.2 NR_121654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1616814 c.5328A>G p.S1776S Chr.3: 123620247-123620247 on Build GRCh38
3057132 c.5301C>A p.L1767L Chr.3: 123620274-123620274 on Build GRCh38
6719667 c.5329C>T p.P1777S Chr.3: 123620246-123620246 on Build GRCh38
3057135 c.5285C>T p.A1762V Chr.3: 123620290-123620290 on Build GRCh38
3057136 c.5243C>T p.T1748M Chr.3: 123620332-123620332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747903270 Chr.3: 123620454-123620454 on Build GRCh38
rs769749622 Chr.3: 123620326-123620326 on Build GRCh38
rs989310520 Chr.3: 123620409-123620409 on Build GRCh38
rs760272590 Chr.3: 123620167-123620170 on Build GRCh38
rs537640257 Chr.3: 123620487-123620487 on Build GRCh38
rs538629159 Chr.3: 123620390-123620390 on Build GRCh38
rs820456 Chr.3: 123620491-123620491 on Build GRCh38
rs149866482 Chr.3: 123620273-123620273 on Build GRCh38
rs763825048 Chr.3: 123620206-123620206 on Build GRCh38
rs974834451 Chr.3: 123620260-123620260 on Build GRCh38
rs758032144 Chr.3: 123620194-123620194 on Build GRCh38
rs763701684 Chr.3: 123620365-123620365 on Build GRCh38
rs749707023 Chr.3: 123620228-123620228 on Build GRCh38
rs1030340647 Chr.3: 123620589-123620589 on Build GRCh38
rs780478599 Chr.3: 123620227-123620227 on Build GRCh38
rs551204823 Chr.3: 123620207-123620207 on Build GRCh38
rs778168311 Chr.3: 123620393-123620393 on Build GRCh38
rs115869339 Chr.3: 123620495-123620495 on Build GRCh38
rs772211380 Chr.3: 123620264-123620264 on Build GRCh38
rs777749875 Chr.3: 123620515-123620515 on Build GRCh38
rs530450065 Chr.3: 123620270-123620270 on Build GRCh38
rs982281687 Chr.3: 123620457-123620457 on Build GRCh38
rs533809644 Chr.3: 123620509-123620509 on Build GRCh38
rs200088151 Chr.3: 123620236-123620236 on Build GRCh38
rs770585066 Chr.3: 123620294-123620294 on Build GRCh38
rs759542748 Chr.3: 123620165-123620165 on Build GRCh38
rs756479350 Chr.3: 123620222-123620222 on Build GRCh38
rs112539139 Chr.3: 123620571-123620571 on Build GRCh38
rs780455131 Chr.3: 123620331-123620331 on Build GRCh38
rs1046139896 Chr.3: 123620469-123620469 on Build GRCh38
rs776377711 Chr.3: 123620162-123620162 on Build GRCh38
rs1050924144 Chr.3: 123620503-123620503 on Build GRCh38
rs753392124 Chr.3: 123620196-123620198 on Build GRCh38
rs751197356 Chr.3: 123620371-123620371 on Build GRCh38
rs886819437 Chr.3: 123620518-123620518 on Build GRCh38
rs988521821 Chr.3: 123620485-123620485 on Build GRCh38
rs751323718 Chr.3: 123620216-123620216 on Build GRCh38
rs1012576678 Chr.3: 123620592-123620592 on Build GRCh38
rs779324323 Chr.3: 123620238-123620238 on Build GRCh38
rs112900417 Chr.3: 123620176-123620176 on Build GRCh38
rs1016427545 Chr.3: 123620508-123620508 on Build GRCh38
rs759291796 Chr.3: 123620311-123620311 on Build GRCh38
rs749676754 Chr.3: 123620417-123620417 on Build GRCh38
rs376908563 Chr.3: 123620379-123620379 on Build GRCh38
rs749176614 Chr.3: 123620564-123620564 on Build GRCh38
rs756607557 Chr.3: 123620384-123620384 on Build GRCh38
rs547331411 Chr.3: 123620295-123620295 on Build GRCh38
rs764614812 Chr.3: 123620170-123620170 on Build GRCh38
rs906288139 Chr.3: 123620481-123620481 on Build GRCh38
rs899384635 Chr.3: 123620382-123620382 on Build GRCh38
rs755343047 Chr.3: 123620386-123620386 on Build GRCh38
rs146990616 Chr.3: 123620186-123620194 on Build GRCh38
rs192655752 Chr.3: 123620496-123620496 on Build GRCh38
rs929597419 Chr.3: 123620488-123620488 on Build GRCh38
rs188032878 Chr.3: 123620352-123620352 on Build GRCh38
rs1038555951 Chr.3: 123620570-123620570 on Build GRCh38
rs748200926 Chr.3: 123620246-123620246 on Build GRCh38
rs536614981 Chr.3: 123620396-123620396 on Build GRCh38
rs775626666 Chr.3: 123620348-123620348 on Build GRCh38
rs747248257 Chr.3: 123620275-123620275 on Build GRCh38
rs372648070 Chr.3: 123620179-123620179 on Build GRCh38
rs761547459 Chr.3: 123620374-123620374 on Build GRCh38
rs387906781 Chr.3: 123620300-123620300 on Build GRCh38
rs568974320 Chr.3: 123620468-123620468 on Build GRCh38

View Full Product Details