Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 126704138-126704636 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD6
Entrez Gene ID
84303
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
CHCM1, Mic25, PPP1R23
UniGene
Hs.518119
Gene Chromosome Location
Chr.3: 126704220-126960420 on Build GRCh38
Transcripts
NM_001320610.1 NM_032343.2 XM_011513234.1 XM_011513235.2 XM_017007320.1 XM_017007321.1 XM_017007322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7229904 c.12G>A p.T4T Chr.3: 126704324-126704324 on Build GRCh38
4138404 c.18C>T p.S6S Chr.3: 126704330-126704330 on Build GRCh38
5834532 c.3delG p.S3fs*33 Chr.3: 126704315-126704315 on Build GRCh38
6163305 c.1A>C p.M1L Chr.3: 126704313-126704313 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs997013602 Chr.3: 126704253-126704253 on Build GRCh38
rs937948353 Chr.3: 126704261-126704261 on Build GRCh38
rs934485898 Chr.3: 126704185-126704185 on Build GRCh38
rs760672239 Chr.3: 126704396-126704396 on Build GRCh38
rs760494837 Chr.3: 126704529-126704529 on Build GRCh38
rs939760851 Chr.3: 126704274-126704274 on Build GRCh38
rs75098676 Chr.3: 126704439-126704439 on Build GRCh38
rs764761455 Chr.3: 126704291-126704291 on Build GRCh38
rs906997584 Chr.3: 126704182-126704182 on Build GRCh38
rs768753428 Chr.3: 126704305-126704305 on Build GRCh38
rs768047544 Chr.3: 126704425-126704425 on Build GRCh38
rs749577943 Chr.3: 126704303-126704303 on Build GRCh38
rs988203128 Chr.3: 126704225-126704225 on Build GRCh38
rs765810624 Chr.3: 126704280-126704280 on Build GRCh38
rs774457222 Chr.3: 126704370-126704370 on Build GRCh38
rs1036065677 Chr.3: 126704506-126704506 on Build GRCh38
rs749824707 Chr.3: 126704313-126704313 on Build GRCh38
rs917549191 Chr.3: 126704404-126704404 on Build GRCh38
rs757729922 Chr.3: 126704344-126704344 on Build GRCh38
rs6804225 Chr.3: 126704270-126704270 on Build GRCh38
rs1049232246 Chr.3: 126704447-126704447 on Build GRCh38
rs775651098 Chr.3: 126704297-126704297 on Build GRCh38
rs1023169382 Chr.3: 126704475-126704475 on Build GRCh38
rs771304781 Chr.3: 126704256-126704256 on Build GRCh38
rs4679130 Chr.3: 126704594-126704594 on Build GRCh38
rs1037291382 Chr.3: 126704453-126704453 on Build GRCh38
rs386665618 Chr.3: 126704269-126704270 on Build GRCh38
rs375938308 Chr.3: 126704405-126704405 on Build GRCh38
rs1011299244 Chr.3: 126704241-126704241 on Build GRCh38
rs569316621 Chr.3: 126704222-126704222 on Build GRCh38
rs1017193708 Chr.3: 126704243-126704243 on Build GRCh38
rs541790341 Chr.3: 126704284-126704284 on Build GRCh38
rs1018925500 Chr.3: 126704318-126704318 on Build GRCh38
rs13070920 Chr.3: 126704473-126704473 on Build GRCh38
rs748360128 Chr.3: 126704374-126704374 on Build GRCh38
rs755642296 Chr.3: 126704279-126704279 on Build GRCh38
rs773163792 Chr.3: 126704323-126704323 on Build GRCh38
rs202222271 Chr.3: 126704389-126704389 on Build GRCh38
rs371171901 Chr.3: 126704262-126704262 on Build GRCh38
rs747308398 Chr.3: 126704292-126704292 on Build GRCh38
rs375259651 Chr.3: 126704296-126704296 on Build GRCh38
rs780344521 Chr.3: 126704357-126704357 on Build GRCh38
rs745442418 Chr.3: 126704482-126704482 on Build GRCh38
rs568339846 Chr.3: 126704468-126704468 on Build GRCh38
rs371353425 Chr.3: 126704224-126704224 on Build GRCh38
rs1007542096 Chr.3: 126704168-126704168 on Build GRCh38
rs767249214 Chr.3: 126704320-126704320 on Build GRCh38
rs967863416 Chr.3: 126704248-126704248 on Build GRCh38
rs752348716 Chr.3: 126704341-126704341 on Build GRCh38
rs752301163 Chr.3: 126704419-126704419 on Build GRCh38
rs573103239 Chr.3: 126704264-126704264 on Build GRCh38
rs756724510 Chr.3: 126704448-126704448 on Build GRCh38
rs746340185 Chr.3: 126704295-126704295 on Build GRCh38
rs753284418 Chr.3: 126704326-126704326 on Build GRCh38
rs907495030 Chr.3: 126704579-126704579 on Build GRCh38
rs956068132 Chr.3: 126704515-126704515 on Build GRCh38
rs750810359 Chr.3: 126704428-126704428 on Build GRCh38
rs529207043 Chr.3: 126704560-126704560 on Build GRCh38
rs1033958900 Chr.3: 126704354-126704354 on Build GRCh38
rs200360858 Chr.3: 126704293-126704293 on Build GRCh38
rs959470896 Chr.3: 126704525-126704525 on Build GRCh38
rs148129437 Chr.3: 126704324-126704324 on Build GRCh38
rs539586754 Chr.3: 126704223-126704223 on Build GRCh38
rs368894266 Chr.3: 126704406-126704406 on Build GRCh38
rs913698716 Chr.3: 126704242-126704242 on Build GRCh38
rs992055057 Chr.3: 126704545-126704545 on Build GRCh38
rs779766981 Chr.3: 126704364-126704366 on Build GRCh38
rs902128866 Chr.3: 126704462-126704462 on Build GRCh38
rs934624522 Chr.3: 126704289-126704289 on Build GRCh38
rs142017260 Chr.3: 126704349-126704349 on Build GRCh38
rs557924807 Chr.3: 126704230-126704230 on Build GRCh38
rs1033492325 Chr.3: 126704521-126704521 on Build GRCh38
rs770458773 Chr.3: 126704449-126704449 on Build GRCh38
rs933044380 Chr.3: 126704563-126704563 on Build GRCh38
rs771881879 Chr.3: 126704504-126704504 on Build GRCh38
rs370664603 Chr.3: 126704294-126704294 on Build GRCh38
rs931081405 Chr.3: 126704430-126704430 on Build GRCh38
rs6804224 Chr.3: 126704269-126704269 on Build GRCh38
rs377567356 Chr.3: 126704310-126704310 on Build GRCh38
rs545968757 Chr.3: 126704332-126704332 on Build GRCh38
rs564958440 Chr.3: 126704519-126704519 on Build GRCh38
rs745802050 Chr.3: 126704299-126704299 on Build GRCh38
rs756671511 Chr.3: 126704352-126704352 on Build GRCh38
rs974528749 Chr.3: 126704531-126704531 on Build GRCh38
rs768971697 Chr.3: 126704364-126704364 on Build GRCh38
rs903933281 Chr.3: 126704499-126704499 on Build GRCh38
rs200230339 Chr.3: 126704282-126704282 on Build GRCh38
rs778576409 Chr.3: 126704450-126704450 on Build GRCh38
rs771868269 Chr.3: 126704388-126704388 on Build GRCh38
rs748405239 Chr.3: 126704397-126704404 on Build GRCh38
rs979270249 Chr.3: 126704255-126704255 on Build GRCh38
rs1002971097 Chr.3: 126704484-126704484 on Build GRCh38
rs765933839 Chr.3: 126704325-126704325 on Build GRCh38
rs762633231 Chr.3: 126704421-126704421 on Build GRCh38
rs769865726 Chr.3: 126704446-126704447 on Build GRCh38
rs972379671 Chr.3: 126704567-126704567 on Build GRCh38
rs767410739 Chr.3: 126704275-126704275 on Build GRCh38
rs749981701 Chr.3: 126704278-126704278 on Build GRCh38
rs758994949 Chr.3: 126704412-126704412 on Build GRCh38
rs987333915 Chr.3: 126704192-126704192 on Build GRCh38
rs369985851 Chr.3: 126704585-126704585 on Build GRCh38
rs150685996 Chr.3: 126704350-126704350 on Build GRCh38
rs557104503 Chr.3: 126704221-126704221 on Build GRCh38
rs368488136 Chr.3: 126704307-126704307 on Build GRCh38
rs771913044 Chr.3: 126704359-126704360 on Build GRCh38
rs749295325 Chr.3: 126704363-126704363 on Build GRCh38
rs961323329 Chr.3: 126704565-126704565 on Build GRCh38
rs1019394758 Chr.3: 126704181-126704181 on Build GRCh38

View Full Product Details