Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 127608748-127609256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM2
Entrez Gene ID
4171
Gene Name
minichromosome maintenance complex component 2
Gene Aliases
BM28, CCNL1, CDCL1, D3S3194, DFNA70, MITOTIN, cdc19
UniGene
Hs.477481
Gene Chromosome Location
Chr.3: 127598357-127622436 on Build GRCh38
Transcripts
NM_004526.3 NR_073375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5695470 c.1423G>A p.E475K Chr.3: 127609018-127609018 on Build GRCh38
7201904 c.1348A>C p.K450Q Chr.3: 127608943-127608943 on Build GRCh38
4463728 c.1302C>T p.V434V Chr.3: 127608897-127608897 on Build GRCh38
4969871 c.1373_1377delATGAA p.D458fs*7 Chr.3: 127608968-127608972 on Build GRCh38
5937096 c.1297C>T p.P433S Chr.3: 127608892-127608892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755437070 Chr.3: 127608885-127608885 on Build GRCh38
rs368920619 Chr.3: 127609070-127609070 on Build GRCh38
rs1031272484 Chr.3: 127608813-127608813 on Build GRCh38
rs755893464 Chr.3: 127609035-127609035 on Build GRCh38
rs965448999 Chr.3: 127609003-127609003 on Build GRCh38
rs551058916 Chr.3: 127609069-127609069 on Build GRCh38
rs751895936 Chr.3: 127609012-127609012 on Build GRCh38
rs187407254 Chr.3: 127608834-127608834 on Build GRCh38
rs780884057 Chr.3: 127609232-127609232 on Build GRCh38
rs746832546 Chr.3: 127608794-127608794 on Build GRCh38
rs757423043 Chr.3: 127609014-127609014 on Build GRCh38
rs757173924 Chr.3: 127608920-127608920 on Build GRCh38
rs891420872 Chr.3: 127609229-127609229 on Build GRCh38
rs886636939 Chr.3: 127609234-127609234 on Build GRCh38
rs939420001 Chr.3: 127609095-127609095 on Build GRCh38
rs770819139 Chr.3: 127608795-127608795 on Build GRCh38
rs201123662 Chr.3: 127608877-127608877 on Build GRCh38
rs754559800 Chr.3: 127609173-127609173 on Build GRCh38
rs147725704 Chr.3: 127608873-127608873 on Build GRCh38
rs750792235 Chr.3: 127608846-127608846 on Build GRCh38
rs530548856 Chr.3: 127608777-127608777 on Build GRCh38
rs762387649 Chr.3: 127608862-127608862 on Build GRCh38
rs140620010 Chr.3: 127608999-127608999 on Build GRCh38
rs372976124 Chr.3: 127608913-127608913 on Build GRCh38
rs545743607 Chr.3: 127608792-127608792 on Build GRCh38
rs930914189 Chr.3: 127609198-127609198 on Build GRCh38
rs764254320 Chr.3: 127609006-127609006 on Build GRCh38
rs1003941996 Chr.3: 127608997-127609004 on Build GRCh38
rs763762735 Chr.3: 127608863-127608863 on Build GRCh38
rs778180034 Chr.3: 127608900-127608900 on Build GRCh38
rs975351069 Chr.3: 127609004-127609004 on Build GRCh38
rs956097023 Chr.3: 127609165-127609165 on Build GRCh38
rs765709610 Chr.3: 127608888-127608888 on Build GRCh38
rs748832537 Chr.3: 127608946-127608946 on Build GRCh38
rs1056591400 Chr.3: 127609124-127609124 on Build GRCh38
rs780010455 Chr.3: 127609041-127609041 on Build GRCh38
rs765714001 Chr.3: 127609033-127609034 on Build GRCh38
rs753224670 Chr.3: 127608993-127608993 on Build GRCh38
rs895836634 Chr.3: 127608827-127608827 on Build GRCh38
rs372110727 Chr.3: 127608884-127608884 on Build GRCh38
rs758512953 Chr.3: 127608892-127608892 on Build GRCh38
rs904130392 Chr.3: 127609174-127609174 on Build GRCh38
rs1028617131 Chr.3: 127609043-127609043 on Build GRCh38
rs752838407 Chr.3: 127608891-127608891 on Build GRCh38
rs745669398 Chr.3: 127609015-127609015 on Build GRCh38
rs992948683 Chr.3: 127609172-127609172 on Build GRCh38
rs1052725623 Chr.3: 127609194-127609194 on Build GRCh38
rs916643119 Chr.3: 127609142-127609142 on Build GRCh38
rs948275843 Chr.3: 127609158-127609158 on Build GRCh38
rs1029825664 Chr.3: 127608975-127608975 on Build GRCh38
rs749203888 Chr.3: 127609048-127609048 on Build GRCh38
rs566852393 Chr.3: 127609189-127609189 on Build GRCh38
rs775445540 Chr.3: 127608929-127608929 on Build GRCh38
rs776680860 Chr.3: 127608804-127608804 on Build GRCh38
rs924465412 Chr.3: 127609111-127609112 on Build GRCh38
rs376973485 Chr.3: 127608967-127608967 on Build GRCh38
rs759887747 Chr.3: 127608981-127608981 on Build GRCh38
rs745777900 Chr.3: 127608829-127608829 on Build GRCh38
rs534125090 Chr.3: 127609195-127609195 on Build GRCh38
rs780672663 Chr.3: 127609071-127609071 on Build GRCh38
rs182603137 Chr.3: 127608988-127608988 on Build GRCh38
rs796922472 Chr.3: 127608907-127608907 on Build GRCh38
rs371208727 Chr.3: 127608869-127608869 on Build GRCh38
rs142779979 Chr.3: 127609187-127609187 on Build GRCh38
rs747454576 Chr.3: 127609129-127609129 on Build GRCh38
rs199539311 Chr.3: 127608925-127608925 on Build GRCh38
rs768216329 Chr.3: 127609066-127609066 on Build GRCh38
rs918559048 Chr.3: 127609094-127609094 on Build GRCh38
rs766835318 Chr.3: 127608976-127608976 on Build GRCh38
rs369473650 Chr.3: 127608871-127608871 on Build GRCh38
rs776866004 Chr.3: 127608977-127608977 on Build GRCh38
rs539837474 Chr.3: 127609126-127609126 on Build GRCh38
rs569132309 Chr.3: 127609089-127609089 on Build GRCh38
rs780841951 Chr.3: 127608924-127608924 on Build GRCh38
rs531775518 Chr.3: 127608875-127608875 on Build GRCh38
rs142512970 Chr.3: 127608966-127608966 on Build GRCh38
rs972742027 Chr.3: 127609077-127609077 on Build GRCh38
rs904089711 Chr.3: 127608776-127608776 on Build GRCh38
rs190988564 Chr.3: 127608982-127608982 on Build GRCh38
rs962597089 Chr.3: 127609076-127609076 on Build GRCh38

View Full Product Details