Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129061646-129062145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GP9
Entrez Gene ID
2815
Gene Name
glycoprotein IX platelet
Gene Aliases
CD42a, GPIX
UniGene
Hs.1144
Gene Chromosome Location
Chr.3: 129055449-129062411 on Build GRCh38
Transcripts
NM_000174.4 XM_005247374.3 XM_011512701.1 XM_011512702.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4535043 c.214G>A p.D72N Chr.3: 129061953-129061953 on Build GRCh38
7252409 c.132G>A p.T44T Chr.3: 129061871-129061871 on Build GRCh38
4687704 c.265T>A p.C89S Chr.3: 129062004-129062004 on Build GRCh38
1208506 c.335G>A p.R112H Chr.3: 129062074-129062074 on Build GRCh38
4474888 c.194C>T p.S65F Chr.3: 129061933-129061933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775994327 Chr.3: 129061899-129061899 on Build GRCh38
rs768351451 Chr.3: 129061795-129061795 on Build GRCh38
rs200640594 Chr.3: 129061975-129061975 on Build GRCh38
rs755371475 Chr.3: 129062109-129062109 on Build GRCh38
rs768867472 Chr.3: 129061905-129061905 on Build GRCh38
rs780722740 Chr.3: 129061948-129061948 on Build GRCh38
rs780057910 Chr.3: 129062014-129062014 on Build GRCh38
rs369980915 Chr.3: 129061918-129061918 on Build GRCh38
rs747209258 Chr.3: 129062117-129062117 on Build GRCh38
rs771201708 Chr.3: 129062122-129062122 on Build GRCh38
rs756853190 Chr.3: 129062047-129062047 on Build GRCh38
rs188407487 Chr.3: 129061670-129061670 on Build GRCh38
rs758791695 Chr.3: 129062115-129062115 on Build GRCh38
rs764919230 Chr.3: 129062028-129062028 on Build GRCh38
rs28933378 Chr.3: 129061809-129061809 on Build GRCh38
rs749784464 Chr.3: 129062051-129062051 on Build GRCh38
rs148622572 Chr.3: 129062042-129062042 on Build GRCh38
rs201755437 Chr.3: 129061862-129061862 on Build GRCh38
rs201745505 Chr.3: 129061699-129061699 on Build GRCh38
rs138811539 Chr.3: 129061894-129061894 on Build GRCh38
rs763255801 Chr.3: 129061847-129061847 on Build GRCh38
rs140929300 Chr.3: 129061841-129061841 on Build GRCh38
rs761917144 Chr.3: 129061913-129061913 on Build GRCh38
rs779861329 Chr.3: 129061763-129061763 on Build GRCh38
rs767469270 Chr.3: 129061914-129061914 on Build GRCh38
rs746730389 Chr.3: 129062016-129062016 on Build GRCh38
rs962839753 Chr.3: 129061864-129061864 on Build GRCh38
rs755563532 Chr.3: 129062054-129062054 on Build GRCh38
rs753000146 Chr.3: 129061725-129061725 on Build GRCh38
rs750293970 Chr.3: 129061870-129061870 on Build GRCh38
rs752262357 Chr.3: 129061935-129061935 on Build GRCh38
rs556468076 Chr.3: 129062102-129062102 on Build GRCh38
rs753570637 Chr.3: 129061926-129061926 on Build GRCh38
rs112163054 Chr.3: 129061683-129061683 on Build GRCh38
rs895691026 Chr.3: 129061792-129061792 on Build GRCh38
rs201341449 Chr.3: 129062068-129062068 on Build GRCh38
rs376032777 Chr.3: 129062126-129062126 on Build GRCh38
rs773906187 Chr.3: 129061804-129061804 on Build GRCh38
rs751593201 Chr.3: 129061858-129061858 on Build GRCh38
rs142692011 Chr.3: 129061988-129061988 on Build GRCh38
rs992855691 Chr.3: 129061919-129061919 on Build GRCh38
rs775081774 Chr.3: 129062090-129062090 on Build GRCh38
rs550480973 Chr.3: 129061889-129061889 on Build GRCh38
rs923137982 Chr.3: 129061991-129061991 on Build GRCh38
rs61732506 Chr.3: 129062114-129062114 on Build GRCh38
rs745621166 Chr.3: 129061756-129061756 on Build GRCh38
rs200376043 Chr.3: 129061757-129061757 on Build GRCh38
rs763418097 Chr.3: 129061900-129061900 on Build GRCh38
rs776687574 Chr.3: 129062124-129062124 on Build GRCh38
rs6069 Chr.3: 129061871-129061871 on Build GRCh38
rs764247973 Chr.3: 129061738-129061738 on Build GRCh38
rs921030356 Chr.3: 129061916-129061916 on Build GRCh38
rs772126176 Chr.3: 129062058-129062058 on Build GRCh38
rs773442662 Chr.3: 129062060-129062060 on Build GRCh38
rs750557788 Chr.3: 129061915-129061915 on Build GRCh38
rs137994346 Chr.3: 129061980-129061980 on Build GRCh38
rs573091452 Chr.3: 129061735-129061735 on Build GRCh38
rs771512899 Chr.3: 129061891-129061891 on Build GRCh38
rs5030764 Chr.3: 129061921-129061921 on Build GRCh38
rs770361254 Chr.3: 129061945-129061945 on Build GRCh38
rs772819137 Chr.3: 129061893-129061893 on Build GRCh38
rs121918037 Chr.3: 129061951-129061951 on Build GRCh38
rs121918036 Chr.3: 129061849-129061849 on Build GRCh38
rs757217460 Chr.3: 129061744-129061744 on Build GRCh38
rs983071890 Chr.3: 129061878-129061878 on Build GRCh38
rs761923868 Chr.3: 129061977-129061977 on Build GRCh38
rs745846700 Chr.3: 129061987-129061987 on Build GRCh38
rs61734182 Chr.3: 129062113-129062113 on Build GRCh38
rs754789523 Chr.3: 129061934-129061934 on Build GRCh38
rs777573986 Chr.3: 129062045-129062045 on Build GRCh38
rs121918038 Chr.3: 129061759-129061759 on Build GRCh38
rs750011732 Chr.3: 129062108-129062108 on Build GRCh38
rs889759228 Chr.3: 129062037-129062037 on Build GRCh38
rs749625028 Chr.3: 129061950-129061950 on Build GRCh38
rs759941195 Chr.3: 129061821-129061821 on Build GRCh38
rs887132252 Chr.3: 129062081-129062081 on Build GRCh38
rs200489927 Chr.3: 129062073-129062073 on Build GRCh38
rs768833037 Chr.3: 129062116-129062116 on Build GRCh38
rs865852486 Chr.3: 129061996-129061996 on Build GRCh38
rs779138158 Chr.3: 129062055-129062055 on Build GRCh38
rs770152913 Chr.3: 129062093-129062093 on Build GRCh38
rs751649007 Chr.3: 129061739-129061739 on Build GRCh38
rs748912527 Chr.3: 129061780-129061780 on Build GRCh38
rs763793634 Chr.3: 129062096-129062096 on Build GRCh38
rs960932963 Chr.3: 129061880-129061880 on Build GRCh38
rs767444247 Chr.3: 129061863-129061863 on Build GRCh38
rs896687623 Chr.3: 129062080-129062080 on Build GRCh38
rs747051704 Chr.3: 129062066-129062066 on Build GRCh38
rs758119211 Chr.3: 129061938-129061938 on Build GRCh38
rs145472050 Chr.3: 129061942-129061942 on Build GRCh38
rs28933377 Chr.3: 129061906-129061906 on Build GRCh38
rs765666170 Chr.3: 129061838-129061838 on Build GRCh38
rs1024317986 Chr.3: 129062110-129062110 on Build GRCh38
rs377254159 Chr.3: 129061820-129061820 on Build GRCh38
rs368116821 Chr.3: 129062094-129062094 on Build GRCh38
rs780671537 Chr.3: 129062050-129062050 on Build GRCh38
rs901787643 Chr.3: 129061773-129061773 on Build GRCh38
rs765287859 Chr.3: 129062074-129062074 on Build GRCh38
rs146025958 Chr.3: 129062029-129062029 on Build GRCh38
rs889822586 Chr.3: 129061857-129061857 on Build GRCh38
rs781001089 Chr.3: 129061748-129061748 on Build GRCh38
rs141151793 Chr.3: 129062057-129062057 on Build GRCh38
rs779808233 Chr.3: 129061886-129061886 on Build GRCh38
rs766253334 Chr.3: 129061998-129061998 on Build GRCh38
rs749144595 Chr.3: 129061888-129061888 on Build GRCh38
rs555017924 Chr.3: 129062046-129062046 on Build GRCh38
rs147675904 Chr.3: 129061819-129061819 on Build GRCh38
rs756744791 Chr.3: 129061671-129061671 on Build GRCh38
rs202229101 Chr.3: 129062107-129062107 on Build GRCh38
rs764292260 Chr.3: 129061851-129061851 on Build GRCh38
rs1051092529 Chr.3: 129061831-129061831 on Build GRCh38
rs373723419 Chr.3: 129062059-129062059 on Build GRCh38
rs759829973 Chr.3: 129061694-129061694 on Build GRCh38

View Full Product Details