Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130690383-130690881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R4
Entrez Gene ID
30849
Gene Name
phosphoinositide-3-kinase regulatory subunit 4
Gene Aliases
VPS15, p150
UniGene
Hs.149032
Gene Chromosome Location
Chr.3: 130678934-130746852 on Build GRCh38
Transcripts
NM_014602.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
98950 c.3103A>G p.I1035V Chr.3: 130690650-130690650 on Build GRCh38
5863513 c.3118C>T p.R1040* Chr.3: 130690635-130690635 on Build GRCh38
7141967 c.3144C>A p.L1048L Chr.3: 130690609-130690609 on Build GRCh38
290826 c.3144C>G p.L1048L Chr.3: 130690609-130690609 on Build GRCh38
5886294 c.3239C>T p.P1080L Chr.3: 130690514-130690514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558980604 Chr.3: 130690798-130690798 on Build GRCh38
rs779104744 Chr.3: 130690619-130690619 on Build GRCh38
rs763720763 Chr.3: 130690658-130690658 on Build GRCh38
rs753712561 Chr.3: 130690534-130690534 on Build GRCh38
rs199700522 Chr.3: 130690481-130690481 on Build GRCh38
rs751727600 Chr.3: 130690443-130690447 on Build GRCh38
rs56160735 Chr.3: 130690625-130690625 on Build GRCh38
rs760751940 Chr.3: 130690628-130690628 on Build GRCh38
rs777458805 Chr.3: 130690597-130690597 on Build GRCh38
rs375119113 Chr.3: 130690460-130690460 on Build GRCh38
rs937044315 Chr.3: 130690740-130690759 on Build GRCh38
rs780000804 Chr.3: 130690455-130690455 on Build GRCh38
rs776127275 Chr.3: 130690458-130690458 on Build GRCh38
rs756822733 Chr.3: 130690611-130690611 on Build GRCh38
rs1043630104 Chr.3: 130690833-130690833 on Build GRCh38
rs554897210 Chr.3: 130690831-130690831 on Build GRCh38
rs1006867506 Chr.3: 130690812-130690812 on Build GRCh38
rs944055794 Chr.3: 130690804-130690804 on Build GRCh38
rs985398552 Chr.3: 130690462-130690462 on Build GRCh38
rs1016871592 Chr.3: 130690846-130690846 on Build GRCh38
rs776572857 Chr.3: 130690635-130690635 on Build GRCh38
rs962551755 Chr.3: 130690487-130690487 on Build GRCh38
rs759431388 Chr.3: 130690636-130690636 on Build GRCh38
rs769162853 Chr.3: 130690616-130690616 on Build GRCh38
rs770940364 Chr.3: 130690634-130690634 on Build GRCh38
rs760798247 Chr.3: 130690513-130690513 on Build GRCh38
rs374424679 Chr.3: 130690702-130690702 on Build GRCh38
rs147413868 Chr.3: 130690612-130690612 on Build GRCh38
rs752428407 Chr.3: 130690588-130690588 on Build GRCh38
rs755247236 Chr.3: 130690723-130690723 on Build GRCh38
rs1047055240 Chr.3: 130690790-130690790 on Build GRCh38
rs903589976 Chr.3: 130690841-130690841 on Build GRCh38
rs998310592 Chr.3: 130690785-130690785 on Build GRCh38
rs936169624 Chr.3: 130690684-130690684 on Build GRCh38
rs755579347 Chr.3: 130690680-130690680 on Build GRCh38
rs552536210 Chr.3: 130690594-130690594 on Build GRCh38
rs1041577082 Chr.3: 130690529-130690529 on Build GRCh38
rs780796163 Chr.3: 130690668-130690668 on Build GRCh38
rs749751291 Chr.3: 130690613-130690613 on Build GRCh38
rs770471371 Chr.3: 130690442-130690442 on Build GRCh38
rs575354758 Chr.3: 130690751-130690751 on Build GRCh38
rs139814973 Chr.3: 130690711-130690711 on Build GRCh38
rs891950844 Chr.3: 130690713-130690713 on Build GRCh38
rs900109287 Chr.3: 130690812-130690813 on Build GRCh38
rs754781434 Chr.3: 130690552-130690552 on Build GRCh38
rs889809179 Chr.3: 130690803-130690803 on Build GRCh38
rs997011766 Chr.3: 130690832-130690832 on Build GRCh38
rs981039493 Chr.3: 130690750-130690750 on Build GRCh38
rs766374049 Chr.3: 130690527-130690527 on Build GRCh38
rs748527435 Chr.3: 130690622-130690622 on Build GRCh38
rs200211626 Chr.3: 130690660-130690660 on Build GRCh38
rs767663289 Chr.3: 130690486-130690486 on Build GRCh38
rs200778585 Chr.3: 130690577-130690577 on Build GRCh38
rs376477491 Chr.3: 130690659-130690659 on Build GRCh38
rs769072698 Chr.3: 130690471-130690471 on Build GRCh38
rs373328532 Chr.3: 130690670-130690670 on Build GRCh38
rs17692351 Chr.3: 130690787-130690787 on Build GRCh38
rs200448951 Chr.3: 130690695-130690695 on Build GRCh38
rs143085620 Chr.3: 130690589-130690589 on Build GRCh38
rs779465049 Chr.3: 130690692-130690692 on Build GRCh38
rs762793792 Chr.3: 130690656-130690656 on Build GRCh38
rs147350320 Chr.3: 130690650-130690650 on Build GRCh38
rs772242905 Chr.3: 130690623-130690623 on Build GRCh38
rs1008488722 Chr.3: 130690438-130690438 on Build GRCh38
rs972561057 Chr.3: 130690498-130690498 on Build GRCh38
rs779979292 Chr.3: 130690649-130690649 on Build GRCh38
rs56006483 Chr.3: 130690564-130690564 on Build GRCh38
rs773216329 Chr.3: 130690500-130690500 on Build GRCh38
rs761889390 Chr.3: 130690485-130690485 on Build GRCh38

View Full Product Details