Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 133824158-133824659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB6B
Entrez Gene ID
51560
Gene Name
RAB6B, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.715344
Gene Chromosome Location
Chr.3: 133824235-133895847 on Build GRCh38
Transcripts
NM_016577.3 XM_011512893.2

SNP Detail

SNP ID Chromosome Location
rs963867160 Chr.3: 133824426-133824426 on Build GRCh38
rs1010653521 Chr.3: 133824316-133824316 on Build GRCh38
rs936061031 Chr.3: 133824189-133824189 on Build GRCh38
rs534150145 Chr.3: 133824589-133824589 on Build GRCh38
rs372219760 Chr.3: 133824440-133824440 on Build GRCh38
rs761947140 Chr.3: 133824506-133824506 on Build GRCh38
rs1010457597 Chr.3: 133824531-133824531 on Build GRCh38
rs1044826746 Chr.3: 133824502-133824502 on Build GRCh38
rs1006958030 Chr.3: 133824565-133824565 on Build GRCh38
rs936149154 Chr.3: 133824513-133824513 on Build GRCh38
rs1000481391 Chr.3: 133824186-133824186 on Build GRCh38
rs962510837 Chr.3: 133824374-133824374 on Build GRCh38
rs373331036 Chr.3: 133824401-133824401 on Build GRCh38
rs898077633 Chr.3: 133824247-133824247 on Build GRCh38
rs951394709 Chr.3: 133824617-133824617 on Build GRCh38
rs1014419334 Chr.3: 133824197-133824197 on Build GRCh38
rs188045253 Chr.3: 133824594-133824594 on Build GRCh38
rs929876756 Chr.3: 133824408-133824408 on Build GRCh38
rs575165400 Chr.3: 133824613-133824614 on Build GRCh38
rs962729316 Chr.3: 133824586-133824586 on Build GRCh38
rs955467925 Chr.3: 133824325-133824325 on Build GRCh38
rs759719961 Chr.3: 133824384-133824385 on Build GRCh38
rs771027334 Chr.3: 133824553-133824554 on Build GRCh38
rs768694703 Chr.3: 133824210-133824210 on Build GRCh38
rs891108208 Chr.3: 133824557-133824557 on Build GRCh38
rs368004495 Chr.3: 133824208-133824208 on Build GRCh38
rs992072823 Chr.3: 133824623-133824623 on Build GRCh38
rs977827408 Chr.3: 133824510-133824510 on Build GRCh38
rs76485805 Chr.3: 133824524-133824524 on Build GRCh38
rs115525603 Chr.3: 133824530-133824530 on Build GRCh38
rs531635539 Chr.3: 133824438-133824438 on Build GRCh38
rs1046038043 Chr.3: 133824217-133824218 on Build GRCh38
rs571653470 Chr.3: 133824546-133824546 on Build GRCh38
rs922553316 Chr.3: 133824453-133824453 on Build GRCh38
rs974056924 Chr.3: 133824385-133824385 on Build GRCh38
rs537993788 Chr.3: 133824439-133824439 on Build GRCh38
rs60912492 Chr.3: 133824398-133824398 on Build GRCh38
rs1018311566 Chr.3: 133824578-133824578 on Build GRCh38
rs183164063 Chr.3: 133824329-133824329 on Build GRCh38
rs1029628986 Chr.3: 133824317-133824317 on Build GRCh38
rs891832407 Chr.3: 133824261-133824261 on Build GRCh38
rs967284365 Chr.3: 133824493-133824494 on Build GRCh38
rs898089358 Chr.3: 133824627-133824628 on Build GRCh38
rs1018086284 Chr.3: 133824340-133824340 on Build GRCh38
rs1055913670 Chr.3: 133824537-133824537 on Build GRCh38
rs569094614 Chr.3: 133824191-133824192 on Build GRCh38
rs1025448793 Chr.3: 133824614-133824614 on Build GRCh38
rs1054589168 Chr.3: 133824525-133824525 on Build GRCh38
rs894617760 Chr.3: 133824192-133824192 on Build GRCh38
rs992249815 Chr.3: 133824410-133824410 on Build GRCh38

View Full Product Details