Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138030781-138031283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN18
Entrez Gene ID
51208
Gene Name
claudin 18
Gene Aliases
SFTA5, SFTPJ
UniGene
Hs.655324
Gene Chromosome Location
Chr.3: 137998816-138033652 on Build GRCh38
Transcripts
NM_001002026.2 NM_016369.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5459024 c.736C>T p.R246C Chr.3: 138031091-138031091 on Build GRCh38
5580627 c.742G>A p.E248K Chr.3: 138031097-138031097 on Build GRCh38
5355436 c.724G>A p.D242N Chr.3: 138031079-138031079 on Build GRCh38
7263327 c.642A>G p.S214S Chr.3: 138030997-138030997 on Build GRCh38
6629987 c.737G>A p.R246H Chr.3: 138031092-138031092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961525937 Chr.3: 138031000-138031000 on Build GRCh38
rs781530876 Chr.3: 138030914-138030914 on Build GRCh38
rs964701497 Chr.3: 138031143-138031143 on Build GRCh38
rs556733884 Chr.3: 138031116-138031116 on Build GRCh38
rs999860873 Chr.3: 138030909-138030909 on Build GRCh38
rs374671741 Chr.3: 138031123-138031123 on Build GRCh38
rs775385710 Chr.3: 138031106-138031106 on Build GRCh38
rs1011461500 Chr.3: 138030896-138030896 on Build GRCh38
rs919906363 Chr.3: 138031007-138031007 on Build GRCh38
rs998908475 Chr.3: 138030868-138030868 on Build GRCh38
rs55923693 Chr.3: 138031214-138031214 on Build GRCh38
rs971943909 Chr.3: 138031086-138031086 on Build GRCh38
rs147152301 Chr.3: 138030845-138030845 on Build GRCh38
rs777364119 Chr.3: 138031154-138031154 on Build GRCh38
rs376386414 Chr.3: 138031083-138031083 on Build GRCh38
rs564632238 Chr.3: 138030849-138030849 on Build GRCh38
rs770153777 Chr.3: 138031148-138031148 on Build GRCh38
rs931601165 Chr.3: 138030834-138030834 on Build GRCh38
rs141484279 Chr.3: 138031079-138031079 on Build GRCh38
rs201892247 Chr.3: 138031177-138031177 on Build GRCh38
rs754216040 Chr.3: 138030854-138030854 on Build GRCh38
rs199985015 Chr.3: 138031166-138031166 on Build GRCh38
rs779996032 Chr.3: 138031176-138031176 on Build GRCh38
rs761882973 Chr.3: 138031124-138031124 on Build GRCh38
rs771871196 Chr.3: 138031175-138031175 on Build GRCh38
rs922555406 Chr.3: 138031183-138031183 on Build GRCh38
rs771962732 Chr.3: 138031104-138031104 on Build GRCh38
rs543309126 Chr.3: 138031261-138031261 on Build GRCh38
rs756927400 Chr.3: 138031163-138031163 on Build GRCh38
rs576521499 Chr.3: 138031162-138031162 on Build GRCh38
rs754026762 Chr.3: 138031016-138031016 on Build GRCh38
rs966031562 Chr.3: 138031239-138031239 on Build GRCh38
rs574690650 Chr.3: 138031259-138031259 on Build GRCh38
rs372664208 Chr.3: 138031167-138031167 on Build GRCh38
rs752075988 Chr.3: 138031136-138031136 on Build GRCh38
rs751399907 Chr.3: 138031175-138031175 on Build GRCh38
rs1053286810 Chr.3: 138031103-138031103 on Build GRCh38
rs768680816 Chr.3: 138031115-138031115 on Build GRCh38
rs370767483 Chr.3: 138031102-138031102 on Build GRCh38
rs952715745 Chr.3: 138031033-138031033 on Build GRCh38
rs764350012 Chr.3: 138031014-138031014 on Build GRCh38
rs893034016 Chr.3: 138030891-138030891 on Build GRCh38
rs758762263 Chr.3: 138031055-138031055 on Build GRCh38
rs1041591608 Chr.3: 138031203-138031203 on Build GRCh38
rs949540806 Chr.3: 138031066-138031066 on Build GRCh38
rs763361247 Chr.3: 138031130-138031130 on Build GRCh38
rs778117984 Chr.3: 138031192-138031193 on Build GRCh38
rs780698250 Chr.3: 138031073-138031073 on Build GRCh38
rs138222227 Chr.3: 138031125-138031125 on Build GRCh38
rs772982623 Chr.3: 138030930-138030930 on Build GRCh38
rs767039113 Chr.3: 138031132-138031132 on Build GRCh38
rs933855438 Chr.3: 138030851-138030851 on Build GRCh38
rs560519244 Chr.3: 138031226-138031226 on Build GRCh38
rs994223037 Chr.3: 138031258-138031258 on Build GRCh38
rs571741712 Chr.3: 138031198-138031198 on Build GRCh38
rs766998141 Chr.3: 138030853-138030853 on Build GRCh38
rs761250039 Chr.3: 138030823-138030823 on Build GRCh38
rs767640635 Chr.3: 138030985-138030985 on Build GRCh38
rs373585650 Chr.3: 138030958-138030958 on Build GRCh38
rs750815878 Chr.3: 138031129-138031129 on Build GRCh38
rs760733189 Chr.3: 138031006-138031006 on Build GRCh38
rs374979503 Chr.3: 138031180-138031180 on Build GRCh38
rs200380058 Chr.3: 138031145-138031145 on Build GRCh38
rs778506065 Chr.3: 138031092-138031092 on Build GRCh38
rs765470182 Chr.3: 138031026-138031026 on Build GRCh38
rs774372525 Chr.3: 138030979-138030979 on Build GRCh38
rs567940448 Chr.3: 138031078-138031078 on Build GRCh38
rs375252888 Chr.3: 138031044-138031044 on Build GRCh38
rs769964897 Chr.3: 138031190-138031190 on Build GRCh38
rs768735958 Chr.3: 138031179-138031179 on Build GRCh38
rs765684908 Chr.3: 138031091-138031091 on Build GRCh38
rs16847742 Chr.3: 138030997-138030997 on Build GRCh38
rs747803032 Chr.3: 138030919-138030919 on Build GRCh38
rs757669567 Chr.3: 138031022-138031022 on Build GRCh38
rs766193972 Chr.3: 138031107-138031132 on Build GRCh38
rs376205087 Chr.3: 138030990-138030990 on Build GRCh38
rs769635313 Chr.3: 138030927-138030927 on Build GRCh38
rs1028263915 Chr.3: 138030905-138030905 on Build GRCh38
rs991006794 Chr.3: 138031042-138031042 on Build GRCh38
rs139404952 Chr.3: 138030846-138030846 on Build GRCh38
rs150815158 Chr.3: 138031111-138031111 on Build GRCh38
rs1052827423 Chr.3: 138030875-138030875 on Build GRCh38
rs571713437 Chr.3: 138030968-138030968 on Build GRCh38
rs754748790 Chr.3: 138031185-138031186 on Build GRCh38
rs180927693 Chr.3: 138030980-138030980 on Build GRCh38

View Full Product Details