Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138400451-138400954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRAS
Entrez Gene ID
22808
Gene Name
muscle RAS oncogene homolog
Gene Aliases
M-RAs, R-RAS3, RRAS3
UniGene
Hs.527021
Gene Chromosome Location
Chr.3: 138347648-138405535 on Build GRCh38
Transcripts
NM_001085049.2 NM_001252090.1 NM_001252091.1 NM_001252092.1 NM_001252093.1 NM_012219.4 XM_005247228.1 XM_005247229.2 XM_017005887.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5398252 c.451C>T p.P151S Chr.3: 138400537-138400537 on Build GRCh38
1693509 c.493G>A p.D165N Chr.3: 138400579-138400579 on Build GRCh38
6022588 c.474G>A p.K158K Chr.3: 138400560-138400560 on Build GRCh38
1670595 c.460G>T p.E154* Chr.3: 138400546-138400546 on Build GRCh38
3846355 c.492C>A p.V164V Chr.3: 138400578-138400578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777312436 Chr.3: 138400583-138400583 on Build GRCh38
rs765822649 Chr.3: 138400596-138400596 on Build GRCh38
rs749546756 Chr.3: 138400926-138400926 on Build GRCh38
rs1056375413 Chr.3: 138400669-138400669 on Build GRCh38
rs950963001 Chr.3: 138400854-138400854 on Build GRCh38
rs777705388 Chr.3: 138400492-138400492 on Build GRCh38
rs749183340 Chr.3: 138400499-138400499 on Build GRCh38
rs868704071 Chr.3: 138400857-138400857 on Build GRCh38
rs762347485 Chr.3: 138400586-138400586 on Build GRCh38
rs566299958 Chr.3: 138400474-138400474 on Build GRCh38
rs142079021 Chr.3: 138400799-138400799 on Build GRCh38
rs149904534 Chr.3: 138400539-138400539 on Build GRCh38
rs746649291 Chr.3: 138400686-138400686 on Build GRCh38
rs778714349 Chr.3: 138400641-138400641 on Build GRCh38
rs112240787 Chr.3: 138400531-138400531 on Build GRCh38
rs1022969571 Chr.3: 138400847-138400847 on Build GRCh38
rs755934877 Chr.3: 138400627-138400627 on Build GRCh38
rs769097483 Chr.3: 138400579-138400579 on Build GRCh38
rs970105655 Chr.3: 138400849-138400849 on Build GRCh38
rs568889988 Chr.3: 138400611-138400611 on Build GRCh38
rs909212024 Chr.3: 138400920-138400920 on Build GRCh38
rs752238245 Chr.3: 138400625-138400625 on Build GRCh38
rs531133052 Chr.3: 138400700-138400700 on Build GRCh38
rs1011534272 Chr.3: 138400812-138400812 on Build GRCh38
rs745920937 Chr.3: 138400541-138400541 on Build GRCh38
rs111513189 Chr.3: 138400533-138400533 on Build GRCh38
rs997317153 Chr.3: 138400705-138400705 on Build GRCh38
rs370661294 Chr.3: 138400640-138400640 on Build GRCh38
rs976199052 Chr.3: 138400829-138400829 on Build GRCh38
rs746001002 Chr.3: 138400662-138400662 on Build GRCh38
rs1038777453 Chr.3: 138400657-138400657 on Build GRCh38
rs758935565 Chr.3: 138400600-138400600 on Build GRCh38
rs115608872 Chr.3: 138400910-138400910 on Build GRCh38
rs552912667 Chr.3: 138400814-138400814 on Build GRCh38
rs903558385 Chr.3: 138400787-138400787 on Build GRCh38
rs879364551 Chr.3: 138400816-138400816 on Build GRCh38
rs761170472 Chr.3: 138400575-138400575 on Build GRCh38
rs964831208 Chr.3: 138400803-138400803 on Build GRCh38
rs929568881 Chr.3: 138400506-138400506 on Build GRCh38
rs896377981 Chr.3: 138400673-138400673 on Build GRCh38
rs950977096 Chr.3: 138400932-138400932 on Build GRCh38
rs550769490 Chr.3: 138400711-138400711 on Build GRCh38
rs928800634 Chr.3: 138400924-138400926 on Build GRCh38
rs769982876 Chr.3: 138400771-138400771 on Build GRCh38
rs753786137 Chr.3: 138400639-138400639 on Build GRCh38
rs535509894 Chr.3: 138400871-138400871 on Build GRCh38
rs770096230 Chr.3: 138400887-138400887 on Build GRCh38
rs937854399 Chr.3: 138400648-138400648 on Build GRCh38
rs775641289 Chr.3: 138400828-138400828 on Build GRCh38
rs771001666 Chr.3: 138400504-138400504 on Build GRCh38
rs772406756 Chr.3: 138400570-138400570 on Build GRCh38
rs376989664 Chr.3: 138400628-138400628 on Build GRCh38
rs900255868 Chr.3: 138400692-138400692 on Build GRCh38
rs555293201 Chr.3: 138400664-138400664 on Build GRCh38
rs570762740 Chr.3: 138400754-138400754 on Build GRCh38
rs751145113 Chr.3: 138400597-138400597 on Build GRCh38
rs536326964 Chr.3: 138400574-138400574 on Build GRCh38
rs568651825 Chr.3: 138400802-138400802 on Build GRCh38
rs755877770 Chr.3: 138400484-138400484 on Build GRCh38
rs2306373 Chr.3: 138400840-138400840 on Build GRCh38

View Full Product Details