Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110174701-110175207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A17
Entrez Gene ID
388662
Gene Name
solute carrier family 6 member 17
Gene Aliases
MRT48, NTT4
UniGene
Hs.128382
Gene Chromosome Location
Chr.1: 110150510-110202202 on Build GRCh38
Transcripts
NM_001010898.2 XM_006710643.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6057597 c.654_655GG>CT p.E219* Chr.1: 110174861-110174862 on Build GRCh38
3471028 c.732G>A p.K244K Chr.1: 110174939-110174939 on Build GRCh38
3471027 c.629C>T p.A210V Chr.1: 110174836-110174836 on Build GRCh38
4557150 c.716G>A p.G239E Chr.1: 110174923-110174923 on Build GRCh38
5483501 c.576G>A p.V192V Chr.1: 110174783-110174783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934265936 Chr.1: 110175046-110175046 on Build GRCh38
rs374410937 Chr.1: 110175011-110175011 on Build GRCh38
rs749231933 Chr.1: 110174871-110174871 on Build GRCh38
rs754886223 Chr.1: 110174999-110174999 on Build GRCh38
rs764116383 Chr.1: 110174812-110174812 on Build GRCh38
rs752269820 Chr.1: 110174972-110174972 on Build GRCh38
rs773881509 Chr.1: 110174739-110174739 on Build GRCh38
rs146876115 Chr.1: 110175141-110175141 on Build GRCh38
rs572816333 Chr.1: 110175111-110175111 on Build GRCh38
rs767666097 Chr.1: 110174820-110174820 on Build GRCh38
rs188000169 Chr.1: 110174825-110174825 on Build GRCh38
rs763398547 Chr.1: 110174939-110174939 on Build GRCh38
rs904763528 Chr.1: 110174798-110174798 on Build GRCh38
rs142547110 Chr.1: 110174861-110174861 on Build GRCh38
rs916965928 Chr.1: 110175154-110175154 on Build GRCh38
rs994002772 Chr.1: 110174796-110174796 on Build GRCh38
rs368481590 Chr.1: 110175126-110175126 on Build GRCh38
rs536039070 Chr.1: 110175031-110175031 on Build GRCh38
rs751594760 Chr.1: 110174815-110174815 on Build GRCh38
rs544874208 Chr.1: 110175140-110175140 on Build GRCh38
rs768107906 Chr.1: 110174735-110174735 on Build GRCh38
rs538366520 Chr.1: 110174951-110174951 on Build GRCh38
rs59871604 Chr.1: 110175000-110175000 on Build GRCh38
rs377380238 Chr.1: 110174977-110174977 on Build GRCh38
rs754905375 Chr.1: 110174874-110174874 on Build GRCh38
rs774997399 Chr.1: 110174747-110174747 on Build GRCh38
rs759916740 Chr.1: 110174759-110174759 on Build GRCh38
rs770108919 Chr.1: 110174934-110174934 on Build GRCh38
rs753621104 Chr.1: 110174997-110174997 on Build GRCh38
rs1025945443 Chr.1: 110175139-110175139 on Build GRCh38
rs747833914 Chr.1: 110174740-110174740 on Build GRCh38
rs79143268 Chr.1: 110174904-110174904 on Build GRCh38
rs766126977 Chr.1: 110174996-110174996 on Build GRCh38
rs552706330 Chr.1: 110175067-110175067 on Build GRCh38
rs1017510428 Chr.1: 110175026-110175026 on Build GRCh38
rs753401145 Chr.1: 110175178-110175178 on Build GRCh38
rs758042488 Chr.1: 110175008-110175008 on Build GRCh38
rs761812409 Chr.1: 110174957-110174957 on Build GRCh38
rs760929066 Chr.1: 110174987-110174987 on Build GRCh38
rs199792205 Chr.1: 110174752-110174752 on Build GRCh38
rs774425229 Chr.1: 110174954-110174954 on Build GRCh38
rs746648884 Chr.1: 110174933-110174933 on Build GRCh38
rs763498406 Chr.1: 110174810-110174810 on Build GRCh38
rs967683130 Chr.1: 110174918-110174918 on Build GRCh38
rs373414163 Chr.1: 110174903-110174903 on Build GRCh38
rs973254547 Chr.1: 110175078-110175078 on Build GRCh38
rs777404607 Chr.1: 110175010-110175010 on Build GRCh38
rs889800477 Chr.1: 110175114-110175114 on Build GRCh38
rs764608426 Chr.1: 110174953-110174953 on Build GRCh38
rs755927851 Chr.1: 110174860-110174860 on Build GRCh38
rs772717899 Chr.1: 110174925-110174925 on Build GRCh38
rs530309447 Chr.1: 110174723-110174723 on Build GRCh38
rs769368426 Chr.1: 110174875-110174875 on Build GRCh38
rs771857280 Chr.1: 110174919-110174919 on Build GRCh38
rs201642757 Chr.1: 110174753-110174753 on Build GRCh38
rs1057251502 Chr.1: 110174726-110174726 on Build GRCh38
rs775924184 Chr.1: 110174787-110174787 on Build GRCh38
rs765632054 Chr.1: 110174769-110174769 on Build GRCh38
rs777150694 Chr.1: 110174755-110174755 on Build GRCh38
rs757491378 Chr.1: 110174817-110174817 on Build GRCh38
rs370200407 Chr.1: 110174872-110174872 on Build GRCh38
rs750222964 Chr.1: 110174829-110174829 on Build GRCh38
rs1035382942 Chr.1: 110174844-110174844 on Build GRCh38
rs1005603858 Chr.1: 110174754-110174754 on Build GRCh38
rs771804058 Chr.1: 110174741-110174741 on Build GRCh38
rs1030130194 Chr.1: 110174967-110174967 on Build GRCh38
rs763717336 Chr.1: 110175158-110175158 on Build GRCh38

View Full Product Details