Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110453869-110454364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROK1
Entrez Gene ID
84432
Gene Name
prokineticin 1
Gene Aliases
EGVEGF, PK1, PRK1
UniGene
Hs.514793
Gene Chromosome Location
Chr.1: 110451166-110457354 on Build GRCh38
Transcripts
NM_032414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1472403 c.182A>C p.H61P Chr.1: 110454070-110454070 on Build GRCh38
7257114 c.96T>C p.C32C Chr.1: 110453984-110453984 on Build GRCh38
4717927 c.169G>T p.G57C Chr.1: 110454057-110454057 on Build GRCh38
1332601 c.171C>T p.G57G Chr.1: 110454059-110454059 on Build GRCh38
1332600 c.143G>A p.R48Q Chr.1: 110454031-110454031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762618568 Chr.1: 110453929-110453929 on Build GRCh38
rs992027561 Chr.1: 110454180-110454180 on Build GRCh38
rs746949920 Chr.1: 110454074-110454074 on Build GRCh38
rs141353448 Chr.1: 110454031-110454031 on Build GRCh38
rs17628304 Chr.1: 110454328-110454328 on Build GRCh38
rs141461032 Chr.1: 110454051-110454051 on Build GRCh38
rs917698372 Chr.1: 110453914-110453914 on Build GRCh38
rs767050858 Chr.1: 110453944-110453944 on Build GRCh38
rs946292365 Chr.1: 110454308-110454308 on Build GRCh38
rs111441481 Chr.1: 110454089-110454089 on Build GRCh38
rs753551131 Chr.1: 110454052-110454052 on Build GRCh38
rs774210245 Chr.1: 110453932-110453932 on Build GRCh38
rs776177506 Chr.1: 110454077-110454077 on Build GRCh38
rs138146720 Chr.1: 110454059-110454059 on Build GRCh38
rs778697807 Chr.1: 110454067-110454067 on Build GRCh38
rs749908060 Chr.1: 110453946-110453946 on Build GRCh38
rs748129530 Chr.1: 110454002-110454002 on Build GRCh38
rs771947784 Chr.1: 110454007-110454007 on Build GRCh38
rs985184350 Chr.1: 110454304-110454304 on Build GRCh38
rs774890402 Chr.1: 110454112-110454112 on Build GRCh38
rs867258000 Chr.1: 110454179-110454179 on Build GRCh38
rs781470960 Chr.1: 110453975-110453975 on Build GRCh38
rs116061833 Chr.1: 110454180-110454180 on Build GRCh38
rs774097959 Chr.1: 110454001-110454001 on Build GRCh38
rs751031662 Chr.1: 110454134-110454134 on Build GRCh38
rs760342875 Chr.1: 110453959-110453959 on Build GRCh38
rs768644969 Chr.1: 110453930-110453930 on Build GRCh38
rs763604097 Chr.1: 110453996-110453997 on Build GRCh38
rs768102447 Chr.1: 110454046-110454046 on Build GRCh38
rs773074032 Chr.1: 110454071-110454071 on Build GRCh38
rs909508876 Chr.1: 110454005-110454005 on Build GRCh38
rs1037256521 Chr.1: 110454029-110454029 on Build GRCh38
rs758631433 Chr.1: 110453969-110453969 on Build GRCh38
rs761731107 Chr.1: 110453940-110453940 on Build GRCh38
rs113229704 Chr.1: 110454087-110454087 on Build GRCh38
rs760109897 Chr.1: 110454022-110454022 on Build GRCh38
rs764406554 Chr.1: 110454033-110454033 on Build GRCh38
rs149887149 Chr.1: 110453970-110453970 on Build GRCh38
rs181071753 Chr.1: 110454314-110454314 on Build GRCh38
rs775382022 Chr.1: 110453912-110453912 on Build GRCh38
rs936370187 Chr.1: 110454256-110454256 on Build GRCh38
rs764763469 Chr.1: 110453963-110453963 on Build GRCh38
rs923496004 Chr.1: 110454252-110454262 on Build GRCh38
rs142388637 Chr.1: 110454050-110454050 on Build GRCh38
rs62623571 Chr.1: 110454030-110454030 on Build GRCh38
rs532479267 Chr.1: 110453971-110453971 on Build GRCh38
rs571070891 Chr.1: 110454150-110454150 on Build GRCh38
rs745897801 Chr.1: 110453981-110453981 on Build GRCh38
rs749125320 Chr.1: 110453993-110453993 on Build GRCh38
rs772739896 Chr.1: 110454010-110454010 on Build GRCh38
rs146968434 Chr.1: 110454111-110454111 on Build GRCh38
rs962397976 Chr.1: 110454316-110454316 on Build GRCh38
rs536227805 Chr.1: 110453951-110453951 on Build GRCh38
rs987746115 Chr.1: 110454287-110454287 on Build GRCh38
rs780076706 Chr.1: 110453982-110453982 on Build GRCh38
rs372495524 Chr.1: 110453910-110453910 on Build GRCh38
rs528950996 Chr.1: 110454234-110454234 on Build GRCh38
rs757864792 Chr.1: 110454044-110454044 on Build GRCh38
rs759160619 Chr.1: 110454091-110454091 on Build GRCh38
rs200271858 Chr.1: 110454076-110454076 on Build GRCh38
rs756092160 Chr.1: 110454052-110454052 on Build GRCh38
rs761438937 Chr.1: 110453898-110453898 on Build GRCh38
rs951274783 Chr.1: 110454144-110454144 on Build GRCh38
rs200512156 Chr.1: 110453998-110453998 on Build GRCh38
rs565581756 Chr.1: 110454133-110454133 on Build GRCh38
rs772001059 Chr.1: 110454070-110454070 on Build GRCh38
rs755200880 Chr.1: 110454060-110454060 on Build GRCh38
rs559326098 Chr.1: 110454095-110454095 on Build GRCh38
rs751951334 Chr.1: 110454043-110454043 on Build GRCh38
rs548816420 Chr.1: 110454237-110454237 on Build GRCh38
rs202098995 Chr.1: 110453962-110453962 on Build GRCh38
rs969545869 Chr.1: 110454227-110454227 on Build GRCh38

View Full Product Details