Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111476694-111477204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf162
Entrez Gene ID
128346
Gene Name
chromosome 1 open reading frame 162
Gene Aliases
-
UniGene
Hs.288010
Gene Chromosome Location
Chr.1: 111462694-111478512 on Build GRCh38
Transcripts
NM_001300834.1 NM_001300835.1 NM_174896.3 XM_011540712.2 XM_017000323.1 XM_017000324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615123 c.43C>T p.Q15* Chr.1: 111476803-111476803 on Build GRCh38
3471243 c.64G>T p.A22S Chr.1: 111476824-111476824 on Build GRCh38
6284746 c.43C>G p.Q15E Chr.1: 111476803-111476803 on Build GRCh38
1246662 c.50C>G p.T17S Chr.1: 111476810-111476810 on Build GRCh38
462609 c.60A>C p.T20T Chr.1: 111476820-111476820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs193297910 Chr.1: 111476999-111476999 on Build GRCh38
rs564428561 Chr.1: 111477002-111477002 on Build GRCh38
rs556179888 Chr.1: 111476770-111476770 on Build GRCh38
rs1025838491 Chr.1: 111476928-111476928 on Build GRCh38
rs776855443 Chr.1: 111476829-111476829 on Build GRCh38
rs112021080 Chr.1: 111476906-111476906 on Build GRCh38
rs116446242 Chr.1: 111476925-111476925 on Build GRCh38
rs201845325 Chr.1: 111476820-111476820 on Build GRCh38
rs778356541 Chr.1: 111476918-111476918 on Build GRCh38
rs913154356 Chr.1: 111476772-111476772 on Build GRCh38
rs905171009 Chr.1: 111477160-111477160 on Build GRCh38
rs1015119072 Chr.1: 111476976-111476976 on Build GRCh38
rs900904275 Chr.1: 111477142-111477142 on Build GRCh38
rs540432008 Chr.1: 111477148-111477148 on Build GRCh38
rs929763513 Chr.1: 111477144-111477144 on Build GRCh38
rs1005451168 Chr.1: 111476965-111476965 on Build GRCh38
rs947296921 Chr.1: 111477095-111477095 on Build GRCh38
rs74109580 Chr.1: 111477166-111477166 on Build GRCh38
rs763483566 Chr.1: 111476859-111476859 on Build GRCh38
rs978655774 Chr.1: 111477117-111477117 on Build GRCh38
rs754452504 Chr.1: 111476776-111476776 on Build GRCh38
rs747109721 Chr.1: 111476786-111476786 on Build GRCh38
rs943917149 Chr.1: 111477092-111477092 on Build GRCh38
rs1023635335 Chr.1: 111476946-111476946 on Build GRCh38
rs150674974 Chr.1: 111476845-111476845 on Build GRCh38
rs913102454 Chr.1: 111477075-111477075 on Build GRCh38
rs150912013 Chr.1: 111476827-111476827 on Build GRCh38
rs767760986 Chr.1: 111476869-111476869 on Build GRCh38
rs771750394 Chr.1: 111476797-111476797 on Build GRCh38
rs373597824 Chr.1: 111476764-111476764 on Build GRCh38
rs990163765 Chr.1: 111477056-111477056 on Build GRCh38
rs41301285 Chr.1: 111477130-111477130 on Build GRCh38
rs761983469 Chr.1: 111476864-111476864 on Build GRCh38
rs887918893 Chr.1: 111476963-111476963 on Build GRCh38
rs372291235 Chr.1: 111476747-111476747 on Build GRCh38
rs753788162 Chr.1: 111476897-111476897 on Build GRCh38
rs781718310 Chr.1: 111476805-111476805 on Build GRCh38
rs958702981 Chr.1: 111477018-111477018 on Build GRCh38
rs778430791 Chr.1: 111476792-111476792 on Build GRCh38
rs774474478 Chr.1: 111476861-111476861 on Build GRCh38
rs533049296 Chr.1: 111476980-111476980 on Build GRCh38
rs561686332 Chr.1: 111477149-111477149 on Build GRCh38
rs977655628 Chr.1: 111477076-111477076 on Build GRCh38
rs531855022 Chr.1: 111477127-111477127 on Build GRCh38
rs201028931 Chr.1: 111476800-111476800 on Build GRCh38
rs74109579 Chr.1: 111477153-111477153 on Build GRCh38
rs775113654 Chr.1: 111476932-111476932 on Build GRCh38
rs768628858 Chr.1: 111476860-111476860 on Build GRCh38
rs748325571 Chr.1: 111476886-111476886 on Build GRCh38
rs753735365 Chr.1: 111476771-111476771 on Build GRCh38
rs912450878 Chr.1: 111476803-111476803 on Build GRCh38
rs750190651 Chr.1: 111476748-111476748 on Build GRCh38
rs912428822 Chr.1: 111477082-111477082 on Build GRCh38
rs750598713 Chr.1: 111476885-111476885 on Build GRCh38
rs377423729 Chr.1: 111476890-111476890 on Build GRCh38
rs576232943 Chr.1: 111476806-111476806 on Build GRCh38
rs112752296 Chr.1: 111477059-111477059 on Build GRCh38
rs967512013 Chr.1: 111477064-111477064 on Build GRCh38
rs752218235 Chr.1: 111476919-111476919 on Build GRCh38
rs767323108 Chr.1: 111476858-111476860 on Build GRCh38
rs770234485 Chr.1: 111476817-111476817 on Build GRCh38
rs767473245 Chr.1: 111477040-111477040 on Build GRCh38
rs1039624974 Chr.1: 111477106-111477106 on Build GRCh38
rs779993923 Chr.1: 111476766-111476766 on Build GRCh38
rs1048188668 Chr.1: 111476958-111476958 on Build GRCh38
rs768555109 Chr.1: 111476794-111476794 on Build GRCh38
rs1039766559 Chr.1: 111476853-111476853 on Build GRCh38
rs775102955 Chr.1: 111476865-111476865 on Build GRCh38
rs759437189 Chr.1: 111477019-111477019 on Build GRCh38
rs564088323 Chr.1: 111476935-111476935 on Build GRCh38
rs988721977 Chr.1: 111476725-111476725 on Build GRCh38
rs1035532924 Chr.1: 111476985-111476985 on Build GRCh38
rs1035057140 Chr.1: 111477179-111477179 on Build GRCh38
rs756096736 Chr.1: 111476750-111476750 on Build GRCh38

View Full Product Details