Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113879518-113880057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4B1-AS1
Entrez Gene ID
100287722
Gene Name
AP4B1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.664669
Gene Chromosome Location
Chr.1: 113812612-113901237 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Gene Symbol
BCL2L15
Entrez Gene ID
440603
Gene Name
BCL2 like 15
Gene Aliases
Bfk, C1orf178
UniGene
Hs.123106
Gene Chromosome Location
Chr.1: 113876814-113887547 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

SNP Detail

SNP ID Chromosome Location
rs947669190 Chr.1: 113879656-113879656 on Build GRCh38
rs781579334 Chr.1: 113879910-113879910 on Build GRCh38
rs973370878 Chr.1: 113879934-113879934 on Build GRCh38
rs963377151 Chr.1: 113879932-113879932 on Build GRCh38
rs1041136965 Chr.1: 113879720-113879720 on Build GRCh38
rs977601448 Chr.1: 113879660-113879660 on Build GRCh38
rs191716869 Chr.1: 113879869-113879869 on Build GRCh38
rs753684166 Chr.1: 113879717-113879717 on Build GRCh38
rs1054073219 Chr.1: 113879758-113879758 on Build GRCh38
rs761778622 Chr.1: 113879719-113879719 on Build GRCh38
rs914840163 Chr.1: 113879612-113879612 on Build GRCh38
rs1041462651 Chr.1: 113879700-113879700 on Build GRCh38
rs76583534 Chr.1: 113879798-113879798 on Build GRCh38
rs567566759 Chr.1: 113879880-113879880 on Build GRCh38
rs1035432515 Chr.1: 113879580-113879580 on Build GRCh38
rs569830939 Chr.1: 113879570-113879570 on Build GRCh38
rs924797666 Chr.1: 113879691-113879691 on Build GRCh38
rs141578669 Chr.1: 113879607-113879607 on Build GRCh38
rs74728716 Chr.1: 113879638-113879638 on Build GRCh38
rs927190557 Chr.1: 113879573-113879573 on Build GRCh38
rs534729324 Chr.1: 113879621-113879621 on Build GRCh38
rs563082790 Chr.1: 113879661-113879661 on Build GRCh38
rs577719962 Chr.1: 113879643-113879643 on Build GRCh38
rs927274193 Chr.1: 113879942-113879942 on Build GRCh38
rs868549128 Chr.1: 113880024-113880024 on Build GRCh38
rs138535337 Chr.1: 113879879-113879879 on Build GRCh38
rs897690417 Chr.1: 113879927-113879927 on Build GRCh38
rs1036250640 Chr.1: 113879900-113879900 on Build GRCh38
rs958471211 Chr.1: 113880000-113880000 on Build GRCh38
rs535332282 Chr.1: 113879578-113879578 on Build GRCh38
rs898711208 Chr.1: 113879834-113879834 on Build GRCh38
rs917053060 Chr.1: 113879670-113879670 on Build GRCh38
rs564092500 Chr.1: 113879930-113879930 on Build GRCh38
rs373849589 Chr.1: 113879954-113879955 on Build GRCh38
rs546470498 Chr.1: 113879648-113879648 on Build GRCh38
rs1035885312 Chr.1: 113879962-113879962 on Build GRCh38
rs555424211 Chr.1: 113879605-113879605 on Build GRCh38
rs533201370 Chr.1: 113880023-113880023 on Build GRCh38
rs996404897 Chr.1: 113879797-113879797 on Build GRCh38
rs188760734 Chr.1: 113879630-113879630 on Build GRCh38
rs1039932632 Chr.1: 113879739-113879739 on Build GRCh38
rs992727691 Chr.1: 113880012-113880012 on Build GRCh38
rs184227078 Chr.1: 113879876-113879876 on Build GRCh38
rs576568721 Chr.1: 113879748-113879748 on Build GRCh38
rs750387489 Chr.1: 113879894-113879894 on Build GRCh38
rs932679003 Chr.1: 113879793-113879793 on Build GRCh38

View Full Product Details