Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138946708-138947250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL2
Entrez Gene ID
668
Gene Name
forkhead box L2
Gene Aliases
BPES, BPES1, PFRK, PINTO, POF3
UniGene
Hs.289292
Gene Chromosome Location
Chr.3: 138944224-138947140 on Build GRCh38
Transcripts
NM_023067.3 NM_001040061.2 XM_005247443.3

Gene Symbol
FOXL2NB
Entrez Gene ID
401089
Gene Name
FOXL2 neighbor
Gene Aliases
C3orf72
UniGene
Hs.591303
Gene Chromosome Location
Chr.3: 138947234-138953992 on Build GRCh38
Transcripts
NM_023067.3 NM_001040061.2 XM_005247443.3

SNP Detail

SNP ID Chromosome Location
rs745676338 Chr.3: 138946747-138946747 on Build GRCh38
rs539109610 Chr.3: 138946882-138946882 on Build GRCh38
rs576439625 Chr.3: 138946868-138946868 on Build GRCh38
rs994655165 Chr.3: 138947205-138947205 on Build GRCh38
rs534227199 Chr.3: 138946729-138946729 on Build GRCh38
rs928364190 Chr.3: 138947101-138947101 on Build GRCh38
rs1006121427 Chr.3: 138947089-138947089 on Build GRCh38
rs879804830 Chr.3: 138946877-138946877 on Build GRCh38
rs893285499 Chr.3: 138947119-138947119 on Build GRCh38
rs970126224 Chr.3: 138946951-138946951 on Build GRCh38
rs561023238 Chr.3: 138947000-138947000 on Build GRCh38
rs756989770 Chr.3: 138946741-138946741 on Build GRCh38
rs914224829 Chr.3: 138947116-138947116 on Build GRCh38
rs1053780927 Chr.3: 138946948-138946948 on Build GRCh38
rs145924266 Chr.3: 138946973-138946973 on Build GRCh38
rs944186472 Chr.3: 138947193-138947193 on Build GRCh38
rs1016213433 Chr.3: 138947213-138947213 on Build GRCh38
rs889933449 Chr.3: 138946987-138946987 on Build GRCh38
rs746906405 Chr.3: 138946764-138946764 on Build GRCh38
rs950573234 Chr.3: 138946967-138946967 on Build GRCh38
rs1011267885 Chr.3: 138946926-138946926 on Build GRCh38
rs200169078 Chr.3: 138946763-138946763 on Build GRCh38
rs1048456863 Chr.3: 138947207-138947207 on Build GRCh38
rs192325010 Chr.3: 138947063-138947063 on Build GRCh38
rs1053327674 Chr.3: 138947111-138947111 on Build GRCh38
rs755181359 Chr.3: 138946806-138946806 on Build GRCh38
rs1022736785 Chr.3: 138946949-138946949 on Build GRCh38
rs888423213 Chr.3: 138947227-138947227 on Build GRCh38
rs1023128934 Chr.3: 138947125-138947125 on Build GRCh38
rs1051781437 Chr.3: 138946863-138946864 on Build GRCh38
rs1008748288 Chr.3: 138946996-138946996 on Build GRCh38
rs1026987825 Chr.3: 138947072-138947072 on Build GRCh38
rs532092423 Chr.3: 138947179-138947179 on Build GRCh38
rs897279294 Chr.3: 138947204-138947204 on Build GRCh38
rs955979189 Chr.3: 138947103-138947103 on Build GRCh38
rs558805988 Chr.3: 138946886-138946886 on Build GRCh38
rs746544902 Chr.3: 138947210-138947210 on Build GRCh38
rs771930968 Chr.3: 138946750-138946750 on Build GRCh38
rs574415705 Chr.3: 138947059-138947059 on Build GRCh38
rs941874514 Chr.3: 138947027-138947027 on Build GRCh38
rs996884213 Chr.3: 138946797-138946797 on Build GRCh38
rs547635156 Chr.3: 138946734-138946734 on Build GRCh38
rs551814994 Chr.3: 138947200-138947200 on Build GRCh38
rs934308579 Chr.3: 138946944-138946944 on Build GRCh38
rs1022820959 Chr.3: 138947095-138947095 on Build GRCh38
rs778422326 Chr.3: 138946742-138946742 on Build GRCh38
rs1003037092 Chr.3: 138947162-138947162 on Build GRCh38
rs556654191 Chr.3: 138946842-138946842 on Build GRCh38
rs911405048 Chr.3: 138947163-138947163 on Build GRCh38
rs1049914329 Chr.3: 138946975-138946975 on Build GRCh38
rs981604888 Chr.3: 138947100-138947100 on Build GRCh38
rs951977992 Chr.3: 138947077-138947077 on Build GRCh38
rs748722930 Chr.3: 138946737-138946737 on Build GRCh38
rs11924939 Chr.3: 138946954-138946954 on Build GRCh38
rs1035769406 Chr.3: 138946965-138946965 on Build GRCh38
rs897157459 Chr.3: 138947025-138947025 on Build GRCh38
rs969887077 Chr.3: 138947098-138947098 on Build GRCh38
rs921400566 Chr.3: 138947052-138947052 on Build GRCh38
rs905577229 Chr.3: 138947159-138947159 on Build GRCh38
rs960184242 Chr.3: 138947051-138947051 on Build GRCh38
rs981188257 Chr.3: 138946912-138946912 on Build GRCh38
rs984664902 Chr.3: 138946810-138946810 on Build GRCh38
rs879018674 Chr.3: 138947040-138947040 on Build GRCh38
rs974547520 Chr.3: 138947051-138947052 on Build GRCh38
rs754194513 Chr.3: 138946783-138946783 on Build GRCh38
rs972404907 Chr.3: 138946752-138946752 on Build GRCh38
rs567696496 Chr.3: 138946756-138946756 on Build GRCh38
rs768686586 Chr.3: 138946767-138946767 on Build GRCh38
rs928301280 Chr.3: 138946916-138946916 on Build GRCh38
rs947664013 Chr.3: 138947121-138947121 on Build GRCh38
rs755652331 Chr.3: 138946731-138946731 on Build GRCh38
rs1049944234 Chr.3: 138947149-138947149 on Build GRCh38
rs951863654 Chr.3: 138946796-138946796 on Build GRCh38
rs563265663 Chr.3: 138947109-138947109 on Build GRCh38

View Full Product Details