Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 142664109-142664647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLS1
Entrez Gene ID
5357
Gene Name
plastin 1
Gene Aliases
-
UniGene
Hs.203637
Gene Chromosome Location
Chr.3: 142596387-142713664 on Build GRCh38
Transcripts
NM_001145319.1 NM_001172312.1 NM_002670.2 XM_006713660.3 XM_011512900.2 XM_011512901.1 XM_011512903.2 XM_017006626.1 XM_017006627.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
218752 c.11G>A p.S4N Chr.3: 142664248-142664248 on Build GRCh38
3408298 c.46C>G p.L16V Chr.3: 142664283-142664283 on Build GRCh38
6450617 c.29G>A p.R10Q Chr.3: 142664266-142664266 on Build GRCh38
3940326 c.36G>A p.E12E Chr.3: 142664273-142664273 on Build GRCh38
4762186 c.40G>C p.E14Q Chr.3: 142664277-142664277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778735363 Chr.3: 142664240-142664240 on Build GRCh38
rs760684545 Chr.3: 142664251-142664251 on Build GRCh38
rs748981259 Chr.3: 142664330-142664332 on Build GRCh38
rs763240317 Chr.3: 142664291-142664291 on Build GRCh38
rs755633871 Chr.3: 142664334-142664334 on Build GRCh38
rs772320882 Chr.3: 142664247-142664247 on Build GRCh38
rs768235395 Chr.3: 142664270-142664270 on Build GRCh38
rs768758489 Chr.3: 142664255-142664255 on Build GRCh38
rs77961515 Chr.3: 142664153-142664153 on Build GRCh38
rs981761003 Chr.3: 142664545-142664545 on Build GRCh38
rs768836781 Chr.3: 142664255-142664255 on Build GRCh38
rs750795154 Chr.3: 142664290-142664290 on Build GRCh38
rs182914807 Chr.3: 142664210-142664210 on Build GRCh38
rs1055926576 Chr.3: 142664167-142664167 on Build GRCh38
rs961341499 Chr.3: 142664514-142664514 on Build GRCh38
rs532402477 Chr.3: 142664404-142664404 on Build GRCh38
rs777811284 Chr.3: 142664213-142664213 on Build GRCh38
rs192922375 Chr.3: 142664552-142664552 on Build GRCh38
rs371422827 Chr.3: 142664189-142664189 on Build GRCh38
rs567137077 Chr.3: 142664508-142664508 on Build GRCh38
rs752427577 Chr.3: 142664316-142664316 on Build GRCh38
rs778769706 Chr.3: 142664354-142664354 on Build GRCh38
rs200819457 Chr.3: 142664319-142664319 on Build GRCh38
rs373717334 Chr.3: 142664248-142664248 on Build GRCh38
rs866296829 Chr.3: 142664609-142664609 on Build GRCh38
rs770820118 Chr.3: 142664233-142664233 on Build GRCh38
rs745483232 Chr.3: 142664192-142664192 on Build GRCh38
rs2272150 Chr.3: 142664476-142664476 on Build GRCh38
rs529917813 Chr.3: 142664415-142664415 on Build GRCh38
rs921052903 Chr.3: 142664555-142664555 on Build GRCh38
rs367822162 Chr.3: 142664341-142664341 on Build GRCh38
rs147765025 Chr.3: 142664506-142664506 on Build GRCh38
rs990514767 Chr.3: 142664583-142664583 on Build GRCh38
rs187467555 Chr.3: 142664526-142664526 on Build GRCh38
rs13075015 Chr.3: 142664448-142664448 on Build GRCh38
rs894639199 Chr.3: 142664175-142664176 on Build GRCh38
rs995729158 Chr.3: 142664428-142664428 on Build GRCh38
rs745712760 Chr.3: 142664246-142664246 on Build GRCh38
rs939684229 Chr.3: 142664138-142664138 on Build GRCh38
rs770221691 Chr.3: 142664248-142664249 on Build GRCh38
rs927701291 Chr.3: 142664147-142664147 on Build GRCh38
rs892404460 Chr.3: 142664182-142664182 on Build GRCh38
rs1019292522 Chr.3: 142664504-142664504 on Build GRCh38
rs749306895 Chr.3: 142664489-142664489 on Build GRCh38
rs768477627 Chr.3: 142664573-142664573 on Build GRCh38
rs767129680 Chr.3: 142664306-142664306 on Build GRCh38
rs552770026 Chr.3: 142664521-142664521 on Build GRCh38
rs749259192 Chr.3: 142664230-142664230 on Build GRCh38
rs1028195262 Chr.3: 142664381-142664381 on Build GRCh38
rs765704001 Chr.3: 142664277-142664277 on Build GRCh38
rs907190033 Chr.3: 142664314-142664314 on Build GRCh38
rs569571675 Chr.3: 142664470-142664470 on Build GRCh38
rs534851528 Chr.3: 142664518-142664518 on Build GRCh38
rs777076310 Chr.3: 142664265-142664265 on Build GRCh38
rs182676444 Chr.3: 142664582-142664582 on Build GRCh38
rs763673034 Chr.3: 142664340-142664340 on Build GRCh38
rs1034363520 Chr.3: 142664357-142664357 on Build GRCh38
rs762287294 Chr.3: 142664266-142664266 on Build GRCh38
rs757211047 Chr.3: 142664348-142664348 on Build GRCh38
rs1046590889 Chr.3: 142664140-142664140 on Build GRCh38

View Full Product Details