Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 147391880-147392385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZIC4
Entrez Gene ID
84107
Gene Name
Zic family member 4
Gene Aliases
-
UniGene
Hs.415766
Gene Chromosome Location
Chr.3: 147386048-147406893 on Build GRCh38
Transcripts
NM_001168378.1 NM_001168379.1 NM_001243256.1 NM_032153.5 NR_033118.2 NR_033119.2 NR_040762.1

SNP Detail

SNP ID Chromosome Location
rs919093071 Chr.3: 147392152-147392152 on Build GRCh38
rs1052260899 Chr.3: 147392132-147392132 on Build GRCh38
rs906837221 Chr.3: 147392272-147392272 on Build GRCh38
rs371422967 Chr.3: 147392235-147392235 on Build GRCh38
rs994101743 Chr.3: 147392296-147392296 on Build GRCh38
rs954302771 Chr.3: 147392299-147392299 on Build GRCh38
rs550966870 Chr.3: 147392136-147392136 on Build GRCh38
rs922372533 Chr.3: 147392122-147392122 on Build GRCh38
rs962559454 Chr.3: 147392019-147392019 on Build GRCh38
rs530842226 Chr.3: 147392108-147392108 on Build GRCh38
rs566799487 Chr.3: 147392049-147392049 on Build GRCh38
rs571494693 Chr.3: 147392081-147392081 on Build GRCh38
rs905171687 Chr.3: 147392198-147392198 on Build GRCh38
rs1007780319 Chr.3: 147392316-147392316 on Build GRCh38
rs1003041750 Chr.3: 147392281-147392281 on Build GRCh38
rs1045013868 Chr.3: 147391922-147391922 on Build GRCh38
rs538826686 Chr.3: 147392303-147392303 on Build GRCh38
rs1048932925 Chr.3: 147391929-147391929 on Build GRCh38
rs887724536 Chr.3: 147392202-147392202 on Build GRCh38
rs1035204572 Chr.3: 147391957-147391957 on Build GRCh38
rs1031378662 Chr.3: 147392284-147392284 on Build GRCh38
rs191471174 Chr.3: 147391954-147391954 on Build GRCh38
rs577595862 Chr.3: 147391928-147391928 on Build GRCh38
rs976942605 Chr.3: 147392083-147392083 on Build GRCh38
rs905302675 Chr.3: 147391936-147391936 on Build GRCh38
rs200629412 Chr.3: 147392239-147392243 on Build GRCh38
rs1048872595 Chr.3: 147392172-147392172 on Build GRCh38
rs1020046209 Chr.3: 147392352-147392352 on Build GRCh38
rs777057850 Chr.3: 147391908-147391908 on Build GRCh38
rs897893503 Chr.3: 147392288-147392288 on Build GRCh38
rs374684018 Chr.3: 147391998-147391998 on Build GRCh38
rs750745261 Chr.3: 147392162-147392162 on Build GRCh38
rs1027796323 Chr.3: 147392297-147392297 on Build GRCh38
rs963796940 Chr.3: 147392082-147392082 on Build GRCh38
rs1037090726 Chr.3: 147392151-147392151 on Build GRCh38
rs1017861631 Chr.3: 147392319-147392319 on Build GRCh38
rs371666201 Chr.3: 147391931-147391931 on Build GRCh38
rs546648035 Chr.3: 147392224-147392224 on Build GRCh38
rs887692082 Chr.3: 147391991-147391991 on Build GRCh38
rs1032268808 Chr.3: 147392050-147392054 on Build GRCh38
rs891537927 Chr.3: 147392295-147392295 on Build GRCh38
rs749980646 Chr.3: 147392225-147392225 on Build GRCh38
rs983802036 Chr.3: 147392050-147392050 on Build GRCh38
rs888048263 Chr.3: 147392306-147392306 on Build GRCh38
rs183618097 Chr.3: 147392140-147392140 on Build GRCh38
rs58262807 Chr.3: 147392042-147392042 on Build GRCh38
rs1021791234 Chr.3: 147392298-147392298 on Build GRCh38
rs367888174 Chr.3: 147391921-147391921 on Build GRCh38
rs866272009 Chr.3: 147391909-147391909 on Build GRCh38
rs1045044999 Chr.3: 147392195-147392195 on Build GRCh38
rs1056991495 Chr.3: 147392264-147392264 on Build GRCh38
rs529678587 Chr.3: 147392032-147392032 on Build GRCh38
rs985489968 Chr.3: 147392131-147392131 on Build GRCh38
rs985686430 Chr.3: 147392320-147392320 on Build GRCh38
rs990402120 Chr.3: 147392078-147392078 on Build GRCh38
rs759907661 Chr.3: 147391928-147391928 on Build GRCh38
rs559023533 Chr.3: 147392349-147392349 on Build GRCh38
rs529022274 Chr.3: 147391980-147391980 on Build GRCh38
rs563104700 Chr.3: 147391963-147391963 on Build GRCh38
rs567293700 Chr.3: 147392138-147392138 on Build GRCh38
rs935183150 Chr.3: 147392126-147392126 on Build GRCh38
rs551431195 Chr.3: 147392060-147392060 on Build GRCh38

View Full Product Details