Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149026990-149027501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYG1
Entrez Gene ID
2992
Gene Name
glycogenin 1
Gene Aliases
GSD15, GYG
UniGene
Hs.477892
Gene Chromosome Location
Chr.3: 148991408-149027669 on Build GRCh38
Transcripts
NM_001184720.1 NM_001184721.1 NM_004130.3 XM_017006275.1 XM_017006276.1

SNP Detail

SNP ID Chromosome Location
rs548548843 Chr.3: 149027179-149027179 on Build GRCh38
rs747618973 Chr.3: 149027094-149027094 on Build GRCh38
rs180791582 Chr.3: 149027145-149027145 on Build GRCh38
rs779185556 Chr.3: 149027057-149027058 on Build GRCh38
rs755885817 Chr.3: 149027058-149027058 on Build GRCh38
rs549567633 Chr.3: 149027309-149027309 on Build GRCh38
rs1020285244 Chr.3: 149027269-149027269 on Build GRCh38
rs1024035973 Chr.3: 149027229-149027230 on Build GRCh38
rs779869156 Chr.3: 149027060-149027060 on Build GRCh38
rs191817695 Chr.3: 149027024-149027024 on Build GRCh38
rs1057348109 Chr.3: 149027079-149027079 on Build GRCh38
rs532787062 Chr.3: 149027263-149027263 on Build GRCh38
rs927458591 Chr.3: 149027347-149027347 on Build GRCh38
rs998512675 Chr.3: 149027195-149027195 on Build GRCh38
rs761762601 Chr.3: 149027135-149027135 on Build GRCh38
rs115945803 Chr.3: 149027364-149027364 on Build GRCh38
rs3347 Chr.3: 149027449-149027449 on Build GRCh38
rs79788143 Chr.3: 149027255-149027255 on Build GRCh38
rs890308700 Chr.3: 149027065-149027065 on Build GRCh38
rs190363713 Chr.3: 149027353-149027353 on Build GRCh38
rs112590964 Chr.3: 149027074-149027074 on Build GRCh38
rs917485718 Chr.3: 149027404-149027404 on Build GRCh38
rs147583759 Chr.3: 149027334-149027334 on Build GRCh38
rs773794159 Chr.3: 149027072-149027072 on Build GRCh38
rs11541524 Chr.3: 149027064-149027064 on Build GRCh38
rs777963746 Chr.3: 149027041-149027041 on Build GRCh38
rs899661893 Chr.3: 149027149-149027149 on Build GRCh38
rs572380867 Chr.3: 149027407-149027407 on Build GRCh38
rs758586179 Chr.3: 149027054-149027054 on Build GRCh38
rs138925323 Chr.3: 149027081-149027082 on Build GRCh38
rs145168414 Chr.3: 149027123-149027123 on Build GRCh38
rs755691102 Chr.3: 149027017-149027018 on Build GRCh38
rs749219967 Chr.3: 149027063-149027063 on Build GRCh38
rs921943143 Chr.3: 149027368-149027369 on Build GRCh38
rs771713792 Chr.3: 149027102-149027102 on Build GRCh38
rs745631121 Chr.3: 149027057-149027057 on Build GRCh38
rs768639978 Chr.3: 149027071-149027071 on Build GRCh38
rs754442802 Chr.3: 149027129-149027129 on Build GRCh38
rs538686115 Chr.3: 149027230-149027231 on Build GRCh38
rs958365017 Chr.3: 149027148-149027148 on Build GRCh38
rs537865633 Chr.3: 149027394-149027395 on Build GRCh38
rs561952462 Chr.3: 149027196-149027196 on Build GRCh38
rs1046814867 Chr.3: 149027112-149027112 on Build GRCh38
rs547392072 Chr.3: 149027257-149027257 on Build GRCh38
rs184040045 Chr.3: 149027052-149027052 on Build GRCh38
rs993501890 Chr.3: 149027390-149027390 on Build GRCh38
rs374995146 Chr.3: 149027278-149027278 on Build GRCh38
rs777890348 Chr.3: 149027055-149027055 on Build GRCh38

View Full Product Details