Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149761013-149761512 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKUB1
Entrez Gene ID
389161
Gene Name
ankyrin repeat and ubiquitin domain containing 1
Gene Aliases
C3orf16
UniGene
Hs.665988
Gene Chromosome Location
Chr.3: 149760648-149792823 on Build GRCh38
Transcripts
NM_001144960.2 NM_001315505.1 XM_011512798.1 XM_011512801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4987083 c.1622A>G p.E541G Chr.3: 149761497-149761497 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000179838 Chr.3: 149761212-149761212 on Build GRCh38
rs930283359 Chr.3: 149761403-149761403 on Build GRCh38
rs559063813 Chr.3: 149761119-149761119 on Build GRCh38
rs538871230 Chr.3: 149761051-149761051 on Build GRCh38
rs889966016 Chr.3: 149761138-149761138 on Build GRCh38
rs565696662 Chr.3: 149761383-149761383 on Build GRCh38
rs940652424 Chr.3: 149761349-149761349 on Build GRCh38
rs529637328 Chr.3: 149761234-149761234 on Build GRCh38
rs993923680 Chr.3: 149761276-149761276 on Build GRCh38
rs879579610 Chr.3: 149761185-149761185 on Build GRCh38
rs896996688 Chr.3: 149761275-149761275 on Build GRCh38
rs927868083 Chr.3: 149761039-149761039 on Build GRCh38
rs538300928 Chr.3: 149761239-149761239 on Build GRCh38
rs555709322 Chr.3: 149761288-149761288 on Build GRCh38
rs965698179 Chr.3: 149761206-149761206 on Build GRCh38
rs989686474 Chr.3: 149761470-149761470 on Build GRCh38
rs756262247 Chr.3: 149761267-149761267 on Build GRCh38
rs1007999687 Chr.3: 149761248-149761248 on Build GRCh38
rs868737174 Chr.3: 149761214-149761214 on Build GRCh38
rs547376962 Chr.3: 149761353-149761353 on Build GRCh38
rs763008788 Chr.3: 149761136-149761136 on Build GRCh38
rs540627014 Chr.3: 149761297-149761297 on Build GRCh38
rs971113386 Chr.3: 149761372-149761372 on Build GRCh38
rs1006979494 Chr.3: 149761176-149761176 on Build GRCh38
rs927783807 Chr.3: 149761317-149761317 on Build GRCh38
rs765806155 Chr.3: 149761165-149761165 on Build GRCh38
rs183091511 Chr.3: 149761270-149761270 on Build GRCh38
rs898817395 Chr.3: 149761111-149761111 on Build GRCh38
rs982232347 Chr.3: 149761376-149761376 on Build GRCh38
rs1026826241 Chr.3: 149761097-149761097 on Build GRCh38
rs1054047567 Chr.3: 149761169-149761172 on Build GRCh38
rs533762328 Chr.3: 149761146-149761147 on Build GRCh38
rs949526135 Chr.3: 149761146-149761146 on Build GRCh38
rs190575408 Chr.3: 149761193-149761193 on Build GRCh38
rs574912386 Chr.3: 149761296-149761296 on Build GRCh38
rs940351982 Chr.3: 149761072-149761072 on Build GRCh38
rs1522237 Chr.3: 149761298-149761298 on Build GRCh38
rs990821908 Chr.3: 149761131-149761131 on Build GRCh38
rs954715 Chr.3: 149761469-149761469 on Build GRCh38
rs915455847 Chr.3: 149761180-149761180 on Build GRCh38
rs1036068054 Chr.3: 149761073-149761073 on Build GRCh38
rs924152347 Chr.3: 149761156-149761157 on Build GRCh38
rs34971698 Chr.3: 149761157-149761157 on Build GRCh38

View Full Product Details