Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150742611-150743105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIAH2
Entrez Gene ID
6478
Gene Name
siah E3 ubiquitin protein ligase 2
Gene Aliases
hSiah2
UniGene
Hs.477959
Gene Chromosome Location
Chr.3: 150741123-150763476 on Build GRCh38
Transcripts
NM_005067.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6362281 c.431G>A p.G144D Chr.3: 150742685-150742685 on Build GRCh38
1039892 c.442A>G p.T148A Chr.3: 150742674-150742674 on Build GRCh38
4725971 c.427A>G p.T143A Chr.3: 150742689-150742689 on Build GRCh38
4548794 c.456G>A p.T152T Chr.3: 150742660-150742660 on Build GRCh38
4114414 c.490C>T p.R164C Chr.3: 150742626-150742626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148151216 Chr.3: 150742715-150742715 on Build GRCh38
rs546350530 Chr.3: 150742934-150742934 on Build GRCh38
rs762261885 Chr.3: 150742785-150742785 on Build GRCh38
rs373195810 Chr.3: 150742687-150742687 on Build GRCh38
rs751467793 Chr.3: 150742679-150742679 on Build GRCh38
rs1023174014 Chr.3: 150742887-150742887 on Build GRCh38
rs142360179 Chr.3: 150742660-150742660 on Build GRCh38
rs141944116 Chr.3: 150742920-150742920 on Build GRCh38
rs778141081 Chr.3: 150742709-150742709 on Build GRCh38
rs971824946 Chr.3: 150742942-150742942 on Build GRCh38
rs868711139 Chr.3: 150742694-150742694 on Build GRCh38
rs376697845 Chr.3: 150742765-150742765 on Build GRCh38
rs770320086 Chr.3: 150742857-150742857 on Build GRCh38
rs932540038 Chr.3: 150743079-150743079 on Build GRCh38
rs901959275 Chr.3: 150742764-150742764 on Build GRCh38
rs934636052 Chr.3: 150743080-150743080 on Build GRCh38
rs955137222 Chr.3: 150742789-150742789 on Build GRCh38
rs974771896 Chr.3: 150742910-150742910 on Build GRCh38
rs377755599 Chr.3: 150742706-150742706 on Build GRCh38
rs773132524 Chr.3: 150742782-150742782 on Build GRCh38
rs770988224 Chr.3: 150742723-150742723 on Build GRCh38
rs746424385 Chr.3: 150742739-150742739 on Build GRCh38
rs772432383 Chr.3: 150742745-150742745 on Build GRCh38
rs781081520 Chr.3: 150742688-150742688 on Build GRCh38
rs767595834 Chr.3: 150742737-150742737 on Build GRCh38
rs1026040250 Chr.3: 150742943-150742943 on Build GRCh38
rs1054802697 Chr.3: 150743032-150743032 on Build GRCh38
rs373248583 Chr.3: 150742638-150742638 on Build GRCh38
rs566281525 Chr.3: 150742944-150742944 on Build GRCh38
rs943292880 Chr.3: 150743026-150743026 on Build GRCh38
rs962904936 Chr.3: 150742871-150742871 on Build GRCh38
rs377407314 Chr.3: 150742661-150742661 on Build GRCh38
rs945415150 Chr.3: 150742669-150742669 on Build GRCh38
rs549604637 Chr.3: 150742693-150742693 on Build GRCh38
rs570756255 Chr.3: 150743063-150743063 on Build GRCh38
rs1053147459 Chr.3: 150743081-150743081 on Build GRCh38
rs910499841 Chr.3: 150742999-150742999 on Build GRCh38
rs774748783 Chr.3: 150742731-150742731 on Build GRCh38
rs1011674935 Chr.3: 150742883-150742883 on Build GRCh38
rs370922801 Chr.3: 150743000-150743000 on Build GRCh38
rs761003951 Chr.3: 150742913-150742913 on Build GRCh38
rs775886677 Chr.3: 150742747-150742747 on Build GRCh38
rs114684938 Chr.3: 150742819-150742819 on Build GRCh38
rs910438075 Chr.3: 150742710-150742710 on Build GRCh38
rs984583758 Chr.3: 150742977-150742977 on Build GRCh38
rs548880201 Chr.3: 150742873-150742873 on Build GRCh38
rs960760682 Chr.3: 150742936-150742936 on Build GRCh38
rs924474054 Chr.3: 150743005-150743005 on Build GRCh38
rs923280627 Chr.3: 150743077-150743077 on Build GRCh38
rs992174950 Chr.3: 150742933-150742933 on Build GRCh38
rs978136595 Chr.3: 150742970-150742971 on Build GRCh38
rs372893534 Chr.3: 150743004-150743004 on Build GRCh38
rs766834931 Chr.3: 150742804-150742806 on Build GRCh38
rs976519423 Chr.3: 150743042-150743042 on Build GRCh38
rs529138780 Chr.3: 150742917-150742917 on Build GRCh38
rs1015724302 Chr.3: 150742845-150742845 on Build GRCh38
rs761497923 Chr.3: 150742637-150742637 on Build GRCh38
rs1040373777 Chr.3: 150742726-150742726 on Build GRCh38

View Full Product Details