Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151328694-151329239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
UniGene
Hs.744234
Gene Chromosome Location
Chr.3: 151085665-151436677 on Build GRCh38
Transcripts
NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1 NM_176894.2 XM_006713664.1

Gene Symbol
P2RY13
Entrez Gene ID
53829
Gene Name
purinergic receptor P2Y13
Gene Aliases
FKSG77, GPCR1, GPR86, GPR94, P2Y13, SP174
UniGene
Hs.546396
Gene Chromosome Location
Chr.3: 151326308-151329549 on Build GRCh38
Transcripts
NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1 NM_176894.2 XM_006713664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7229676 c.58C>A p.R20R Chr.3: 151328935-151328935 on Build GRCh38
4710675 c.274G>C p.A92P Chr.3: 151328719-151328719 on Build GRCh38
3846510 c.137T>G p.V46G Chr.3: 151328919-151328919 on Build GRCh38
7169211 c.105G>A p.L35L Chr.3: 151328888-151328888 on Build GRCh38
4826514 c.296C>T p.S99L Chr.3: 151328697-151328697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749980757 Chr.3: 151328997-151328997 on Build GRCh38
rs774813657 Chr.3: 151328842-151328842 on Build GRCh38
rs940470516 Chr.3: 151329102-151329102 on Build GRCh38
rs139399025 Chr.3: 151328800-151328800 on Build GRCh38
rs560572748 Chr.3: 151328831-151328831 on Build GRCh38
rs546426718 Chr.3: 151329089-151329089 on Build GRCh38
rs531906927 Chr.3: 151329178-151329178 on Build GRCh38
rs755787621 Chr.3: 151328720-151328720 on Build GRCh38
rs577175996 Chr.3: 151329054-151329054 on Build GRCh38
rs936533736 Chr.3: 151328884-151328884 on Build GRCh38
rs1037434573 Chr.3: 151329192-151329192 on Build GRCh38
rs772423641 Chr.3: 151328980-151328980 on Build GRCh38
rs754884650 Chr.3: 151329033-151329033 on Build GRCh38
rs746749067 Chr.3: 151328793-151328793 on Build GRCh38
rs937256742 Chr.3: 151329186-151329186 on Build GRCh38
rs141361811 Chr.3: 151328990-151328990 on Build GRCh38
rs759959036 Chr.3: 151328846-151328849 on Build GRCh38
rs769029900 Chr.3: 151328799-151328799 on Build GRCh38
rs916568072 Chr.3: 151329204-151329204 on Build GRCh38
rs771476846 Chr.3: 151328963-151328963 on Build GRCh38
rs748249195 Chr.3: 151328814-151328814 on Build GRCh38
rs868119745 Chr.3: 151328883-151328883 on Build GRCh38
rs371578438 Chr.3: 151328952-151328952 on Build GRCh38
rs754757932 Chr.3: 151328935-151328935 on Build GRCh38
rs753351579 Chr.3: 151328768-151328768 on Build GRCh38
rs940453885 Chr.3: 151329135-151329135 on Build GRCh38
rs767769751 Chr.3: 151329012-151329013 on Build GRCh38
rs372729106 Chr.3: 151328880-151328880 on Build GRCh38
rs776306831 Chr.3: 151328988-151328988 on Build GRCh38
rs6794283 Chr.3: 151329206-151329206 on Build GRCh38
rs753476138 Chr.3: 151328858-151328858 on Build GRCh38
rs758381185 Chr.3: 151328902-151328902 on Build GRCh38
rs764745405 Chr.3: 151328995-151328995 on Build GRCh38
rs779815610 Chr.3: 151328908-151328908 on Build GRCh38
rs759831858 Chr.3: 151328848-151328848 on Build GRCh38
rs867914537 Chr.3: 151328915-151328915 on Build GRCh38
rs973183139 Chr.3: 151329146-151329146 on Build GRCh38
rs1055009153 Chr.3: 151328789-151328789 on Build GRCh38
rs781100764 Chr.3: 151329035-151329035 on Build GRCh38
rs756701413 Chr.3: 151328771-151328771 on Build GRCh38
rs1046083571 Chr.3: 151329069-151329069 on Build GRCh38
rs746804027 Chr.3: 151328928-151328928 on Build GRCh38
rs766350629 Chr.3: 151329007-151329007 on Build GRCh38
rs367643722 Chr.3: 151329038-151329038 on Build GRCh38
rs1055421055 Chr.3: 151328892-151328892 on Build GRCh38
rs753432791 Chr.3: 151329035-151329039 on Build GRCh38
rs760223982 Chr.3: 151329020-151329030 on Build GRCh38
rs763699238 Chr.3: 151328723-151328723 on Build GRCh38
rs756829710 Chr.3: 151328860-151328860 on Build GRCh38
rs958243867 Chr.3: 151329034-151329034 on Build GRCh38
rs766749093 Chr.3: 151328839-151328839 on Build GRCh38
rs563195353 Chr.3: 151329092-151329092 on Build GRCh38
rs1055671953 Chr.3: 151329190-151329190 on Build GRCh38
rs750375754 Chr.3: 151328885-151328885 on Build GRCh38
rs751524418 Chr.3: 151329015-151329015 on Build GRCh38
rs150522091 Chr.3: 151328788-151328788 on Build GRCh38
rs3732757 Chr.3: 151328816-151328816 on Build GRCh38
rs749193587 Chr.3: 151328970-151328970 on Build GRCh38
rs770957622 Chr.3: 151329014-151329014 on Build GRCh38
rs987614742 Chr.3: 151329099-151329099 on Build GRCh38
rs920431477 Chr.3: 151329148-151329153 on Build GRCh38
rs967488878 Chr.3: 151328794-151328794 on Build GRCh38
rs886178605 Chr.3: 151329098-151329098 on Build GRCh38
rs763750367 Chr.3: 151328849-151328849 on Build GRCh38
rs773655982 Chr.3: 151328819-151328819 on Build GRCh38
rs764774867 Chr.3: 151328867-151328867 on Build GRCh38
rs781282890 Chr.3: 151328937-151328937 on Build GRCh38
rs779876492 Chr.3: 151328792-151328792 on Build GRCh38
rs752434984 Chr.3: 151329018-151329018 on Build GRCh38
rs749291316 Chr.3: 151328957-151328957 on Build GRCh38
rs987493533 Chr.3: 151328749-151328749 on Build GRCh38
rs763793559 Chr.3: 151329024-151329024 on Build GRCh38
rs146597143 Chr.3: 151328944-151328944 on Build GRCh38
rs899300964 Chr.3: 151329203-151329203 on Build GRCh38
rs144128158 Chr.3: 151328718-151328718 on Build GRCh38
rs773366464 Chr.3: 151328953-151328953 on Build GRCh38
rs747246139 Chr.3: 151328791-151328791 on Build GRCh38
rs759952511 Chr.3: 151328975-151328975 on Build GRCh38
rs902661215 Chr.3: 151328896-151328896 on Build GRCh38
rs954437328 Chr.3: 151329041-151329041 on Build GRCh38
rs774782565 Chr.3: 151328964-151328964 on Build GRCh38
rs778773836 Chr.3: 151328785-151328785 on Build GRCh38
rs369369502 Chr.3: 151329101-151329101 on Build GRCh38

View Full Product Details