Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 156291275-156291770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB1
Entrez Gene ID
7881
Gene Name
potassium voltage-gated channel subfamily A member regulatory beta subunit 1
Gene Aliases
AKR6A3, KCNA1B, KV-BETA-1, Kvb1.3, hKvBeta3, hKvb3
UniGene
Hs.654519
Gene Chromosome Location
Chr.3: 156118211-156540603 on Build GRCh38
Transcripts
NM_001308217.1 NM_001308222.1 NM_003471.3 NM_172159.3 NM_172160.2 XM_017007171.1 XM_017007174.1

SNP Detail

SNP ID Chromosome Location
rs1021516032 Chr.3: 156291482-156291482 on Build GRCh38
rs948997278 Chr.3: 156291363-156291363 on Build GRCh38
rs1022848269 Chr.3: 156291449-156291449 on Build GRCh38
rs887458785 Chr.3: 156291395-156291395 on Build GRCh38
rs987191705 Chr.3: 156291502-156291502 on Build GRCh38
rs1004274760 Chr.3: 156291393-156291393 on Build GRCh38
rs948862050 Chr.3: 156291574-156291574 on Build GRCh38
rs773074771 Chr.3: 156291538-156291538 on Build GRCh38
rs910173500 Chr.3: 156291580-156291580 on Build GRCh38
rs564452320 Chr.3: 156291616-156291616 on Build GRCh38
rs987136323 Chr.3: 156291745-156291745 on Build GRCh38
rs938810560 Chr.3: 156291519-156291519 on Build GRCh38
rs765772119 Chr.3: 156291401-156291401 on Build GRCh38
rs894623871 Chr.3: 156291360-156291360 on Build GRCh38
rs1057337838 Chr.3: 156291332-156291332 on Build GRCh38
rs921518763 Chr.3: 156291564-156291564 on Build GRCh38
rs959869916 Chr.3: 156291408-156291408 on Build GRCh38
rs1036058192 Chr.3: 156291638-156291638 on Build GRCh38
rs1012747447 Chr.3: 156291649-156291649 on Build GRCh38
rs552425081 Chr.3: 156291487-156291487 on Build GRCh38
rs887188740 Chr.3: 156291346-156291346 on Build GRCh38
rs1048361852 Chr.3: 156291578-156291578 on Build GRCh38
rs999168480 Chr.3: 156291744-156291744 on Build GRCh38
rs934385385 Chr.3: 156291492-156291492 on Build GRCh38
rs887136659 Chr.3: 156291591-156291591 on Build GRCh38
rs977508867 Chr.3: 156291299-156291299 on Build GRCh38
rs993304203 Chr.3: 156291535-156291535 on Build GRCh38
rs1026976893 Chr.3: 156291344-156291344 on Build GRCh38
rs376778469 Chr.3: 156291651-156291651 on Build GRCh38
rs748354094 Chr.3: 156291418-156291418 on Build GRCh38
rs991735662 Chr.3: 156291438-156291438 on Build GRCh38
rs1048604524 Chr.3: 156291380-156291380 on Build GRCh38
rs938763864 Chr.3: 156291329-156291329 on Build GRCh38
rs968659114 Chr.3: 156291742-156291742 on Build GRCh38
rs866794558 Chr.3: 156291317-156291317 on Build GRCh38
rs969076211 Chr.3: 156291450-156291450 on Build GRCh38
rs7620003 Chr.3: 156291720-156291720 on Build GRCh38
rs769726382 Chr.3: 156291459-156291459 on Build GRCh38
rs181223496 Chr.3: 156291674-156291674 on Build GRCh38
rs997661859 Chr.3: 156291435-156291435 on Build GRCh38
rs1004180953 Chr.3: 156291618-156291618 on Build GRCh38
rs16825964 Chr.3: 156291631-156291631 on Build GRCh38
rs895898449 Chr.3: 156291642-156291642 on Build GRCh38
rs944179785 Chr.3: 156291554-156291554 on Build GRCh38
rs551099358 Chr.3: 156291732-156291732 on Build GRCh38
rs931604346 Chr.3: 156291569-156291569 on Build GRCh38
rs1040282974 Chr.3: 156291563-156291563 on Build GRCh38
rs924600771 Chr.3: 156291514-156291514 on Build GRCh38
rs1031607313 Chr.3: 156291715-156291715 on Build GRCh38
rs977490627 Chr.3: 156291508-156291508 on Build GRCh38
rs1000142927 Chr.3: 156291664-156291664 on Build GRCh38
rs924297489 Chr.3: 156291469-156291469 on Build GRCh38
rs968646908 Chr.3: 156291656-156291656 on Build GRCh38
rs924681963 Chr.3: 156291310-156291310 on Build GRCh38
rs530427724 Chr.3: 156291352-156291352 on Build GRCh38
rs1005801913 Chr.3: 156291399-156291399 on Build GRCh38

View Full Product Details