Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 159273506-159274002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCJ-SCHIP1
Entrez Gene ID
100505385
Gene Name
IQCJ-SCHIP1 readthrough
Gene Aliases
-
UniGene
Hs.134665
Gene Chromosome Location
Chr.3: 159069252-159897366 on Build GRCh38
Transcripts
NM_001197113.1 NM_001197114.1 NM_001197107.1 NM_001197108.1 NM_014575.3

Gene Symbol
SCHIP1
Entrez Gene ID
29970
Gene Name
schwannomin interacting protein 1
Gene Aliases
SCHIP-1
UniGene
Hs.134665
Gene Chromosome Location
Chr.3: 159273247-159897366 on Build GRCh38
Transcripts
NM_001197113.1 NM_001197114.1 NM_001197107.1 NM_001197108.1 NM_014575.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5675751 c.16C>G p.Q6E Chr.3: 159273836-159273836 on Build GRCh38
323212 c.19C>A p.R7S Chr.3: 159273839-159273839 on Build GRCh38
1632946 c.5A>T p.E2V Chr.3: 159273825-159273825 on Build GRCh38
5820238 c.45G>A p.Q15Q Chr.3: 159273865-159273865 on Build GRCh38
1670674 c.28A>C p.T10P Chr.3: 159273848-159273848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995639105 Chr.3: 159273579-159273579 on Build GRCh38
rs754769562 Chr.3: 159273794-159273794 on Build GRCh38
rs765524157 Chr.3: 159273886-159273886 on Build GRCh38
rs140796600 Chr.3: 159273820-159273820 on Build GRCh38
rs762014363 Chr.3: 159273880-159273880 on Build GRCh38
rs770025184 Chr.3: 159273732-159273732 on Build GRCh38
rs935681412 Chr.3: 159273780-159273780 on Build GRCh38
rs370535535 Chr.3: 159273755-159273755 on Build GRCh38
rs935417246 Chr.3: 159273895-159273895 on Build GRCh38
rs1042493169 Chr.3: 159273605-159273605 on Build GRCh38
rs958387500 Chr.3: 159273707-159273707 on Build GRCh38
rs756866549 Chr.3: 159273821-159273821 on Build GRCh38
rs2621285 Chr.3: 159273935-159273935 on Build GRCh38
rs886390615 Chr.3: 159273954-159273954 on Build GRCh38
rs191127215 Chr.3: 159273920-159273920 on Build GRCh38
rs965851359 Chr.3: 159273720-159273720 on Build GRCh38
rs777209063 Chr.3: 159273815-159273815 on Build GRCh38
rs945627997 Chr.3: 159273857-159273857 on Build GRCh38
rs771538318 Chr.3: 159273797-159273798 on Build GRCh38
rs776056798 Chr.3: 159273845-159273845 on Build GRCh38
rs774377202 Chr.3: 159273661-159273661 on Build GRCh38
rs34135941 Chr.3: 159273599-159273599 on Build GRCh38
rs3732847 Chr.3: 159273764-159273764 on Build GRCh38
rs545336019 Chr.3: 159273900-159273900 on Build GRCh38
rs772375075 Chr.3: 159273842-159273842 on Build GRCh38
rs368112222 Chr.3: 159273922-159273922 on Build GRCh38
rs184725132 Chr.3: 159273926-159273926 on Build GRCh38
rs898656341 Chr.3: 159273574-159273574 on Build GRCh38
rs767152416 Chr.3: 159273916-159273916 on Build GRCh38
rs143326283 Chr.3: 159273860-159273860 on Build GRCh38
rs563844530 Chr.3: 159273556-159273556 on Build GRCh38
rs906679635 Chr.3: 159273884-159273884 on Build GRCh38
rs1047198420 Chr.3: 159273962-159273962 on Build GRCh38
rs778580906 Chr.3: 159273824-159273824 on Build GRCh38
rs751358761 Chr.3: 159273790-159273790 on Build GRCh38
rs959617750 Chr.3: 159273678-159273678 on Build GRCh38
rs774925078 Chr.3: 159273851-159273851 on Build GRCh38
rs1009697823 Chr.3: 159273590-159273590 on Build GRCh38
rs767477070 Chr.3: 159273798-159273798 on Build GRCh38
rs143207517 Chr.3: 159273849-159273849 on Build GRCh38
rs149218707 Chr.3: 159273852-159273852 on Build GRCh38
rs1033714639 Chr.3: 159273673-159273673 on Build GRCh38
rs528483159 Chr.3: 159273713-159273713 on Build GRCh38
rs75319750 Chr.3: 159273638-159273638 on Build GRCh38
rs989742646 Chr.3: 159273807-159273807 on Build GRCh38
rs776801024 Chr.3: 159273878-159273878 on Build GRCh38
rs531196459 Chr.3: 159273571-159273571 on Build GRCh38
rs939474993 Chr.3: 159273899-159273899 on Build GRCh38
rs924456094 Chr.3: 159273864-159273864 on Build GRCh38
rs932944043 Chr.3: 159273554-159273555 on Build GRCh38
rs1035348246 Chr.3: 159273687-159273687 on Build GRCh38
rs756083722 Chr.3: 159273814-159273814 on Build GRCh38
rs983323691 Chr.3: 159273692-159273692 on Build GRCh38
rs535311586 Chr.3: 159273906-159273906 on Build GRCh38
rs145384643 Chr.3: 159273850-159273850 on Build GRCh38
rs1027586190 Chr.3: 159273681-159273681 on Build GRCh38
rs759969799 Chr.3: 159273852-159273865 on Build GRCh38
rs1004790036 Chr.3: 159273972-159273972 on Build GRCh38
rs767714446 Chr.3: 159273923-159273924 on Build GRCh38
rs746154959 Chr.3: 159273647-159273647 on Build GRCh38
rs1040019784 Chr.3: 159273542-159273542 on Build GRCh38
rs994815237 Chr.3: 159273679-159273679 on Build GRCh38
rs765577294 Chr.3: 159273786-159273786 on Build GRCh38
rs1003046781 Chr.3: 159273671-159273671 on Build GRCh38
rs1039733918 Chr.3: 159273924-159273924 on Build GRCh38
rs749054619 Chr.3: 159273589-159273589 on Build GRCh38
rs1004026004 Chr.3: 159273664-159273664 on Build GRCh38
rs752644749 Chr.3: 159273805-159273805 on Build GRCh38
rs570902051 Chr.3: 159273701-159273701 on Build GRCh38
rs746225987 Chr.3: 159273839-159273839 on Build GRCh38
rs768578443 Chr.3: 159273609-159273609 on Build GRCh38

View Full Product Details