Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 164996446-164996943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SI
Entrez Gene ID
6476
Gene Name
sucrase-isomaltase
Gene Aliases
-
UniGene
Hs.429596
Gene Chromosome Location
Chr.3: 164978898-165078495 on Build GRCh38
Transcripts
NM_001041.3 XM_011513078.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096661 c.4668G>T p.R1556S Chr.3: 164996559-164996559 on Build GRCh38
1265564 c.4643T>G p.L1548R Chr.3: 164996584-164996584 on Build GRCh38
5736815 c.4638G>A p.M1546I Chr.3: 164996589-164996589 on Build GRCh38
5651094 c.4623C>A p.L1541L Chr.3: 164996604-164996604 on Build GRCh38
1265566 c.4595G>T p.G1532V Chr.3: 164996632-164996632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147980815 Chr.3: 164996882-164996882 on Build GRCh38
rs780003350 Chr.3: 164996534-164996534 on Build GRCh38
rs557372761 Chr.3: 164996584-164996584 on Build GRCh38
rs1040060732 Chr.3: 164996765-164996765 on Build GRCh38
rs973915706 Chr.3: 164996729-164996729 on Build GRCh38
rs969368790 Chr.3: 164996529-164996529 on Build GRCh38
rs200851311 Chr.3: 164996521-164996521 on Build GRCh38
rs764325020 Chr.3: 164996698-164996698 on Build GRCh38
rs550813094 Chr.3: 164996872-164996872 on Build GRCh38
rs773712567 Chr.3: 164996513-164996513 on Build GRCh38
rs755731709 Chr.3: 164996753-164996753 on Build GRCh38
rs758042047 Chr.3: 164996780-164996783 on Build GRCh38
rs532955508 Chr.3: 164996916-164996916 on Build GRCh38
rs766266827 Chr.3: 164996717-164996717 on Build GRCh38
rs769560102 Chr.3: 164996563-164996563 on Build GRCh38
rs773712921 Chr.3: 164996566-164996566 on Build GRCh38
rs763616023 Chr.3: 164996515-164996515 on Build GRCh38
rs748923904 Chr.3: 164996661-164996661 on Build GRCh38
rs754961413 Chr.3: 164996736-164996736 on Build GRCh38
rs113969994 Chr.3: 164996711-164996711 on Build GRCh38
rs763366493 Chr.3: 164996574-164996574 on Build GRCh38
rs768226156 Chr.3: 164996506-164996506 on Build GRCh38
rs151234636 Chr.3: 164996541-164996541 on Build GRCh38
rs756341887 Chr.3: 164996600-164996600 on Build GRCh38
rs772665950 Chr.3: 164996815-164996815 on Build GRCh38
rs1039248856 Chr.3: 164996913-164996913 on Build GRCh38
rs766815320 Chr.3: 164996579-164996579 on Build GRCh38
rs901618532 Chr.3: 164996487-164996488 on Build GRCh38
rs934437430 Chr.3: 164996864-164996866 on Build GRCh38
rs747880728 Chr.3: 164996557-164996557 on Build GRCh38
rs751302984 Chr.3: 164996730-164996730 on Build GRCh38
rs539127421 Chr.3: 164996641-164996641 on Build GRCh38
rs114497045 Chr.3: 164996473-164996473 on Build GRCh38
rs759927786 Chr.3: 164996594-164996594 on Build GRCh38
rs577327619 Chr.3: 164996623-164996623 on Build GRCh38
rs768264563 Chr.3: 164996546-164996546 on Build GRCh38
rs779227756 Chr.3: 164996625-164996626 on Build GRCh38
rs745560836 Chr.3: 164996726-164996729 on Build GRCh38
rs767701775 Chr.3: 164996597-164996597 on Build GRCh38
rs775975573 Chr.3: 164996555-164996555 on Build GRCh38
rs779492314 Chr.3: 164996782-164996783 on Build GRCh38
rs377619918 Chr.3: 164996517-164996517 on Build GRCh38
rs900612391 Chr.3: 164996601-164996601 on Build GRCh38
rs779692980 Chr.3: 164996634-164996634 on Build GRCh38
rs950756075 Chr.3: 164996688-164996688 on Build GRCh38
rs769598593 Chr.3: 164996647-164996647 on Build GRCh38
rs199581970 Chr.3: 164996523-164996523 on Build GRCh38
rs561697975 Chr.3: 164996771-164996771 on Build GRCh38
rs868525953 Chr.3: 164996773-164996773 on Build GRCh38
rs760145273 Chr.3: 164996674-164996674 on Build GRCh38
rs1053379218 Chr.3: 164996877-164996877 on Build GRCh38
rs746546923 Chr.3: 164996785-164996785 on Build GRCh38
rs771364276 Chr.3: 164996663-164996663 on Build GRCh38
rs763589746 Chr.3: 164996588-164996588 on Build GRCh38
rs571920146 Chr.3: 164996530-164996530 on Build GRCh38
rs746151855 Chr.3: 164996810-164996810 on Build GRCh38
rs780817586 Chr.3: 164996644-164996644 on Build GRCh38
rs764235215 Chr.3: 164996609-164996609 on Build GRCh38
rs771169688 Chr.3: 164996520-164996520 on Build GRCh38
rs750249689 Chr.3: 164996610-164996610 on Build GRCh38
rs534673736 Chr.3: 164996497-164996497 on Build GRCh38
rs776979788 Chr.3: 164996531-164996531 on Build GRCh38
rs111418304 Chr.3: 164996790-164996790 on Build GRCh38
rs774573006 Chr.3: 164996670-164996670 on Build GRCh38
rs747607478 Chr.3: 164996645-164996645 on Build GRCh38
rs776299891 Chr.3: 164996507-164996507 on Build GRCh38
rs200559542 Chr.3: 164996696-164996696 on Build GRCh38
rs373292945 Chr.3: 164996504-164996504 on Build GRCh38
rs765739672 Chr.3: 164996509-164996509 on Build GRCh38
rs554576453 Chr.3: 164996502-164996502 on Build GRCh38
rs779347864 Chr.3: 164996804-164996804 on Build GRCh38
rs775551355 Chr.3: 164996692-164996692 on Build GRCh38
rs115606442 Chr.3: 164996656-164996656 on Build GRCh38
rs375448215 Chr.3: 164996471-164996471 on Build GRCh38
rs778738268 Chr.3: 164996524-164996524 on Build GRCh38
rs370402596 Chr.3: 164996738-164996738 on Build GRCh38
rs772401454 Chr.3: 164996678-164996678 on Build GRCh38
rs4855271 Chr.3: 164996744-164996744 on Build GRCh38
rs928338500 Chr.3: 164996802-164996802 on Build GRCh38
rs377168819 Chr.3: 164996525-164996525 on Build GRCh38
rs78437999 Chr.3: 164996508-164996508 on Build GRCh38
rs755033393 Chr.3: 164996519-164996519 on Build GRCh38
rs766839191 Chr.3: 164996518-164996518 on Build GRCh38
rs1040089426 Chr.3: 164996477-164996477 on Build GRCh38
rs749013790 Chr.3: 164996774-164996774 on Build GRCh38
rs866426226 Chr.3: 164996742-164996742 on Build GRCh38
rs984666217 Chr.3: 164996721-164996721 on Build GRCh38
rs569184256 Chr.3: 164996888-164996888 on Build GRCh38

View Full Product Details