Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 165772796-165773342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCHE
Entrez Gene ID
590
Gene Name
butyrylcholinesterase
Gene Aliases
CHE1, CHE2, E1
UniGene
Hs.420483
Gene Chromosome Location
Chr.3: 165772904-165837467 on Build GRCh38
Transcripts
XM_017006967.1

SNP Detail

SNP ID Chromosome Location
rs964093758 Chr.3: 165773034-165773034 on Build GRCh38
rs371032418 Chr.3: 165773094-165773094 on Build GRCh38
rs1030063230 Chr.3: 165773039-165773039 on Build GRCh38
rs896343496 Chr.3: 165773273-165773273 on Build GRCh38
rs199567692 Chr.3: 165773088-165773088 on Build GRCh38
rs1004477118 Chr.3: 165773244-165773244 on Build GRCh38
rs571279565 Chr.3: 165773172-165773172 on Build GRCh38
rs974264291 Chr.3: 165773292-165773292 on Build GRCh38
rs1013543031 Chr.3: 165773089-165773090 on Build GRCh38
rs1011402022 Chr.3: 165773201-165773201 on Build GRCh38
rs556141230 Chr.3: 165773250-165773250 on Build GRCh38
rs369786680 Chr.3: 165773067-165773067 on Build GRCh38
rs145327297 Chr.3: 165773243-165773243 on Build GRCh38
rs150581482 Chr.3: 165773133-165773133 on Build GRCh38
rs548870275 Chr.3: 165773062-165773062 on Build GRCh38
rs966014566 Chr.3: 165773191-165773191 on Build GRCh38
rs963978835 Chr.3: 165772820-165772820 on Build GRCh38
rs1042406 Chr.3: 165773176-165773176 on Build GRCh38
rs55765003 Chr.3: 165773192-165773192 on Build GRCh38
rs934217730 Chr.3: 165772867-165772867 on Build GRCh38
rs1045412725 Chr.3: 165773113-165773113 on Build GRCh38
rs886058152 Chr.3: 165772982-165772982 on Build GRCh38
rs144542474 Chr.3: 165772915-165772915 on Build GRCh38
rs999147516 Chr.3: 165772950-165772950 on Build GRCh38
rs185680601 Chr.3: 165772896-165772896 on Build GRCh38
rs1010952136 Chr.3: 165772995-165772995 on Build GRCh38
rs955663824 Chr.3: 165773041-165773041 on Build GRCh38
rs1044973756 Chr.3: 165773297-165773297 on Build GRCh38
rs975811165 Chr.3: 165772827-165772827 on Build GRCh38
rs193009639 Chr.3: 165772853-165772853 on Build GRCh38
rs941108112 Chr.3: 165772890-165772890 on Build GRCh38
rs1023573783 Chr.3: 165773095-165773096 on Build GRCh38
rs187886111 Chr.3: 165772927-165772927 on Build GRCh38
rs969849571 Chr.3: 165773309-165773309 on Build GRCh38
rs568512358 Chr.3: 165773085-165773085 on Build GRCh38
rs55729116 Chr.3: 165773097-165773097 on Build GRCh38
rs139885577 Chr.3: 165773202-165773202 on Build GRCh38
rs984516681 Chr.3: 165773236-165773236 on Build GRCh38
rs55849903 Chr.3: 165773033-165773033 on Build GRCh38
rs3836432 Chr.3: 165773088-165773089 on Build GRCh38
rs55753636 Chr.3: 165773101-165773101 on Build GRCh38
rs55781241 Chr.3: 165773181-165773181 on Build GRCh38
rs886058154 Chr.3: 165773208-165773208 on Build GRCh38
rs949290369 Chr.3: 165773289-165773289 on Build GRCh38
rs1015889129 Chr.3: 165773246-165773246 on Build GRCh38
rs757059420 Chr.3: 165772918-165772918 on Build GRCh38
rs567077263 Chr.3: 165773220-165773220 on Build GRCh38
rs3495 Chr.3: 165773193-165773193 on Build GRCh38
rs915705995 Chr.3: 165773099-165773099 on Build GRCh38
rs1036597352 Chr.3: 165773255-165773255 on Build GRCh38
rs916070916 Chr.3: 165773303-165773303 on Build GRCh38
rs77307863 Chr.3: 165772869-165772869 on Build GRCh38
rs1000822422 Chr.3: 165773315-165773315 on Build GRCh38
rs886058153 Chr.3: 165773145-165773145 on Build GRCh38
rs1031520845 Chr.3: 165772951-165772951 on Build GRCh38
rs529114188 Chr.3: 165773054-165773054 on Build GRCh38
rs865783912 Chr.3: 165772966-165772966 on Build GRCh38
rs940537185 Chr.3: 165772986-165772986 on Build GRCh38
rs760760763 Chr.3: 165773035-165773035 on Build GRCh38
rs1045808268 Chr.3: 165772932-165772932 on Build GRCh38
rs868606986 Chr.3: 165773130-165773130 on Build GRCh38
rs759407697 Chr.3: 165773278-165773278 on Build GRCh38
rs908713939 Chr.3: 165773076-165773076 on Build GRCh38
rs931650494 Chr.3: 165773207-165773207 on Build GRCh38
rs908676483 Chr.3: 165772877-165772877 on Build GRCh38
rs1019811739 Chr.3: 165773157-165773157 on Build GRCh38
rs899955848 Chr.3: 165773210-165773210 on Build GRCh38
rs981557321 Chr.3: 165773310-165773310 on Build GRCh38
rs1029682736 Chr.3: 165773228-165773228 on Build GRCh38
rs1038087200 Chr.3: 165772901-165772901 on Build GRCh38
rs892929809 Chr.3: 165772992-165772992 on Build GRCh38
rs962954716 Chr.3: 165773263-165773263 on Build GRCh38
rs148490591 Chr.3: 165772934-165772934 on Build GRCh38
rs746868218 Chr.3: 165773091-165773092 on Build GRCh38
rs887579734 Chr.3: 165772983-165772983 on Build GRCh38
rs1032409528 Chr.3: 165773109-165773112 on Build GRCh38

View Full Product Details