Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 165837095-165837596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCHE
Entrez Gene ID
590
Gene Name
butyrylcholinesterase
Gene Aliases
CHE1, CHE2, E1
UniGene
Hs.420483
Gene Chromosome Location
Chr.3: 165772904-165837467 on Build GRCh38
Transcripts
XM_017006967.1

SNP Detail

SNP ID Chromosome Location
rs1034355056 Chr.3: 165837281-165837281 on Build GRCh38
rs919794458 Chr.3: 165837531-165837531 on Build GRCh38
rs769880025 Chr.3: 165837425-165837425 on Build GRCh38
rs910460693 Chr.3: 165837472-165837472 on Build GRCh38
rs377763653 Chr.3: 165837504-165837504 on Build GRCh38
rs552858194 Chr.3: 165837275-165837275 on Build GRCh38
rs776581766 Chr.3: 165837272-165837272 on Build GRCh38
rs562054493 Chr.3: 165837437-165837437 on Build GRCh38
rs541713553 Chr.3: 165837409-165837409 on Build GRCh38
rs894094548 Chr.3: 165837559-165837559 on Build GRCh38
rs540455411 Chr.3: 165837137-165837137 on Build GRCh38
rs1021541736 Chr.3: 165837334-165837334 on Build GRCh38
rs919324795 Chr.3: 165837505-165837505 on Build GRCh38
rs1055379507 Chr.3: 165837556-165837556 on Build GRCh38
rs942016921 Chr.3: 165837124-165837124 on Build GRCh38
rs1036912388 Chr.3: 165837529-165837529 on Build GRCh38
rs1021320529 Chr.3: 165837343-165837343 on Build GRCh38
rs960960484 Chr.3: 165837257-165837257 on Build GRCh38
rs979623569 Chr.3: 165837358-165837358 on Build GRCh38
rs547400578 Chr.3: 165837436-165837436 on Build GRCh38
rs1034994711 Chr.3: 165837128-165837128 on Build GRCh38
rs953739417 Chr.3: 165837488-165837488 on Build GRCh38
rs938181815 Chr.3: 165837540-165837540 on Build GRCh38
rs906715815 Chr.3: 165837239-165837239 on Build GRCh38
rs1046585680 Chr.3: 165837220-165837220 on Build GRCh38
rs906668153 Chr.3: 165837539-165837539 on Build GRCh38
rs765253646 Chr.3: 165837463-165837463 on Build GRCh38
rs910522500 Chr.3: 165837122-165837122 on Build GRCh38
rs768588350 Chr.3: 165837362-165837362 on Build GRCh38
rs775391550 Chr.3: 165837551-165837551 on Build GRCh38
rs954712352 Chr.3: 165837455-165837455 on Build GRCh38
rs920998371 Chr.3: 165837395-165837395 on Build GRCh38
rs544254061 Chr.3: 165837476-165837476 on Build GRCh38
rs993654851 Chr.3: 165837211-165837211 on Build GRCh38
rs897948385 Chr.3: 165837183-165837183 on Build GRCh38
rs545578322 Chr.3: 165837165-165837165 on Build GRCh38
rs939942872 Chr.3: 165837525-165837525 on Build GRCh38
rs746031095 Chr.3: 165837345-165837345 on Build GRCh38
rs977205520 Chr.3: 165837428-165837428 on Build GRCh38
rs958327448 Chr.3: 165837293-165837293 on Build GRCh38
rs761912325 Chr.3: 165837462-165837462 on Build GRCh38
rs530515511 Chr.3: 165837461-165837461 on Build GRCh38
rs1126680 Chr.3: 165837337-165837337 on Build GRCh38
rs564154251 Chr.3: 165837512-165837512 on Build GRCh38
rs986450995 Chr.3: 165837497-165837497 on Build GRCh38
rs779676057 Chr.3: 165837360-165837360 on Build GRCh38
rs776506432 Chr.3: 165837402-165837402 on Build GRCh38
rs923105684 Chr.3: 165837453-165837453 on Build GRCh38

View Full Product Details