Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 170384441-170384954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SKIL
Entrez Gene ID
6498
Gene Name
SKI-like proto-oncogene
Gene Aliases
SNO, SnoA, SnoI, SnoN
UniGene
Hs.536655
Gene Chromosome Location
Chr.3: 170357678-170396849 on Build GRCh38
Transcripts
NM_001145097.2 NM_001145098.2 NM_001248008.1 NM_005414.4 XM_005247721.1 XM_006713735.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4525976 c.1366G>A p.V456I Chr.3: 170384702-170384702 on Build GRCh38
6868900 c.1418G>A p.C473Y Chr.3: 170384754-170384754 on Build GRCh38
3204268 c.1252G>T p.A418S Chr.3: 170384588-170384588 on Build GRCh38
445945 c.1219A>G p.K407E Chr.3: 170384555-170384555 on Build GRCh38
3204270 c.1424G>A p.R475H Chr.3: 170384760-170384760 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995350705 Chr.3: 170384745-170384745 on Build GRCh38
rs775225712 Chr.3: 170384734-170384734 on Build GRCh38
rs771731578 Chr.3: 170384586-170384586 on Build GRCh38
rs776253617 Chr.3: 170384768-170384768 on Build GRCh38
rs755304469 Chr.3: 170384552-170384552 on Build GRCh38
rs770940600 Chr.3: 170384583-170384583 on Build GRCh38
rs763571153 Chr.3: 170384492-170384492 on Build GRCh38
rs73879198 Chr.3: 170384920-170384920 on Build GRCh38
rs766994035 Chr.3: 170384793-170384793 on Build GRCh38
rs1004912705 Chr.3: 170384771-170384771 on Build GRCh38
rs775312128 Chr.3: 170384601-170384601 on Build GRCh38
rs759961591 Chr.3: 170384806-170384806 on Build GRCh38
rs755179957 Chr.3: 170384705-170384705 on Build GRCh38
rs766832508 Chr.3: 170384699-170384699 on Build GRCh38
rs764862156 Chr.3: 170384636-170384636 on Build GRCh38
rs373705278 Chr.3: 170384581-170384581 on Build GRCh38
rs944952146 Chr.3: 170384591-170384591 on Build GRCh38
rs959977152 Chr.3: 170384816-170384816 on Build GRCh38
rs763306917 Chr.3: 170384791-170384791 on Build GRCh38
rs781376763 Chr.3: 170384564-170384564 on Build GRCh38
rs780634860 Chr.3: 170384543-170384543 on Build GRCh38
rs753367994 Chr.3: 170384501-170384501 on Build GRCh38
rs1028115741 Chr.3: 170384896-170384896 on Build GRCh38
rs770840893 Chr.3: 170384574-170384574 on Build GRCh38
rs937772780 Chr.3: 170384679-170384679 on Build GRCh38
rs751927088 Chr.3: 170384799-170384799 on Build GRCh38
rs760271416 Chr.3: 170384609-170384609 on Build GRCh38
rs201142905 Chr.3: 170384754-170384754 on Build GRCh38
rs79718249 Chr.3: 170384826-170384826 on Build GRCh38
rs969892079 Chr.3: 170384894-170384894 on Build GRCh38
rs757742973 Chr.3: 170384504-170384504 on Build GRCh38
rs140141944 Chr.3: 170384716-170384716 on Build GRCh38
rs369623290 Chr.3: 170384508-170384508 on Build GRCh38
rs113703691 Chr.3: 170384782-170384782 on Build GRCh38
rs772831192 Chr.3: 170384788-170384788 on Build GRCh38
rs917520984 Chr.3: 170384599-170384599 on Build GRCh38
rs750743704 Chr.3: 170384510-170384510 on Build GRCh38
rs997941219 Chr.3: 170384839-170384839 on Build GRCh38
rs748457942 Chr.3: 170384708-170384708 on Build GRCh38
rs956048752 Chr.3: 170384929-170384929 on Build GRCh38
rs781574909 Chr.3: 170384707-170384707 on Build GRCh38
rs761594682 Chr.3: 170384775-170384775 on Build GRCh38
rs992722818 Chr.3: 170384820-170384820 on Build GRCh38
rs748239013 Chr.3: 170384567-170384567 on Build GRCh38
rs751998491 Chr.3: 170384704-170384704 on Build GRCh38
rs745769656 Chr.3: 170384720-170384720 on Build GRCh38
rs200162917 Chr.3: 170384547-170384547 on Build GRCh38
rs758986541 Chr.3: 170384662-170384662 on Build GRCh38
rs750891726 Chr.3: 170384646-170384646 on Build GRCh38
rs183339911 Chr.3: 170384521-170384521 on Build GRCh38
rs934692805 Chr.3: 170384570-170384570 on Build GRCh38
rs777635813 Chr.3: 170384695-170384695 on Build GRCh38
rs772019111 Chr.3: 170384731-170384731 on Build GRCh38
rs1015442699 Chr.3: 170384802-170384802 on Build GRCh38
rs979133924 Chr.3: 170384538-170384538 on Build GRCh38
rs186289738 Chr.3: 170384917-170384917 on Build GRCh38
rs761243511 Chr.3: 170384635-170384635 on Build GRCh38
rs776052409 Chr.3: 170384633-170384633 on Build GRCh38
rs768240946 Chr.3: 170384765-170384765 on Build GRCh38
rs763766901 Chr.3: 170384623-170384623 on Build GRCh38
rs760359226 Chr.3: 170384485-170384485 on Build GRCh38
rs764624982 Chr.3: 170384779-170384779 on Build GRCh38
rs200487211 Chr.3: 170384760-170384760 on Build GRCh38
rs770962705 Chr.3: 170384851-170384851 on Build GRCh38
rs954055279 Chr.3: 170384906-170384906 on Build GRCh38
rs904902156 Chr.3: 170384656-170384656 on Build GRCh38

View Full Product Details