Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 171007006-171007504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A2
Entrez Gene ID
6514
Gene Name
solute carrier family 2 member 2
Gene Aliases
GLUT2
UniGene
Hs.167584
Gene Chromosome Location
Chr.3: 170996341-171026979 on Build GRCh38
Transcripts
NM_000340.1 NM_001278658.1 NM_001278659.1 XM_011513087.2 XM_011513089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4501430 c.588C>T p.I196I Chr.3: 171007172-171007172 on Build GRCh38
4501975 c.601C>T p.L201F Chr.3: 171007159-171007159 on Build GRCh38
76469 c.589G>A p.V197I Chr.3: 171007171-171007171 on Build GRCh38
3590150 c.610C>T p.Q204* Chr.3: 171007150-171007150 on Build GRCh38
4115201 c.547G>A p.A183T Chr.3: 171007213-171007213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965891144 Chr.3: 171007290-171007290 on Build GRCh38
rs374173789 Chr.3: 171007299-171007299 on Build GRCh38
rs768979137 Chr.3: 171007472-171007474 on Build GRCh38
rs773145667 Chr.3: 171007313-171007313 on Build GRCh38
rs992348207 Chr.3: 171007446-171007446 on Build GRCh38
rs913895368 Chr.3: 171007434-171007434 on Build GRCh38
rs974686217 Chr.3: 171007039-171007039 on Build GRCh38
rs776498787 Chr.3: 171007194-171007194 on Build GRCh38
rs754220999 Chr.3: 171007264-171007264 on Build GRCh38
rs8192675 Chr.3: 171007094-171007094 on Build GRCh38
rs921828496 Chr.3: 171007050-171007050 on Build GRCh38
rs567533040 Chr.3: 171007124-171007124 on Build GRCh38
rs144822218 Chr.3: 171007231-171007231 on Build GRCh38
rs540342233 Chr.3: 171007088-171007088 on Build GRCh38
rs554772091 Chr.3: 171007443-171007443 on Build GRCh38
rs192897480 Chr.3: 171007336-171007336 on Build GRCh38
rs1032368934 Chr.3: 171007417-171007417 on Build GRCh38
rs773878999 Chr.3: 171007214-171007214 on Build GRCh38
rs731207 Chr.3: 171007327-171007327 on Build GRCh38
rs936703266 Chr.3: 171007141-171007141 on Build GRCh38
rs533366392 Chr.3: 171007040-171007040 on Build GRCh38
rs374624284 Chr.3: 171007172-171007172 on Build GRCh38
rs960256750 Chr.3: 171007047-171007047 on Build GRCh38
rs778281228 Chr.3: 171007388-171007388 on Build GRCh38
rs759604541 Chr.3: 171007115-171007115 on Build GRCh38
rs980798014 Chr.3: 171007401-171007401 on Build GRCh38
rs751065450 Chr.3: 171007154-171007154 on Build GRCh38
rs555757895 Chr.3: 171007078-171007078 on Build GRCh38
rs752305211 Chr.3: 171007232-171007232 on Build GRCh38
rs139187869 Chr.3: 171007121-171007121 on Build GRCh38
rs776566910 Chr.3: 171007117-171007117 on Build GRCh38
rs760095835 Chr.3: 171007251-171007251 on Build GRCh38
rs779065938 Chr.3: 171007180-171007180 on Build GRCh38
rs968335898 Chr.3: 171007352-171007352 on Build GRCh38
rs1022719266 Chr.3: 171007375-171007375 on Build GRCh38
rs774460489 Chr.3: 171007114-171007114 on Build GRCh38
rs757579143 Chr.3: 171007268-171007268 on Build GRCh38
rs988157545 Chr.3: 171007424-171007424 on Build GRCh38
rs762632420 Chr.3: 171007138-171007138 on Build GRCh38
rs750264208 Chr.3: 171007134-171007134 on Build GRCh38
rs766011603 Chr.3: 171007151-171007151 on Build GRCh38
rs925258113 Chr.3: 171007140-171007140 on Build GRCh38
rs528729853 Chr.3: 171007163-171007163 on Build GRCh38
rs745907191 Chr.3: 171007184-171007184 on Build GRCh38
rs747156256 Chr.3: 171007303-171007303 on Build GRCh38
rs867922504 Chr.3: 171007065-171007065 on Build GRCh38
rs121909741 Chr.3: 171007171-171007171 on Build GRCh38
rs759047405 Chr.3: 171007230-171007230 on Build GRCh38
rs944269745 Chr.3: 171007465-171007469 on Build GRCh38
rs1019696977 Chr.3: 171007234-171007234 on Build GRCh38
rs748052042 Chr.3: 171007198-171007198 on Build GRCh38
rs967725498 Chr.3: 171007054-171007054 on Build GRCh38
rs771799491 Chr.3: 171007203-171007203 on Build GRCh38
rs200213178 Chr.3: 171007201-171007201 on Build GRCh38
rs779710728 Chr.3: 171007102-171007102 on Build GRCh38
rs918121086 Chr.3: 171007136-171007136 on Build GRCh38
rs958397917 Chr.3: 171007418-171007418 on Build GRCh38
rs769704150 Chr.3: 171007311-171007311 on Build GRCh38
rs368626129 Chr.3: 171007213-171007213 on Build GRCh38
rs373989348 Chr.3: 171007116-171007116 on Build GRCh38
rs746096379 Chr.3: 171007281-171007281 on Build GRCh38
rs747576729 Chr.3: 171007392-171007392 on Build GRCh38
rs771182536 Chr.3: 171007173-171007173 on Build GRCh38
rs979124233 Chr.3: 171007381-171007381 on Build GRCh38
rs149460434 Chr.3: 171007167-171007167 on Build GRCh38
rs568964087 Chr.3: 171007205-171007205 on Build GRCh38
rs764767259 Chr.3: 171007261-171007261 on Build GRCh38
rs772071162 Chr.3: 171007193-171007193 on Build GRCh38
rs779128131 Chr.3: 171007275-171007275 on Build GRCh38
rs5404 Chr.3: 171007166-171007166 on Build GRCh38
rs370765080 Chr.3: 171007296-171007296 on Build GRCh38

View Full Product Details