Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183192911-183193338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCF2L2
Entrez Gene ID
23101
Gene Name
MCF.2 cell line derived transforming sequence-like 2
Gene Aliases
ARHGEF22
UniGene
Hs.208267
Gene Chromosome Location
Chr.3: 183176116-183428990 on Build GRCh38
Transcripts
NM_015078.3 XM_011512585.2 XM_011512586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4442151 c.2992G>A p.A998T Chr.3: 183193023-183193023 on Build GRCh38
7376618 c.2941G>A p.G981R Chr.3: 183193074-183193074 on Build GRCh38
6231667 c.2933C>T p.A978V Chr.3: 183193082-183193082 on Build GRCh38
5534368 c.2975C>T p.T992I Chr.3: 183193040-183193040 on Build GRCh38
6500261 c.2939C>G p.S980C Chr.3: 183193076-183193076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033448483 Chr.3: 183193201-183193201 on Build GRCh38
rs143707041 Chr.3: 183193071-183193071 on Build GRCh38
rs745792137 Chr.3: 183192971-183192971 on Build GRCh38
rs184418549 Chr.3: 183193145-183193145 on Build GRCh38
rs535533413 Chr.3: 183193024-183193024 on Build GRCh38
rs546962890 Chr.3: 183192940-183192940 on Build GRCh38
rs1000081085 Chr.3: 183193189-183193189 on Build GRCh38
rs766939202 Chr.3: 183193034-183193034 on Build GRCh38
rs781520060 Chr.3: 183192954-183192954 on Build GRCh38
rs929056860 Chr.3: 183193072-183193072 on Build GRCh38
rs369324763 Chr.3: 183193020-183193020 on Build GRCh38
rs994365374 Chr.3: 183193307-183193307 on Build GRCh38
rs746819406 Chr.3: 183193082-183193082 on Build GRCh38
rs751245719 Chr.3: 183193271-183193271 on Build GRCh38
rs199788516 Chr.3: 183193136-183193136 on Build GRCh38
rs1014118451 Chr.3: 183193131-183193131 on Build GRCh38
rs752000365 Chr.3: 183193037-183193037 on Build GRCh38
rs769392725 Chr.3: 183193117-183193117 on Build GRCh38
rs747208614 Chr.3: 183193158-183193158 on Build GRCh38
rs111635280 Chr.3: 183193142-183193142 on Build GRCh38
rs979893519 Chr.3: 183193205-183193205 on Build GRCh38
rs767869677 Chr.3: 183193056-183193056 on Build GRCh38
rs751877557 Chr.3: 183192951-183192951 on Build GRCh38
rs368677502 Chr.3: 183193173-183193173 on Build GRCh38
rs758247321 Chr.3: 183192982-183192982 on Build GRCh38
rs141274500 Chr.3: 183193068-183193068 on Build GRCh38
rs771565282 Chr.3: 183193132-183193132 on Build GRCh38
rs748764030 Chr.3: 183193119-183193119 on Build GRCh38
rs779928412 Chr.3: 183193075-183193075 on Build GRCh38
rs534369600 Chr.3: 183193220-183193220 on Build GRCh38
rs752831103 Chr.3: 183192959-183192959 on Build GRCh38
rs146776367 Chr.3: 183193074-183193074 on Build GRCh38
rs778970330 Chr.3: 183192970-183192970 on Build GRCh38
rs911502211 Chr.3: 183192949-183192949 on Build GRCh38
rs772617781 Chr.3: 183193118-183193118 on Build GRCh38
rs747722984 Chr.3: 183193115-183193115 on Build GRCh38
rs779803998 Chr.3: 183192984-183192984 on Build GRCh38
rs955989541 Chr.3: 183193089-183193089 on Build GRCh38
rs780962991 Chr.3: 183193113-183193113 on Build GRCh38
rs376483334 Chr.3: 183193016-183193016 on Build GRCh38
rs940366143 Chr.3: 183193283-183193283 on Build GRCh38
rs867725556 Chr.3: 183193208-183193208 on Build GRCh38
rs927070081 Chr.3: 183193211-183193211 on Build GRCh38
rs764050529 Chr.3: 183193070-183193070 on Build GRCh38
rs752853028 Chr.3: 183193058-183193058 on Build GRCh38
rs372015806 Chr.3: 183192998-183192998 on Build GRCh38
rs988759973 Chr.3: 183193252-183193252 on Build GRCh38
rs753916499 Chr.3: 183192994-183192997 on Build GRCh38
rs907491755 Chr.3: 183193238-183193238 on Build GRCh38
rs759916921 Chr.3: 183193055-183193055 on Build GRCh38
rs956037176 Chr.3: 183193212-183193212 on Build GRCh38
rs558522954 Chr.3: 183193150-183193150 on Build GRCh38
rs771896511 Chr.3: 183192981-183192981 on Build GRCh38
rs746758021 Chr.3: 183192989-183192989 on Build GRCh38
rs776089897 Chr.3: 183193006-183193006 on Build GRCh38
rs760039661 Chr.3: 183193146-183193146 on Build GRCh38
rs902047561 Chr.3: 183193003-183193003 on Build GRCh38
rs568506060 Chr.3: 183192962-183192962 on Build GRCh38
rs754754655 Chr.3: 183193088-183193088 on Build GRCh38
rs763343794 Chr.3: 183193025-183193025 on Build GRCh38
rs576780944 Chr.3: 183193154-183193154 on Build GRCh38

View Full Product Details