Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 183697571-183698060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YEATS2
Entrez Gene ID
55689
Gene Name
YEATS domain containing 2
Gene Aliases
-
UniGene
Hs.632575
Gene Chromosome Location
Chr.3: 183697797-183812625 on Build GRCh38
Transcripts
NM_018023.4 XM_005247597.2 XM_011512965.1 XM_011512966.1 XM_017006812.1 XM_017006813.1 XM_017006814.1

SNP Detail

SNP ID Chromosome Location
rs887682222 Chr.3: 183697794-183697794 on Build GRCh38
rs967110722 Chr.3: 183698009-183698009 on Build GRCh38
rs995374020 Chr.3: 183697632-183697632 on Build GRCh38
rs941827917 Chr.3: 183698037-183698037 on Build GRCh38
rs893281249 Chr.3: 183697977-183697977 on Build GRCh38
rs994268865 Chr.3: 183697816-183697816 on Build GRCh38
rs957062208 Chr.3: 183697771-183697771 on Build GRCh38
rs1036662096 Chr.3: 183697793-183697804 on Build GRCh38
rs1014254464 Chr.3: 183697995-183697995 on Build GRCh38
rs1018474182 Chr.3: 183697973-183697973 on Build GRCh38
rs985730976 Chr.3: 183697769-183697769 on Build GRCh38
rs544021013 Chr.3: 183697684-183697684 on Build GRCh38
rs941056809 Chr.3: 183697773-183697786 on Build GRCh38
rs578218018 Chr.3: 183697738-183697739 on Build GRCh38
rs958465548 Chr.3: 183698022-183698022 on Build GRCh38
rs955691635 Chr.3: 183697746-183697746 on Build GRCh38
rs951154002 Chr.3: 183697647-183697647 on Build GRCh38
rs896926159 Chr.3: 183697869-183697889 on Build GRCh38
rs909026125 Chr.3: 183697874-183697874 on Build GRCh38
rs370910274 Chr.3: 183697926-183697926 on Build GRCh38
rs895190503 Chr.3: 183697695-183697695 on Build GRCh38
rs991170050 Chr.3: 183698026-183698026 on Build GRCh38
rs560093566 Chr.3: 183697876-183697876 on Build GRCh38
rs945836278 Chr.3: 183697775-183697775 on Build GRCh38
rs544805775 Chr.3: 183697846-183697846 on Build GRCh38
rs866233813 Chr.3: 183697908-183697908 on Build GRCh38
rs114161963 Chr.3: 183698006-183698006 on Build GRCh38
rs955608524 Chr.3: 183697833-183697833 on Build GRCh38
rs1038351304 Chr.3: 183697804-183697804 on Build GRCh38
rs909055902 Chr.3: 183698029-183698029 on Build GRCh38
rs1025491223 Chr.3: 183697638-183697638 on Build GRCh38
rs573451909 Chr.3: 183697679-183697679 on Build GRCh38
rs1005531839 Chr.3: 183697796-183697796 on Build GRCh38
rs561657797 Chr.3: 183697655-183697655 on Build GRCh38
rs928365175 Chr.3: 183697847-183697847 on Build GRCh38
rs9869779 Chr.3: 183697713-183697713 on Build GRCh38
rs1055603240 Chr.3: 183697690-183697690 on Build GRCh38
rs991263450 Chr.3: 183697872-183697872 on Build GRCh38
rs1005500714 Chr.3: 183697700-183697700 on Build GRCh38
rs1029841795 Chr.3: 183697724-183697724 on Build GRCh38
rs1018215712 Chr.3: 183697591-183697591 on Build GRCh38
rs1001473579 Chr.3: 183697599-183697599 on Build GRCh38
rs1017002599 Chr.3: 183697701-183697701 on Build GRCh38
rs981500361 Chr.3: 183698012-183698012 on Build GRCh38
rs913011433 Chr.3: 183697775-183697784 on Build GRCh38
rs891194275 Chr.3: 183697930-183697930 on Build GRCh38
rs1045863535 Chr.3: 183697782-183697782 on Build GRCh38
rs1036201227 Chr.3: 183697877-183697877 on Build GRCh38
rs897118125 Chr.3: 183697807-183697807 on Build GRCh38
rs533029176 Chr.3: 183697838-183697838 on Build GRCh38
rs1035468573 Chr.3: 183698017-183698017 on Build GRCh38
rs1007057599 Chr.3: 183697835-183697835 on Build GRCh38
rs527326604 Chr.3: 183697988-183697988 on Build GRCh38
rs898413284 Chr.3: 183697600-183697600 on Build GRCh38
rs967414388 Chr.3: 183697845-183697845 on Build GRCh38
rs1019976360 Chr.3: 183697844-183697844 on Build GRCh38
rs990377731 Chr.3: 183697842-183697842 on Build GRCh38
rs971992275 Chr.3: 183698041-183698041 on Build GRCh38
rs897154964 Chr.3: 183697892-183697892 on Build GRCh38
rs1030118732 Chr.3: 183697831-183697831 on Build GRCh38
rs1007088690 Chr.3: 183697951-183697951 on Build GRCh38
rs928496595 Chr.3: 183697784-183697784 on Build GRCh38
rs937131479 Chr.3: 183697785-183697785 on Build GRCh38
rs528524122 Chr.3: 183697771-183697782 on Build GRCh38

View Full Product Details