Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 185918234-185918728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRA2B
Entrez Gene ID
6434
Gene Name
transformer 2 beta homolog (Drosophila)
Gene Aliases
Htra2-beta, PPP1R156, SFRS10, SRFS10, TRA2-BETA, TRAN2B
UniGene
Hs.533122
Gene Chromosome Location
Chr.3: 185914568-185938136 on Build GRCh38
Transcripts
NM_001243879.1 NM_004593.2 XM_005247703.1 XM_011513072.2 XM_017007033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6903116 c.833C>T p.S278F Chr.3: 185918388-185918388 on Build GRCh38
4115916 c.812G>A p.R271H Chr.3: 185918409-185918409 on Build GRCh38
5795790 c.802T>A p.Y268N Chr.3: 185918419-185918419 on Build GRCh38
3332119 c.799C>T p.P267S Chr.3: 185918422-185918422 on Build GRCh38
420095 c.851C>T p.S284L Chr.3: 185918370-185918370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767310132 Chr.3: 185918682-185918682 on Build GRCh38
rs962993528 Chr.3: 185918329-185918329 on Build GRCh38
rs867013951 Chr.3: 185918701-185918701 on Build GRCh38
rs183453662 Chr.3: 185918699-185918699 on Build GRCh38
rs1034575643 Chr.3: 185918529-185918529 on Build GRCh38
rs939393073 Chr.3: 185918555-185918555 on Build GRCh38
rs1014809540 Chr.3: 185918269-185918269 on Build GRCh38
rs1004724761 Chr.3: 185918267-185918267 on Build GRCh38
rs778329811 Chr.3: 185918463-185918463 on Build GRCh38
rs748788348 Chr.3: 185918338-185918338 on Build GRCh38
rs778489418 Chr.3: 185918343-185918343 on Build GRCh38
rs768451429 Chr.3: 185918556-185918556 on Build GRCh38
rs770452393 Chr.3: 185918342-185918342 on Build GRCh38
rs1021001348 Chr.3: 185918461-185918470 on Build GRCh38
rs769523428 Chr.3: 185918433-185918433 on Build GRCh38
rs540961991 Chr.3: 185918422-185918422 on Build GRCh38
rs762997606 Chr.3: 185918393-185918393 on Build GRCh38
rs528429137 Chr.3: 185918371-185918371 on Build GRCh38
rs538116357 Chr.3: 185918679-185918679 on Build GRCh38
rs569073526 Chr.3: 185918564-185918564 on Build GRCh38
rs781633335 Chr.3: 185918318-185918318 on Build GRCh38
rs919806321 Chr.3: 185918689-185918689 on Build GRCh38
rs144976860 Chr.3: 185918657-185918657 on Build GRCh38
rs747200048 Chr.3: 185918694-185918695 on Build GRCh38
rs544772959 Chr.3: 185918257-185918257 on Build GRCh38
rs777371640 Chr.3: 185918326-185918326 on Build GRCh38
rs758000942 Chr.3: 185918475-185918475 on Build GRCh38
rs376402396 Chr.3: 185918497-185918497 on Build GRCh38
rs556410119 Chr.3: 185918693-185918693 on Build GRCh38
rs1020496440 Chr.3: 185918601-185918601 on Build GRCh38
rs774063594 Chr.3: 185918584-185918584 on Build GRCh38
rs780774604 Chr.3: 185918319-185918319 on Build GRCh38
rs577997521 Chr.3: 185918607-185918607 on Build GRCh38
rs372774628 Chr.3: 185918473-185918473 on Build GRCh38
rs895625419 Chr.3: 185918576-185918576 on Build GRCh38
rs919858593 Chr.3: 185918448-185918448 on Build GRCh38
rs200996987 Chr.3: 185918398-185918398 on Build GRCh38
rs1002289464 Chr.3: 185918504-185918504 on Build GRCh38
rs149937016 Chr.3: 185918490-185918490 on Build GRCh38
rs886826382 Chr.3: 185918481-185918481 on Build GRCh38
rs756469772 Chr.3: 185918447-185918447 on Build GRCh38
rs746850295 Chr.3: 185918486-185918486 on Build GRCh38
rs912601588 Chr.3: 185918380-185918380 on Build GRCh38
rs987196009 Chr.3: 185918309-185918309 on Build GRCh38
rs766472567 Chr.3: 185918412-185918412 on Build GRCh38
rs931240595 Chr.3: 185918690-185918690 on Build GRCh38
rs967495769 Chr.3: 185918258-185918258 on Build GRCh38
rs564482611 Chr.3: 185918316-185918316 on Build GRCh38
rs756638794 Chr.3: 185918320-185918320 on Build GRCh38
rs532871203 Chr.3: 185918590-185918590 on Build GRCh38
rs554800138 Chr.3: 185918354-185918355 on Build GRCh38
rs756784491 Chr.3: 185918460-185918460 on Build GRCh38
rs374206338 Chr.3: 185918700-185918701 on Build GRCh38
rs1029071698 Chr.3: 185918609-185918609 on Build GRCh38
rs767268821 Chr.3: 185918511-185918511 on Build GRCh38
rs375817644 Chr.3: 185918482-185918482 on Build GRCh38
rs376085296 Chr.3: 185918442-185918442 on Build GRCh38
rs759772070 Chr.3: 185918432-185918432 on Build GRCh38
rs768440776 Chr.3: 185918388-185918388 on Build GRCh38
rs755735807 Chr.3: 185918695-185918695 on Build GRCh38
rs751971160 Chr.3: 185918386-185918386 on Build GRCh38
rs776604450 Chr.3: 185918391-185918391 on Build GRCh38
rs57681380 Chr.3: 185918603-185918603 on Build GRCh38
rs745515821 Chr.3: 185918353-185918353 on Build GRCh38
rs771860123 Chr.3: 185918361-185918361 on Build GRCh38
rs953075036 Chr.3: 185918645-185918645 on Build GRCh38
rs779073999 Chr.3: 185918344-185918345 on Build GRCh38
rs1046486266 Chr.3: 185918483-185918483 on Build GRCh38
rs567429552 Chr.3: 185918666-185918666 on Build GRCh38
rs764557638 Chr.3: 185918457-185918457 on Build GRCh38
rs373210628 Chr.3: 185918341-185918341 on Build GRCh38
rs866402769 Chr.3: 185918451-185918451 on Build GRCh38
rs551012624 Chr.3: 185918599-185918599 on Build GRCh38
rs745343258 Chr.3: 185918369-185918369 on Build GRCh38
rs564623776 Chr.3: 185918409-185918409 on Build GRCh38
rs369335055 Chr.3: 185918458-185918458 on Build GRCh38
rs1045465651 Chr.3: 185918494-185918495 on Build GRCh38

View Full Product Details