Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 187076415-187076940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST6GAL1
Entrez Gene ID
6480
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 1
Gene Aliases
SIAT1, ST6GalI, ST6N
UniGene
Hs.207459
Gene Chromosome Location
Chr.3: 186930343-187078553 on Build GRCh38
Transcripts
NM_003032.2 NM_173216.2 NM_173217.2 XM_005247719.1 XM_005247720.1 XM_006713734.1 XM_011513085.1 XM_011513086.1 XM_017007065.1 XM_017007066.1 XM_017007067.1 XM_017007068.1

SNP Detail

SNP ID Chromosome Location
rs191256248 Chr.3: 187076541-187076541 on Build GRCh38
rs569027241 Chr.3: 187076614-187076617 on Build GRCh38
rs115115233 Chr.3: 187076487-187076487 on Build GRCh38
rs528490763 Chr.3: 187076578-187076578 on Build GRCh38
rs9819341 Chr.3: 187076722-187076722 on Build GRCh38
rs908504708 Chr.3: 187076850-187076850 on Build GRCh38
rs532858990 Chr.3: 187076744-187076744 on Build GRCh38
rs1038459428 Chr.3: 187076658-187076658 on Build GRCh38
rs763816902 Chr.3: 187076813-187076813 on Build GRCh38
rs180719556 Chr.3: 187076601-187076601 on Build GRCh38
rs368517219 Chr.3: 187076690-187076690 on Build GRCh38
rs11554721 Chr.3: 187076492-187076492 on Build GRCh38
rs957533825 Chr.3: 187076551-187076551 on Build GRCh38
rs957525701 Chr.3: 187076776-187076776 on Build GRCh38
rs751174576 Chr.3: 187076831-187076831 on Build GRCh38
rs994677633 Chr.3: 187076475-187076475 on Build GRCh38
rs566302221 Chr.3: 187076910-187076910 on Build GRCh38
rs967504458 Chr.3: 187076527-187076527 on Build GRCh38
rs961433059 Chr.3: 187076855-187076855 on Build GRCh38
rs894428168 Chr.3: 187076686-187076686 on Build GRCh38
rs936433369 Chr.3: 187076552-187076552 on Build GRCh38
rs1013896885 Chr.3: 187076489-187076489 on Build GRCh38
rs955583688 Chr.3: 187076830-187076830 on Build GRCh38
rs955047743 Chr.3: 187076488-187076488 on Build GRCh38
rs1033558071 Chr.3: 187076549-187076549 on Build GRCh38
rs562034066 Chr.3: 187076650-187076650 on Build GRCh38
rs899006421 Chr.3: 187076463-187076463 on Build GRCh38
rs1034260624 Chr.3: 187076483-187076483 on Build GRCh38
rs762553541 Chr.3: 187076757-187076757 on Build GRCh38
rs906329429 Chr.3: 187076648-187076648 on Build GRCh38
rs147266974 Chr.3: 187076684-187076684 on Build GRCh38
rs1042643 Chr.3: 187076614-187076614 on Build GRCh38
rs1024047548 Chr.3: 187076513-187076513 on Build GRCh38
rs886513929 Chr.3: 187076715-187076715 on Build GRCh38
rs933961854 Chr.3: 187076600-187076600 on Build GRCh38
rs930517025 Chr.3: 187076668-187076668 on Build GRCh38
rs974558947 Chr.3: 187076856-187076856 on Build GRCh38
rs1028508016 Chr.3: 187076818-187076818 on Build GRCh38
rs1032797307 Chr.3: 187076763-187076763 on Build GRCh38
rs761118052 Chr.3: 187076625-187076625 on Build GRCh38
rs551456432 Chr.3: 187076886-187076886 on Build GRCh38
rs975653786 Chr.3: 187076576-187076576 on Build GRCh38
rs1037664468 Chr.3: 187076741-187076741 on Build GRCh38
rs996345032 Chr.3: 187076781-187076781 on Build GRCh38
rs1045789004 Chr.3: 187076709-187076709 on Build GRCh38
rs940060631 Chr.3: 187076639-187076639 on Build GRCh38
rs898993538 Chr.3: 187076665-187076665 on Build GRCh38

View Full Product Details