Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 187725200-187725721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL6
Entrez Gene ID
604
Gene Name
B-cell CLL/lymphoma 6
Gene Aliases
BCL5, BCL6A, LAZ3, ZBTB27, ZNF51
UniGene
Hs.478588
Gene Chromosome Location
Chr.3: 187721377-187745725 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Gene Symbol
LOC100131635
Entrez Gene ID
100131635
Gene Name
hCG1645011-like
Gene Aliases
-
UniGene
Hs.434448
Gene Chromosome Location
Chr.3: 187702366-187732415 on Build GRCh38
Transcripts
NM_001130845.1 NM_001134738.1 NM_001706.4 XM_005247694.3 XM_011513062.2 NR_034062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1042447 c.1709-1G>A p.? Chr.3: 187725630-187725630 on Build GRCh38
6610727 c.1716A>G p.K572K Chr.3: 187725622-187725622 on Build GRCh38
1421597 c.1812C>T p.C604C Chr.3: 187725526-187725526 on Build GRCh38
6999804 c.1822G>A p.G608R Chr.3: 187725516-187725516 on Build GRCh38
5817657 c.1812C>G p.C604W Chr.3: 187725526-187725526 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199598549 Chr.3: 187725459-187725459 on Build GRCh38
rs538391218 Chr.3: 187725517-187725517 on Build GRCh38
rs573828872 Chr.3: 187725295-187725295 on Build GRCh38
rs184983595 Chr.3: 187725480-187725480 on Build GRCh38
rs755666072 Chr.3: 187725615-187725615 on Build GRCh38
rs986864705 Chr.3: 187725661-187725661 on Build GRCh38
rs544079523 Chr.3: 187725442-187725442 on Build GRCh38
rs374261259 Chr.3: 187725379-187725379 on Build GRCh38
rs955464200 Chr.3: 187725632-187725632 on Build GRCh38
rs778653664 Chr.3: 187725462-187725465 on Build GRCh38
rs748646860 Chr.3: 187725480-187725481 on Build GRCh38
rs201068277 Chr.3: 187725462-187725462 on Build GRCh38
rs752044631 Chr.3: 187725558-187725558 on Build GRCh38
rs777469418 Chr.3: 187725621-187725621 on Build GRCh38
rs567242744 Chr.3: 187725438-187725454 on Build GRCh38
rs994402271 Chr.3: 187725393-187725393 on Build GRCh38
rs769071983 Chr.3: 187725472-187725488 on Build GRCh38
rs201366633 Chr.3: 187725487-187725488 on Build GRCh38
rs745750475 Chr.3: 187725669-187725669 on Build GRCh38
rs541880007 Chr.3: 187725415-187725415 on Build GRCh38
rs561641205 Chr.3: 187725421-187725421 on Build GRCh38
rs570767731 Chr.3: 187725241-187725241 on Build GRCh38
rs772163908 Chr.3: 187725670-187725670 on Build GRCh38
rs577922665 Chr.3: 187725377-187725377 on Build GRCh38
rs966599533 Chr.3: 187725418-187725418 on Build GRCh38
rs377286077 Chr.3: 187725298-187725298 on Build GRCh38
rs112603207 Chr.3: 187725453-187725453 on Build GRCh38
rs569264944 Chr.3: 187725491-187725491 on Build GRCh38
rs369569342 Chr.3: 187725334-187725334 on Build GRCh38
rs999794416 Chr.3: 187725342-187725342 on Build GRCh38
rs767829359 Chr.3: 187725583-187725583 on Build GRCh38
rs9759194 Chr.3: 187725315-187725315 on Build GRCh38
rs1013990176 Chr.3: 187725692-187725692 on Build GRCh38
rs984220954 Chr.3: 187725249-187725249 on Build GRCh38
rs752300498 Chr.3: 187725470-187725470 on Build GRCh38
rs73886539 Chr.3: 187725479-187725479 on Build GRCh38
rs762173942 Chr.3: 187725454-187725454 on Build GRCh38
rs911279019 Chr.3: 187725262-187725262 on Build GRCh38
rs750544229 Chr.3: 187725455-187725455 on Build GRCh38
rs1032264970 Chr.3: 187725580-187725580 on Build GRCh38
rs753499318 Chr.3: 187725622-187725622 on Build GRCh38
rs753064923 Chr.3: 187725604-187725604 on Build GRCh38
rs777082850 Chr.3: 187725475-187725475 on Build GRCh38
rs181300648 Chr.3: 187725384-187725384 on Build GRCh38
rs1042221684 Chr.3: 187725516-187725516 on Build GRCh38
rs778615302 Chr.3: 187725654-187725654 on Build GRCh38
rs571551739 Chr.3: 187725690-187725690 on Build GRCh38
rs763086743 Chr.3: 187725522-187725522 on Build GRCh38
rs1032985614 Chr.3: 187725431-187725431 on Build GRCh38
rs781017496 Chr.3: 187725453-187725454 on Build GRCh38
rs386669300 Chr.3: 187725462-187725479 on Build GRCh38
rs528353546 Chr.3: 187725574-187725574 on Build GRCh38
rs535937461 Chr.3: 187725310-187725310 on Build GRCh38
rs922428901 Chr.3: 187725550-187725550 on Build GRCh38
rs61732778 Chr.3: 187725526-187725526 on Build GRCh38
rs560485265 Chr.3: 187725476-187725476 on Build GRCh38
rs781588369 Chr.3: 187725463-187725463 on Build GRCh38
rs9755700 Chr.3: 187725425-187725425 on Build GRCh38
rs984103158 Chr.3: 187725429-187725429 on Build GRCh38
rs569406058 Chr.3: 187725316-187725316 on Build GRCh38
rs557073008 Chr.3: 187725279-187725295 on Build GRCh38
rs542434198 Chr.3: 187725309-187725309 on Build GRCh38
rs759391036 Chr.3: 187725451-187725451 on Build GRCh38
rs370028442 Chr.3: 187725504-187725504 on Build GRCh38
rs746831293 Chr.3: 187725676-187725676 on Build GRCh38
rs894191454 Chr.3: 187725369-187725369 on Build GRCh38
rs768391578 Chr.3: 187725678-187725678 on Build GRCh38
rs558391370 Chr.3: 187725375-187725375 on Build GRCh38
rs996848361 Chr.3: 187725585-187725585 on Build GRCh38
rs372794459 Chr.3: 187725672-187725672 on Build GRCh38
rs1020828615 Chr.3: 187725397-187725397 on Build GRCh38
rs898183873 Chr.3: 187725380-187725380 on Build GRCh38
rs896265439 Chr.3: 187725385-187725385 on Build GRCh38
rs566911943 Chr.3: 187725303-187725303 on Build GRCh38
rs773103820 Chr.3: 187725519-187725519 on Build GRCh38
rs139370058 Chr.3: 187725362-187725362 on Build GRCh38
rs979364054 Chr.3: 187725422-187725422 on Build GRCh38
rs756758766 Chr.3: 187725641-187725641 on Build GRCh38
rs879240244 Chr.3: 187725335-187725335 on Build GRCh38
rs760062751 Chr.3: 187725565-187725565 on Build GRCh38
rs1002331797 Chr.3: 187725382-187725382 on Build GRCh38
rs776915341 Chr.3: 187725479-187725480 on Build GRCh38
rs756120631 Chr.3: 187725281-187725282 on Build GRCh38
rs887351118 Chr.3: 187725363-187725363 on Build GRCh38
rs747298443 Chr.3: 187725467-187725479 on Build GRCh38
rs572531548 Chr.3: 187725396-187725396 on Build GRCh38
rs771969572 Chr.3: 187725488-187725488 on Build GRCh38
rs796133356 Chr.3: 187725453-187725454 on Build GRCh38
rs925967072 Chr.3: 187725299-187725299 on Build GRCh38
rs775446627 Chr.3: 187725496-187725496 on Build GRCh38
rs564832837 Chr.3: 187725388-187725388 on Build GRCh38
rs188599501 Chr.3: 187725281-187725281 on Build GRCh38
rs376846160 Chr.3: 187725330-187725330 on Build GRCh38

View Full Product Details