Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 190307277-190307790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN1
Entrez Gene ID
9076
Gene Name
claudin 1
Gene Aliases
CLD1, ILVASC, SEMP1
UniGene
Hs.439060
Gene Chromosome Location
Chr.3: 190305701-190322446 on Build GRCh38
Transcripts
NM_021101.4

SNP Detail

SNP ID Chromosome Location
rs1047780804 Chr.3: 190307402-190307402 on Build GRCh38
rs73056495 Chr.3: 190307691-190307691 on Build GRCh38
rs991677972 Chr.3: 190307659-190307659 on Build GRCh38
rs775497434 Chr.3: 190307661-190307661 on Build GRCh38
rs749814941 Chr.3: 190307563-190307563 on Build GRCh38
rs903430312 Chr.3: 190307537-190307537 on Build GRCh38
rs1040165029 Chr.3: 190307481-190307481 on Build GRCh38
rs1000481420 Chr.3: 190307538-190307538 on Build GRCh38
rs573986269 Chr.3: 190307658-190307658 on Build GRCh38
rs547587328 Chr.3: 190307411-190307411 on Build GRCh38
rs964944430 Chr.3: 190307364-190307364 on Build GRCh38
rs964805596 Chr.3: 190307712-190307712 on Build GRCh38
rs1000935262 Chr.3: 190307681-190307681 on Build GRCh38
rs80187925 Chr.3: 190307400-190307400 on Build GRCh38
rs971758486 Chr.3: 190307369-190307369 on Build GRCh38
rs531266099 Chr.3: 190307513-190307513 on Build GRCh38
rs546735284 Chr.3: 190307624-190307624 on Build GRCh38
rs146819132 Chr.3: 190307648-190307648 on Build GRCh38
rs537588696 Chr.3: 190307463-190307463 on Build GRCh38
rs942117392 Chr.3: 190307431-190307431 on Build GRCh38
rs949716104 Chr.3: 190307354-190307354 on Build GRCh38
rs562359831 Chr.3: 190307450-190307450 on Build GRCh38
rs780870918 Chr.3: 190307328-190307328 on Build GRCh38
rs976639063 Chr.3: 190307378-190307378 on Build GRCh38
rs901266659 Chr.3: 190307508-190307508 on Build GRCh38
rs567992824 Chr.3: 190307730-190307730 on Build GRCh38
rs1048867859 Chr.3: 190307406-190307406 on Build GRCh38
rs917017288 Chr.3: 190307331-190307331 on Build GRCh38
rs769145693 Chr.3: 190307627-190307627 on Build GRCh38
rs1024434247 Chr.3: 190307668-190307668 on Build GRCh38
rs865960266 Chr.3: 190307543-190307543 on Build GRCh38
rs958423594 Chr.3: 190307310-190307310 on Build GRCh38
rs1033278038 Chr.3: 190307586-190307586 on Build GRCh38
rs533586709 Chr.3: 190307615-190307615 on Build GRCh38
rs780769636 Chr.3: 190307545-190307545 on Build GRCh38
rs760627434 Chr.3: 190307680-190307680 on Build GRCh38
rs1056196455 Chr.3: 190307587-190307587 on Build GRCh38
rs923548186 Chr.3: 190307382-190307382 on Build GRCh38
rs535449802 Chr.3: 190307680-190307680 on Build GRCh38
rs976262715 Chr.3: 190307714-190307714 on Build GRCh38
rs998556618 Chr.3: 190307517-190307517 on Build GRCh38
rs991211876 Chr.3: 190307317-190307317 on Build GRCh38
rs909173639 Chr.3: 190307417-190307417 on Build GRCh38
rs896425596 Chr.3: 190307647-190307647 on Build GRCh38
rs888972581 Chr.3: 190307430-190307430 on Build GRCh38
rs557383916 Chr.3: 190307535-190307535 on Build GRCh38
rs755668498 Chr.3: 190307544-190307544 on Build GRCh38
rs528044643 Chr.3: 190307329-190307329 on Build GRCh38
rs191911854 Chr.3: 190307515-190307515 on Build GRCh38
rs923412514 Chr.3: 190307715-190307715 on Build GRCh38
rs930595627 Chr.3: 190307401-190307401 on Build GRCh38

View Full Product Details